Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › MC19215
          Assay Name: hsa-miR-101-2-5p
          miRBase Accession Number: MI0000739
          miRBase Version: v22.1
          Chromosome Location: Chr.9: 4850297 - 4850375 [+] on Build GRCh38
          Mature miRNA Sequence: UCGGUUAUCAUGGUACCGAUGC
          Species: Human, Rat
          Product Type: mirVana® miRNA mimic
          Assay ID MC19215

          Purification
          Size
          Availability Made To Order
          Catalog # 4464066
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000046, mir-101

          Mature miRNA Sequence:

          UCGGUUAUCAUGGUACCGAUGC

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-101-2-5p MIMAT0037312
          Rat rno-miR-101b-5p MIMAT0017045

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-101-2
          Stem-loop Accession # MI0000739
          Stem-loop Sequence
          ACUGUCCUUUUUCGGUUAUCAUGGUACCGAUGCUGUAUAUCUGAAAGGUACAGUACUGUGAUAACUGAAGAAUGGUGGU
          Chromosome Location Chr. 9 - 4850297 - 4850375 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0037312
          miRBase ID: hsa-miR-101-2-5p
          Mature miRNA Sequence: UCGGUUAUCAUGGUACCGAUGC
          Chromosome Location: Chr. 9 - 4850297 - 4850375 [+] on Build GRCh38

          Rat

          Stem-loop ID rno-mir-101b
          Stem-loop Accession # MI0000648
          Stem-loop Sequence
          AUCUGAGACUGAACUGUCCUUUUUCGGUUAUCAUGGUACCGAUGCUGUAGAUCUGAAAGGUACAGUACUGUGAUAGCUGAAGAAUGGUGGUGCCAUC
          Chromosome Location Chr. 1 - 247263723 - 247263819 [+] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0017045
          miRBase ID: rno-miR-101b-5p
          Mature miRNA Sequence: UCGGUUAUCAUGGUACCGAUGC
          Chromosome Location: Chr. 1 - 247263723 - 247263819 [+] on Build Rnor_6.0


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Rn03465939_pri
          TaqMan™ Pri-miRNA Assay : Hs03303387_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM19215
          Ambion® Pre-miR™ miRNA Precursor : PM19215
          TaqMan™ MicroRNA Assay : 462381_mat
          mirVana® miRNA inhibitor : MH19215
          TaqMan™ Advanced miRNA Assay : rno480890_mir
          TaqMan™ Advanced miRNA Assay : 480890_mir


          miRBase Alias:

          Rat: rno-miR-101b* (v18)

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline