Search
Search

| Assay Name: | hsa-miR-219b-5p |
|---|---|
| miRBase Accession Number: | MI0017299 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | AGAUGUCCAGCCACAAUUCUCG |
| Species: | Human, Mouse, Rat |
| Product Type: | Ambion® Anti-miR™ miRNA Inhibitor |
Gene Family ID: | MIPF0000044, mir-219 |
Mature miRNA Sequence: | AGAUGUCCAGCCACAAUUCUCG |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-219b-5p | MIMAT0019747 |
| Mouse | mmu-miR-219b-5p | MIMAT0029806 |
| Rat | rno-miR-219b | MIMAT0017882 |