Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03303392_pri
          Assay Name:
          hsa-mir-219a-2
          Stem-loop ID:
          hsa-mir-219a-2 hsa-mir-219b
          Mapped to miRBase Version:
          v22.1
          Chromosome Location:
          Chr.9: 128392618 - 128392714 [-] on Build GRCh38
          Stem-loop Location:
          Chr.9: 128392618 - 128392714 [-] on Build GRCh38 Chr.9: 128392621 - 128392708 [+] on Build GRCh38
          Mature miRNA ID:
          hsa-miR-219a-5p hsa-miR-219a-2-3p hsa-miR-219b-3p hsa-miR-219b-5p
          Species:
          Human
          Product Type:
          TaqMan™ Pri-miRNA Assay
          Assay ID Hs03303392_pri
          Size
          Availability Made To Order
          Catalog # 4427012
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Amplicon Length:

          101

          Chromosome Location:

          Chr. 9 - 128392317 - 128392317 [-] on 128392317 Build GRCh38

          Species:

          Human

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-219a-5p MIMAT0000276
          Human hsa-miR-219a-2-3p MIMAT0004675
          Human hsa-miR-219b-3p MIMAT0019748
          Human hsa-miR-219b-5p MIMAT0019747

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-219a-2
          Stem-loop Accession # MI0000740
          Stem-loop Sequence
          ACUCAGGGGCUUCGCCACUGAUUGUCCAAACGCAAUUCUUGUACGAGUCUGCGGCCAACCGAGAAUUGUGGCUGGACAUCUGUGGCUGAGCUCCGGG
          Chromosome Location Chr. 9 - 128392618 - 128392714 [-] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0000276
          miRBase ID: hsa-miR-219a-5p
          Mature miRNA Sequence: UGAUUGUCCAAACGCAAUUCU
          Chromosome Location: Chr. 9 - 128392618 - 128392714 [-] on Build GRCh38

          miRBase Accession #: MIMAT0004675
          miRBase ID: hsa-miR-219a-2-3p
          Mature miRNA Sequence: AGAAUUGUGGCUGGACAUCUGU
          Chromosome Location: Chr. 9 - 128392618 - 128392714 [-] on Build GRCh38

          Gene Family ID MIPF0000044, mir-219
          Distance to Amplicon 229 bases
          Stem-loop ID hsa-mir-219b
          Stem-loop Accession # MI0017299
          Stem-loop Sequence
          GGAGCUCAGCCACAGAUGUCCAGCCACAAUUCUCGGUUGGCCGCAGACUCGUACAAGAAUUGCGUUUGGACAAUCAGUGGCGAAGCCC
          Chromosome Location Chr. 9 - 128392621 - 128392708 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0019748
          miRBase ID: hsa-miR-219b-3p
          Mature miRNA Sequence: AGAAUUGCGUUUGGACAAUCAGU
          Chromosome Location: Chr. 9 - 128392621 - 128392708 [+] on Build GRCh38

          miRBase Accession #: MIMAT0019747
          miRBase ID: hsa-miR-219b-5p
          Mature miRNA Sequence: AGAUGUCCAGCCACAAUUCUCG
          Chromosome Location: Chr. 9 - 128392621 - 128392708 [+] on Build GRCh38

          Gene Family ID MIPF0000044, mir-219
          Distance to Amplicon 232 bases

          Back To Top

          More Information




          Related Products

          Ambion® Anti-miR™ miRNA Inhibitor : AM10664
          Ambion® Pre-miR™ miRNA Precursor : PM10664
          TaqMan™ MicroRNA Assay : 000522
          mirVana® miRNA inhibitor : MH10664
          mirVana® miRNA mimic : MC10664
          TaqMan™ Advanced miRNA Assay : 477980_mir
          TaqMan™ Advanced miRNA Assay : mmu482708_mir
          TaqMan™ Advanced miRNA Assay : rno481348_mir
          Ambion® Anti-miR™ miRNA Inhibitor : AM11150
          Ambion® Pre-miR™ miRNA Precursor : PM11150
          TaqMan™ MicroRNA Assay : 002390
          mirVana® miRNA inhibitor : MH11150
          mirVana® miRNA mimic : MC11150
          TaqMan™ Advanced miRNA Assay : rno481002_mir
          TaqMan™ Advanced miRNA Assay : mmu481002_mir
          TaqMan™ Advanced miRNA Assay : 477979_mir
          Ambion® Anti-miR™ miRNA Inhibitor : AM21422
          Ambion® Pre-miR™ miRNA Precursor : PM21422
          TaqMan™ MicroRNA Assay : 462260_mat
          mirVana® miRNA inhibitor : MH21422
          mirVana® miRNA mimic : MC21422
          TaqMan™ Advanced miRNA Assay : 478776_mir
          TaqMan™ Advanced miRNA Assay : mmu481656_mir
          Ambion® Anti-miR™ miRNA Inhibitor : AM20193
          Ambion® Pre-miR™ miRNA Precursor : PM20193
          TaqMan™ MicroRNA Assay : 462668_mat
          mirVana® miRNA inhibitor : MH20193
          mirVana® miRNA mimic : MC20193
          TaqMan™ Advanced miRNA Assay : 478777_mir
          TaqMan™ Advanced miRNA Assay : mmu481505_mir
          TaqMan™ Advanced miRNA Assay : rno481003_mir

          Back To Top

          Related Products

          • High Capacity RNA-to-cDNA Kit
          • SuperScript® IV VILO™ Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Fast Advanced Master Mix
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-4lm45:80/100.66.75.98:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0