Search
Search

| Assay Name: | mmu-miR-219a-5p |
|---|---|
| Stem-loop Accession Number: | MI0000296 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UGAUUGUCCAAACGCAAUUCU |
| Species: | Human, Mouse, Rat, Drosophila melanogaster, Branchiostoma belcheri, Branchiostoma floridae, Capitella teleta, Chicken, Dog, Goat, Gorilla gorilla, Horse, Monodelphis domestica, Pan troglodytes, Pongo pygmaeus, Pteropus alecto, Rhesus monkey, Tribolium castaneum, Xenopus tropicalis |
| Product Type: | TaqMan™ Advanced miRNA Assay |
Gene Family ID: | MIPF0000044, mir-219 |
Mature miRNA Sequence: | UGAUUGUCCAAACGCAAUUCU |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-219a-5p | MIMAT0000276 |
| Mouse | mmu-miR-219a-5p | MIMAT0000664 |
| Rat | rno-miR-219a-5p | MIMAT0000889 |
| Drosophila melanogaster | dme-miR-219-5p | MIMAT0000335 |
| Branchiostoma belcheri | bbe-miR-219-5p | MIMAT0031650 |
| Branchiostoma floridae | bfl-miR-219 | MIMAT0010012 |
| View More Rows | ||
| Capitella teleta | cte-miR-219 | MIMAT0009556 |
| Chicken | gga-miR-219a | MIMAT0001170 |
| Dog | cfa-miR-219-5p | MIMAT0006611 |
| Goat | chi-miR-219 | MIMAT0036067 |
| Gorilla gorilla | ggo-miR-219 | MIMAT0002576 |
| Horse | eca-miR-219-5p | MIMAT0013099 |
| Monodelphis domestica | mdo-miR-219-5p | MIMAT0004134 |
| Pan troglodytes | ptr-miR-219-5p | MIMAT0009193 |
| Pongo pygmaeus | ppy-miR-219 | MIMAT0002577 |
| Pongo pygmaeus | ppy-miR-219-5p | MIMAT0015804 |
| Pteropus alecto | pal-miR-219a-5p | MIMAT0039927 |
| Rhesus monkey | mml-miR-219 | MIMAT0002575 |
| Rhesus monkey | mml-miR-219-5p | MIMAT0006245 |
| Tribolium castaneum | tca-miR-219-5p | MIMAT0008377 |
| Xenopus tropicalis | xtr-miR-219 | MIMAT0003632 |