Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › AM10664
          Assay Name: hsa-miR-219a-5p
          miRBase Accession Number: MI0000296
          miRBase Version: v22.1
          Chromosome Location: Chr.6: 33207835 - 33207944 [+] on Build GRCh38
          Mature miRNA Sequence: UGAUUGUCCAAACGCAAUUCU
          Species: Human, Mouse, Rat, Drosophila melanogaster, Branchiostoma belcheri, Branchiostoma floridae, Capitella teleta, Chicken, Dog, Goat, Gorilla gorilla, Horse, Monodelphis domestica, Pan troglodytes, Pongo pygmaeus, Pteropus alecto, Rhesus monkey, Tribolium castaneum, Xenopus tropicalis
          Product Type: Ambion® Anti-miR™ miRNA Inhibitor
          Assay ID AM10664
          Size
          Availability Made To Order
          Catalog # AM17000
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000044, mir-219

          Mature miRNA Sequence:

          UGAUUGUCCAAACGCAAUUCU

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-219a-5p MIMAT0000276
          Mouse mmu-miR-219a-5p MIMAT0000664
          Rat rno-miR-219a-5p MIMAT0000889
          Drosophila melanogaster dme-miR-219-5p MIMAT0000335
          Branchiostoma belcheri bbe-miR-219-5p MIMAT0031650
          Branchiostoma floridae bfl-miR-219 MIMAT0010012
          View More Rows
          Capitella teleta cte-miR-219 MIMAT0009556
          Chicken gga-miR-219a MIMAT0001170
          Dog cfa-miR-219-5p MIMAT0006611
          Goat chi-miR-219 MIMAT0036067
          Gorilla gorilla ggo-miR-219 MIMAT0002576
          Horse eca-miR-219-5p MIMAT0013099
          Monodelphis domestica mdo-miR-219-5p MIMAT0004134
          Pan troglodytes ptr-miR-219-5p MIMAT0009193
          Pongo pygmaeus ppy-miR-219 MIMAT0002577
          Pongo pygmaeus ppy-miR-219-5p MIMAT0015804
          Pteropus alecto pal-miR-219a-5p MIMAT0039927
          Rhesus monkey mml-miR-219 MIMAT0002575
          Rhesus monkey mml-miR-219-5p MIMAT0006245
          Tribolium castaneum tca-miR-219-5p MIMAT0008377
          Xenopus tropicalis xtr-miR-219 MIMAT0003632

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-219a-1
          Stem-loop Accession # MI0000296
          Stem-loop Sequence
          CCGCCCCGGGCCGCGGCUCCUGAUUGUCCAAACGCAAUUCUCGAGUCUAUGGCUCCGGCCGAGAGUUGAGUCUGGACGUCCCGAGCCGCCGCCCCCAAACCUCGAGCGGG
          Chromosome Location Chr. 6 - 33207835 - 33207944 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0000276
          miRBase ID: hsa-miR-219a-5p
          Mature miRNA Sequence: UGAUUGUCCAAACGCAAUUCU
          Chromosome Location: Chr. 9 - 128392618 - 128392714 [-] on Build GRCh38

          Stem-loop ID hsa-mir-219a-2
          Stem-loop Accession # MI0000740
          Stem-loop Sequence
          ACUCAGGGGCUUCGCCACUGAUUGUCCAAACGCAAUUCUUGUACGAGUCUGCGGCCAACCGAGAAUUGUGGCUGGACAUCUGUGGCUGAGCUCCGGG
          Chromosome Location Chr. 9 - 128392618 - 128392714 [-] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0000276
          miRBase ID: hsa-miR-219a-5p
          Mature miRNA Sequence: UGAUUGUCCAAACGCAAUUCU
          Chromosome Location: Chr. 9 - 128392618 - 128392714 [-] on Build GRCh38

          Mouse

          Stem-loop ID mmu-mir-219a-1
          Stem-loop Accession # MI0000702
          Stem-loop Sequence
          CCGUCCCGGGCCGCGGCUCCUGAUUGUCCAAACGCAAUUCUCGAGUCUCUGGCUCCGGCCGAGAGUUGCGUCUGGACGUCCCGAGCCGCCGCCCCCAAACCUCGAGGGGG
          Chromosome Location Chr. 17 - 34024983 - 34025092 [-] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0000664
          miRBase ID: mmu-miR-219a-5p
          Mature miRNA Sequence: UGAUUGUCCAAACGCAAUUCU
          Chromosome Location: Chr. 2 - 29845631 - 29845727 [-] on Build GRCm38

          Stem-loop ID mmu-mir-219a-2
          Stem-loop Accession # MI0000741
          Stem-loop Sequence
          ACUCAGGGGCUUCGCCACUGAUUGUCCAAACGCAAUUCUUGUACGAGUCUGCGGCCAACCGAGAAUUGUGGCUGGACAUCUGUGGUUGAGCUCCGGG
          Chromosome Location Chr. 2 - 29845631 - 29845727 [-] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0000664
          miRBase ID: mmu-miR-219a-5p
          Mature miRNA Sequence: UGAUUGUCCAAACGCAAUUCU
          Chromosome Location: Chr. 2 - 29845631 - 29845727 [-] on Build GRCm38

          Rat

          Stem-loop ID rno-mir-219a-1
          Stem-loop Accession # MI0000959
          Stem-loop Sequence
          CUGUCCCGGGCCGCGGCUCCUGAUUGUCCAAACGCAAUUCUCGAGUCUCUGGCUCCGGCCGAGAGUUGCGUCUGGACGUCCCGAGCCGCCGCCCCCAAACCUCGAGGGGG
          Chromosome Location Chr. 20 - 3816158 - 3816267 [-] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0000889
          miRBase ID: rno-miR-219a-5p
          Mature miRNA Sequence: UGAUUGUCCAAACGCAAUUCU
          Chromosome Location: Chr. 20 - 3816158 - 3816267 [-] on Build Rnor_6.0

          Stem-loop ID rno-mir-219a-2
          Stem-loop Accession # MI0000960
          Stem-loop Sequence
          ACUCAGGGGCUUCACCACUGAUUGUCCAAACGCAAUUCUUGUACGAGUCUGCGGCCAACCGAGAAUUGUGGCUGGACAUCUGUGGUUGAGCUCCGG
          Chromosome Location Chr. 3 - 8407430 - 8407525 [-] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0000889
          miRBase ID: rno-miR-219a-5p
          Mature miRNA Sequence: UGAUUGUCCAAACGCAAUUCU
          Chromosome Location: Chr. 20 - 3816158 - 3816267 [-] on Build Rnor_6.0

          Drosophila melanogaster

          Stem-loop ID dme-mir-219
          Stem-loop Accession # MI0000358
          Stem-loop Sequence
          UAAUUCGAUUUUUAGCUAUGAUUGUCCAAACGCAAUUCUUGUUGAUAUUCAAUAUUCAAGGGUUGCGACUGGGCAUCGCGGCUCGAAAUAAGAAUACAAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0000335
          miRBase ID: dme-miR-219-5p
          Mature miRNA Sequence: UGAUUGUCCAAACGCAAUUCU
          Chromosome Location: NA

          Branchiostoma belcheri

          Stem-loop ID bbe-mir-219-1
          Stem-loop Accession # MI0026223
          Stem-loop Sequence
          CCAGUGCUGAUUGUCCAAACGCAAUUCUUGUUUGACCUUUGAACCAAGAACUGCGCUGGACAUCAGCAAUUGACGCUGCCUAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0031650
          miRBase ID: bbe-miR-219-5p
          Mature miRNA Sequence: UGAUUGUCCAAACGCAAUUCU
          Chromosome Location: NA

          Stem-loop ID bbe-mir-219-2
          Stem-loop Accession # MI0026224
          Stem-loop Sequence
          CCAGUGCUGAUUGUCCAAACGCAAUUCUUGUUUGACCUUUGAACCAAGAACUGCGCUGGACAUCAGCAAUUGACGCUGCCUAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0031650
          miRBase ID: bbe-miR-219-5p
          Mature miRNA Sequence: UGAUUGUCCAAACGCAAUUCU
          Chromosome Location: NA

          Branchiostoma floridae

          Stem-loop ID bfl-mir-219
          Stem-loop Accession # MI0010527
          Stem-loop Sequence
          CCAGUGCUGAUUGUCCAAACGCAAUUCUUGUUGACCUUUGAACCAAGAACUGCGCUGGACAUCAGCAAUUGACGCUGCCUAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0010012
          miRBase ID: bfl-miR-219
          Mature miRNA Sequence: UGAUUGUCCAAACGCAAUUCU
          Chromosome Location: NA

          Capitella teleta

          Stem-loop ID cte-mir-219
          Stem-loop Accession # MI0010099
          Stem-loop Sequence
          CAGACAGUCUUUUAGCAUUGAUUGUCCAAACGCAAUUCUUGUUCACUUCGUUCUCCAAGAACUGUGUUCGGACAUCAAUCCUAGCAAGGCAGUACACAACC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0009556
          miRBase ID: cte-miR-219
          Mature miRNA Sequence: UGAUUGUCCAAACGCAAUUCU
          Chromosome Location: NA

          Chicken

          Stem-loop ID gga-mir-219a
          Stem-loop Accession # MI0001246
          Stem-loop Sequence
          CUCUGCUCCUGAUUGUCCAAACGCAAUUCUUGUGCGCUGGAGCCGUACGAACCAAGAAUUGUGUCUGGACAUCUGUAGCAGAGAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0001170
          miRBase ID: gga-miR-219a
          Mature miRNA Sequence: UGAUUGUCCAAACGCAAUUCU
          Chromosome Location: NA

          Dog

          Stem-loop ID cfa-mir-219-1
          Stem-loop Accession # MI0008007
          Stem-loop Sequence
          UGAUUGUCCAAACGCAAUUCUCGAGUCUCUGGCUCUGGCCGAGAGUUGAGUCUGGACGUCCCG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006611
          miRBase ID: cfa-miR-219-5p
          Mature miRNA Sequence: UGAUUGUCCAAACGCAAUUCU
          Chromosome Location: NA

          Stem-loop ID cfa-mir-219-2
          Stem-loop Accession # MI0008150
          Stem-loop Sequence
          UGAUUGUCCAAACGCAAUUCUUGUACGAGUCUGCGGCCAACCGAGAAUUGUGGCUGGACAUCUGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006611
          miRBase ID: cfa-miR-219-5p
          Mature miRNA Sequence: UGAUUGUCCAAACGCAAUUCU
          Chromosome Location: NA

          Goat

          Stem-loop ID chi-mir-219
          Stem-loop Accession # MI0030695
          Stem-loop Sequence
          ACUCAGGGGCUUCGCCACUGAUUGUCCAAACGCAAUUCUUGUACGAGUCUGCGGCCAACCGAGAAUUGUGGCUGGACAUCUGUGGCUGAGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036067
          miRBase ID: chi-miR-219
          Mature miRNA Sequence: UGAUUGUCCAAACGCAAUUCU
          Chromosome Location: NA

          Gorilla gorilla

          Stem-loop ID ggo-mir-219
          Stem-loop Accession # MI0002885
          Stem-loop Sequence
          CCGCCCCGGGCCGCGGCUCCUGAUUGUCCAAACGCAAUUCUCGAGUCUAUGGCUCCGGCCGAGAGUUGAGUCUGGACGUCCCGAGCCGCCGCCCCCAAACCUCGAGCGGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002576
          miRBase ID: ggo-miR-219
          Mature miRNA Sequence: UGAUUGUCCAAACGCAAUUCU
          Chromosome Location: NA

          Horse

          Stem-loop ID eca-mir-219-1
          Stem-loop Accession # MI0012846
          Stem-loop Sequence
          GGGCCGCGGCUCUUGAUUGUCCAAACGCAAUUCUCGAGUCUCUGGCUCCGGCCGAGAGUUGAGUCUGGACGUCCCGAGCCGCCGCCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0013099
          miRBase ID: eca-miR-219-5p
          Mature miRNA Sequence: UGAUUGUCCAAACGCAAUUCU
          Chromosome Location: NA

          Stem-loop ID eca-mir-219-2
          Stem-loop Accession # MI0012923
          Stem-loop Sequence
          ACUCAGGGGCUUCGCCACUGAUUGUCCAAACGCAAUUCUUGUACGAGUCUGCGGCCAACCGAGAAUUGUGGCUGGACAUCUGUGGCUGAGCUCCGGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0013099
          miRBase ID: eca-miR-219-5p
          Mature miRNA Sequence: UGAUUGUCCAAACGCAAUUCU
          Chromosome Location: NA

          Monodelphis domestica

          Stem-loop ID mdo-mir-219-1
          Stem-loop Accession # MI0005323
          Stem-loop Sequence
          CAGGGGUUCCGCCGCUGAUUGUCCAAACGCAAUUCUUGUGCGAGUCUGCAGCCAACCGAGAAUUGUGGCUGGACAUCUGUGGCUGAGCUCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0004134
          miRBase ID: mdo-miR-219-5p
          Mature miRNA Sequence: UGAUUGUCCAAACGCAAUUCU
          Chromosome Location: NA

          Stem-loop ID mdo-mir-219-2
          Stem-loop Accession # MI0023783
          Stem-loop Sequence
          UGAUUGUCCAAACGCAAUUCUCGUGGCUCCGGCCCUCGAGAGUUGGGUCUGGACAUCCCG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0004134
          miRBase ID: mdo-miR-219-5p
          Mature miRNA Sequence: UGAUUGUCCAAACGCAAUUCU
          Chromosome Location: NA

          Pan troglodytes

          Stem-loop ID ptr-mir-219-1
          Stem-loop Accession # MI0008584
          Stem-loop Sequence
          CGCCCCGGGCCGCGGCUCCUGAUUGUCCAAACGCAAUUCUCGAGUCUAUGGCUCCGGCUGAGAGUUGAGUCUGGACGUCCCGAGCCGCCGCCCCCAAACCUCGAGCGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0009193
          miRBase ID: ptr-miR-219-5p
          Mature miRNA Sequence: UGAUUGUCCAAACGCAAUUCU
          Chromosome Location: NA

          Stem-loop ID ptr-mir-219-2
          Stem-loop Accession # MI0008585
          Stem-loop Sequence
          ACUCAGGGGCUUCGCCACUGAUUGUCCAAACGCAAUUCUUGUACGAGUCUGCGGCCAACCGAGAAUUGUGGCUGGACAUCUGUGGCUGAGCUCCGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0009193
          miRBase ID: ptr-miR-219-5p
          Mature miRNA Sequence: UGAUUGUCCAAACGCAAUUCU
          Chromosome Location: NA

          Pongo pygmaeus

          Stem-loop ID ppy-mir-219-1
          Stem-loop Accession # MI0002886
          Stem-loop Sequence
          CCGCCCCGGGCCGCGGCUCCUGAUUGUCCAAACGCAAUUCUCGAGUCUAUGGCCCCGGCCGAGAGUUGAGUCUGGACGUCCCGAGCCGCCGCCCCCAAACCUCGAGCGGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002577
          miRBase ID: ppy-miR-219
          Mature miRNA Sequence: UGAUUGUCCAAACGCAAUUCU
          Chromosome Location: NA

          Stem-loop ID ppy-mir-219-2
          Stem-loop Accession # MI0014869
          Stem-loop Sequence
          ACUCAGGGGCUUCGCCACUGAUUGUCCAAACGCAAUUCUUGUACGAGUCUGCGGCCAACCGAGAAUUGUGGCUGGACAUCUGUGGCUGAGCUCCGGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0015804
          miRBase ID: ppy-miR-219-5p
          Mature miRNA Sequence: UGAUUGUCCAAACGCAAUUCU
          Chromosome Location: NA

          Pteropus alecto

          Stem-loop ID pal-mir-219a-1
          Stem-loop Accession # MI0032478
          Stem-loop Sequence
          UGAUUGUCCAAACGCAAUUCUCGAGUCUCUGGCUCCGGCCGAGAGUUGAGUCUGGACGUCCCG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0039927
          miRBase ID: pal-miR-219a-5p
          Mature miRNA Sequence: UGAUUGUCCAAACGCAAUUCU
          Chromosome Location: NA

          Stem-loop ID pal-mir-219a-2
          Stem-loop Accession # MI0032404
          Stem-loop Sequence
          UGAUUGUCCAAACGCAAUUCUUGUACGAGUCUGCGGCCAACCGAGAAUUGUGGCUGGACAUCUGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0039927
          miRBase ID: pal-miR-219a-5p
          Mature miRNA Sequence: UGAUUGUCCAAACGCAAUUCU
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-219-1
          Stem-loop Accession # MI0002884
          Stem-loop Sequence
          CCGCCCCGGGCCGCGGCUCCUGAUUGUCCAAACGCAAUUCUCGAGUCUAUGGCUCUGGCCGAGAGUUGAGUCUGGACGUCCCGAGCCGCCGCCCCCAAACCUCGAGGGGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002575
          miRBase ID: mml-miR-219
          Mature miRNA Sequence: UGAUUGUCCAAACGCAAUUCU
          Chromosome Location: NA

          Stem-loop ID mml-mir-219-2
          Stem-loop Accession # MI0007676
          Stem-loop Sequence
          ACUCAGGGGCUUCGCCACUGAUUGUCCAAACGCAAUUCUUGUACGAGUCUGCGGCCAACCGAGAAUUGUGGCUGGACAUCUGUGGCUGAGCUCCGGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006245
          miRBase ID: mml-miR-219-5p
          Mature miRNA Sequence: UGAUUGUCCAAACGCAAUUCU
          Chromosome Location: NA

          Tribolium castaneum

          Stem-loop ID tca-mir-219
          Stem-loop Accession # MI0008919
          Stem-loop Sequence
          UUUCAGGCCCUGAUUGUCCAAACGCAAUUCUUGUUUGCAAUUUUAUCAAGGACUGUGUGUGGACAUCAUCGCUUGAGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0008377
          miRBase ID: tca-miR-219-5p
          Mature miRNA Sequence: UGAUUGUCCAAACGCAAUUCU
          Chromosome Location: NA

          Xenopus tropicalis

          Stem-loop ID xtr-mir-219
          Stem-loop Accession # MI0004873
          Stem-loop Sequence
          AUGGAGCUCUCGCCCUUGAUUGUCCAAACGCAAUUCUUGUUCCAAUAGAAAUAUCAAGCCAAGAAUUGUGCCUGGACAUCUGUGGCUGGUGAUCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0003632
          miRBase ID: xtr-miR-219
          Mature miRNA Sequence: UGAUUGUCCAAACGCAAUUCU
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Rn03466093_pri
          TaqMan™ Pri-miRNA Assay : Hs03303392_pri
          TaqMan™ Pri-miRNA Assay : Mm03307150_pri
          TaqMan™ Pri-miRNA Assay : Rn03464654_pri
          TaqMan™ Pri-miRNA Assay : Mm03307312_pri
          TaqMan™ Pri-miRNA Assay : Hs03302994_pri
          Ambion® Pre-miR™ miRNA Precursor : PM10664
          TaqMan™ MicroRNA Assay : 000522
          mirVana® miRNA inhibitor : MH10664
          mirVana® miRNA mimic : MC10664
          TaqMan™ Advanced miRNA Assay : 477980_mir
          TaqMan™ Advanced miRNA Assay : mmu482708_mir
          TaqMan™ Advanced miRNA Assay : rno481348_mir


          miRBase Alias:

          Human: hsa-miR-219 (v9.2)
          Human: hsa-miR-219-5p (v19)
          Mouse: mmu-miR-219 (v15)
          Mouse: mmu-miR-219-5p (v19)
          Rat: rno-miR-219-5p (v19)
          Rat: rno-miR-219 (v9.2)
          Citrobacter farmeri: cfa-miR-219 (v18)
          Gallus gallus: gga-miR-219 (v19)
          Monodelphis domestica: mdo-miR-219 (v19)

          Back To Top

          Related Products

          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-jwmpg:80/100.66.76.145:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0