Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › MH11150
          Assay Name: hsa-miR-219a-2-3p
          miRBase Accession Number: MI0000740
          miRBase Version: v22.1
          Chromosome Location: Chr.9: 128392618 - 128392714 [-] on Build GRCh38
          Mature miRNA Sequence: AGAAUUGUGGCUGGACAUCUGU
          Species: Human, Mouse, Rat, Callithrix jacchus, Dasypus novemcinctus, Dog, Guinea pig, Monodelphis domestica, Pan troglodytes, Pteropus alecto, Rabbit, Rhesus monkey
          Product Type: mirVana® miRNA inhibitor
          Assay ID MH11150

          Purification
          Size
          Availability Made To Order
          Catalog # 4464084
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000044, mir-219

          Mature miRNA Sequence:

          AGAAUUGUGGCUGGACAUCUGU

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-219a-2-3p MIMAT0004675
          Mouse mmu-miR-219a-2-3p MIMAT0022841
          Rat rno-miR-219a-2-3p MIMAT0005446
          Callithrix jacchus cja-miR-219 MIMAT0039366
          Dasypus novemcinctus dno-miR-219b-3p MIMAT0047774
          Dog cfa-miR-219-3p MIMAT0006729
          View More Rows
          Guinea pig cpo-miR-219b-3p MIMAT0047129
          Monodelphis domestica mdo-miR-219-1-3p MIMAT0026681
          Pan troglodytes ptr-miR-219-2-3p MIMAT0009194
          Pteropus alecto pal-miR-219a-2-3p MIMAT0039928
          Rabbit ocu-miR-219b-3p MIMAT0048353
          Rhesus monkey mml-miR-219-3p MIMAT0006246

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-219a-2
          Stem-loop Accession # MI0000740
          Stem-loop Sequence
          ACUCAGGGGCUUCGCCACUGAUUGUCCAAACGCAAUUCUUGUACGAGUCUGCGGCCAACCGAGAAUUGUGGCUGGACAUCUGUGGCUGAGCUCCGGG
          Chromosome Location Chr. 9 - 128392618 - 128392714 [-] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0004675
          miRBase ID: hsa-miR-219a-2-3p
          Mature miRNA Sequence: AGAAUUGUGGCUGGACAUCUGU
          Chromosome Location: Chr. 9 - 128392618 - 128392714 [-] on Build GRCh38

          Mouse

          Stem-loop ID mmu-mir-219a-2
          Stem-loop Accession # MI0000741
          Stem-loop Sequence
          ACUCAGGGGCUUCGCCACUGAUUGUCCAAACGCAAUUCUUGUACGAGUCUGCGGCCAACCGAGAAUUGUGGCUGGACAUCUGUGGUUGAGCUCCGGG
          Chromosome Location Chr. 2 - 29845631 - 29845727 [-] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0022841
          miRBase ID: mmu-miR-219a-2-3p
          Mature miRNA Sequence: AGAAUUGUGGCUGGACAUCUGU
          Chromosome Location: Chr. 2 - 29845631 - 29845727 [-] on Build GRCm38

          Rat

          Stem-loop ID rno-mir-219a-2
          Stem-loop Accession # MI0000960
          Stem-loop Sequence
          ACUCAGGGGCUUCACCACUGAUUGUCCAAACGCAAUUCUUGUACGAGUCUGCGGCCAACCGAGAAUUGUGGCUGGACAUCUGUGGUUGAGCUCCGG
          Chromosome Location Chr. 3 - 8407430 - 8407525 [-] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0005446
          miRBase ID: rno-miR-219a-2-3p
          Mature miRNA Sequence: AGAAUUGUGGCUGGACAUCUGU
          Chromosome Location: Chr. 3 - 8407430 - 8407525 [-] on Build Rnor_6.0

          Callithrix jacchus

          Stem-loop ID cja-mir-219
          Stem-loop Accession # MI0031881
          Stem-loop Sequence
          GGCCCUGAACUCAGGGGCUUCGCCACUGAUUGUCCAAACGCAAUUCUUGUAAGAGUCUGCGGCCAACCGAGAAUUGUGGCUGGACAUCUGUGGCUGAGCUCCGGGCGCAACAGGGGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0039366
          miRBase ID: cja-miR-219
          Mature miRNA Sequence: AGAAUUGUGGCUGGACAUCUGU
          Chromosome Location: NA

          Dasypus novemcinctus

          Stem-loop ID dno-mir-219b
          Stem-loop Accession # MI0039051
          Stem-loop Sequence
          UGAUUGUCCAAACGCAAUUCUUGUAUGAGUCUGCGGCCAACCGAGAAUUGUGGCUGGACAUCUGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047774
          miRBase ID: dno-miR-219b-3p
          Mature miRNA Sequence: AGAAUUGUGGCUGGACAUCUGU
          Chromosome Location: NA

          Dog

          Stem-loop ID cfa-mir-219-2
          Stem-loop Accession # MI0008150
          Stem-loop Sequence
          UGAUUGUCCAAACGCAAUUCUUGUACGAGUCUGCGGCCAACCGAGAAUUGUGGCUGGACAUCUGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006729
          miRBase ID: cfa-miR-219-3p
          Mature miRNA Sequence: AGAAUUGUGGCUGGACAUCUGU
          Chromosome Location: NA

          Guinea pig

          Stem-loop ID cpo-mir-219b
          Stem-loop Accession # MI0038712
          Stem-loop Sequence
          UGAUUGUCCAAACGCAAUUCUUGUACGAGUCUGCGGCCAACCGAGAAUUGUGGCUGGACAUCUGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047129
          miRBase ID: cpo-miR-219b-3p
          Mature miRNA Sequence: AGAAUUGUGGCUGGACAUCUGU
          Chromosome Location: NA

          Monodelphis domestica

          Stem-loop ID mdo-mir-219-1
          Stem-loop Accession # MI0005323
          Stem-loop Sequence
          CAGGGGUUCCGCCGCUGAUUGUCCAAACGCAAUUCUUGUGCGAGUCUGCAGCCAACCGAGAAUUGUGGCUGGACAUCUGUGGCUGAGCUCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0026681
          miRBase ID: mdo-miR-219-1-3p
          Mature miRNA Sequence: AGAAUUGUGGCUGGACAUCUGU
          Chromosome Location: NA

          Pan troglodytes

          Stem-loop ID ptr-mir-219-2
          Stem-loop Accession # MI0008585
          Stem-loop Sequence
          ACUCAGGGGCUUCGCCACUGAUUGUCCAAACGCAAUUCUUGUACGAGUCUGCGGCCAACCGAGAAUUGUGGCUGGACAUCUGUGGCUGAGCUCCGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0009194
          miRBase ID: ptr-miR-219-2-3p
          Mature miRNA Sequence: AGAAUUGUGGCUGGACAUCUGU
          Chromosome Location: NA

          Pteropus alecto

          Stem-loop ID pal-mir-219a-2
          Stem-loop Accession # MI0032404
          Stem-loop Sequence
          UGAUUGUCCAAACGCAAUUCUUGUACGAGUCUGCGGCCAACCGAGAAUUGUGGCUGGACAUCUGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0039928
          miRBase ID: pal-miR-219a-2-3p
          Mature miRNA Sequence: AGAAUUGUGGCUGGACAUCUGU
          Chromosome Location: NA

          Rabbit

          Stem-loop ID ocu-mir-219b
          Stem-loop Accession # MI0039356
          Stem-loop Sequence
          UGAUUGUCCAAACGCAAUUCUUGUACGAGUCUGCGGCCAACCGAGAAUUGUGGCUGGACAUCUGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0048353
          miRBase ID: ocu-miR-219b-3p
          Mature miRNA Sequence: AGAAUUGUGGCUGGACAUCUGU
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-219-2
          Stem-loop Accession # MI0007676
          Stem-loop Sequence
          ACUCAGGGGCUUCGCCACUGAUUGUCCAAACGCAAUUCUUGUACGAGUCUGCGGCCAACCGAGAAUUGUGGCUGGACAUCUGUGGCUGAGCUCCGGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006246
          miRBase ID: mml-miR-219-3p
          Mature miRNA Sequence: AGAAUUGUGGCUGGACAUCUGU
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Rn03464654_pri
          TaqMan™ Pri-miRNA Assay : Hs03303392_pri
          TaqMan™ Pri-miRNA Assay : Mm03307312_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM11150
          Ambion® Pre-miR™ miRNA Precursor : PM11150
          TaqMan™ MicroRNA Assay : 002390
          mirVana® miRNA mimic : MC11150
          TaqMan™ Advanced miRNA Assay : rno481002_mir
          TaqMan™ Advanced miRNA Assay : mmu481002_mir
          TaqMan™ Advanced miRNA Assay : 477979_mir


          miRBase Alias:

          Human: hsa-miR-219-2-3p (v19)
          Mouse: mmu-miR-219-3p (v17)
          Mouse: mmu-miR-219-2-3p (v19)
          Rat: rno-miR-219-2-3p (v19)
          Citrobacter farmeri: cfa-miR-219* (v18)

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-6wvps:80/100.66.75.98:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0