Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › AM21422
          Assay Name: hsa-miR-219b-3p
          miRBase Accession Number: MI0017299
          miRBase Version: v22.1
          Chromosome Location: Chr.9: 128392621 - 128392708 [+] on Build GRCh38
          Mature miRNA Sequence: AGAAUUGCGUUUGGACAAUCAGU
          Species: Human, Mouse, Bovine, Cricetulus griseus
          Product Type: Ambion® Anti-miR™ miRNA Inhibitor
          Assay ID AM21422
          Size
          Availability Made To Order
          Catalog # AM17000
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000044, mir-219

          Mature miRNA Sequence:

          AGAAUUGCGUUUGGACAAUCAGU

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-219b-3p MIMAT0019748
          Mouse mmu-miR-219b-3p MIMAT0029807
          Bovine bta-miR-219b-3p MIMAT0040926
          Cricetulus griseus cgr-miR-219b MIMAT0041136

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-219b
          Stem-loop Accession # MI0017299
          Stem-loop Sequence
          GGAGCUCAGCCACAGAUGUCCAGCCACAAUUCUCGGUUGGCCGCAGACUCGUACAAGAAUUGCGUUUGGACAAUCAGUGGCGAAGCCC
          Chromosome Location Chr. 9 - 128392621 - 128392708 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0019748
          miRBase ID: hsa-miR-219b-3p
          Mature miRNA Sequence: AGAAUUGCGUUUGGACAAUCAGU
          Chromosome Location: Chr. 9 - 128392621 - 128392708 [+] on Build GRCh38

          Mouse

          Stem-loop ID mmu-mir-219b
          Stem-loop Accession # MI0024990
          Stem-loop Sequence
          AGAUGUCCAGCCACAAUUCUCGGUUGGCCGCAGACUCGUACAAGAAUUGCGUUUGGACAAUCAGU
          Chromosome Location Chr. 2 - 29845647 - 29845711 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0029807
          miRBase ID: mmu-miR-219b-3p
          Mature miRNA Sequence: AGAAUUGCGUUUGGACAAUCAGU
          Chromosome Location: Chr. 2 - 29845647 - 29845711 [+] on Build GRCm38

          Bovine

          Stem-loop ID bta-mir-219b
          Stem-loop Accession # MI0032929
          Stem-loop Sequence
          AGAGCUCAGCCACAGAUGUCCAGCCACAAUUCUCGGUUGGCCGCAGACUCGUACAAGAAUUGCGUUUGGACAAUCAGUGGCGAAGCUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0040926
          miRBase ID: bta-miR-219b-3p
          Mature miRNA Sequence: AGAAUUGCGUUUGGACAAUCAGU
          Chromosome Location: NA

          Cricetulus griseus

          Stem-loop ID cgr-mir-219b
          Stem-loop Accession # MI0033064
          Stem-loop Sequence
          GGAGCUCAACCACAGAUGUCCAGCCACAAUUCUCGGUUGGCCGCAGACUCGUACAAGAAUUGCGUUUGGACAAUCAGUGGCGAAGCCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0041136
          miRBase ID: cgr-miR-219b
          Mature miRNA Sequence: AGAAUUGCGUUUGGACAAUCAGU
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Hs03303392_pri
          TaqMan™ Pri-miRNA Assay : Mm03307312_pri
          Ambion® Pre-miR™ miRNA Precursor : PM21422
          TaqMan™ MicroRNA Assay : 462260_mat
          mirVana® miRNA inhibitor : MH21422
          mirVana® miRNA mimic : MC21422
          TaqMan™ Advanced miRNA Assay : 478776_mir
          TaqMan™ Advanced miRNA Assay : mmu481656_mir


          miRBase Alias:

          Human: hsa-miR-2964a-3p (v19)

          Back To Top

          Related Products

          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-jwmpg:80/100.66.76.145:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0