Search
Search

| Assay Name: | rno-miR-32-3p |
|---|---|
| miRBase Accession Number: | MI0000873 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | GCAAUUUAGUGUGUGUGAUAUU |
| Species: | Rat |
| Product Type: | Ambion® Anti-miR™ miRNA Inhibitor |
Gene Family ID: | MIPF0000069, mir-32 |
Mature miRNA Sequence: | GCAAUUUAGUGUGUGUGAUAUU |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Rat | rno-miR-32-3p | MIMAT0017103 |