Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ?
Create Account
Applications
Real-Time PCR
Oligos, Primers, Probes and Genes
Cloning
Protein Biology
Cell Culture and Transfection
Cell and Gene Therapy
Cell Analysis
Mass Spectrometry
Chromatography
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Online Order
Custom Primers & TaqMan Probes
miRNA Mimics & Inhibitors
Stealth RNAi / siRNA
Silencer Select siRNAs
Custom DNA Oligos
GeneArt Services
GeneArt Strings DNA Fragments
TrueGuide CRISPR gRNA
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all online orderable products
Services
Custom Services
Instrument Qualification Services
Technical Services
Pipette Services
Sample Request
Instrument Maintenance Services
Repairs and Relocation Services
OEM and Licensing Services
Events, Seminars, Training
Unity Lab Services
See all services
Support
Catalogs
Application Notes
Safety Data Sheets
Legal and Regulatory
Certificates of Analysis Search
Nunc/Nalgene Product Certificates
e-learning
FAQs
Learning Centers
Technical Support Centers
See all help and support topics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign In
Sign In
Don't have an account ?
Create Account
Connect Your Lab
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Rn03464551_pri
Assay Name:
rno-mir-32
Stem-loop ID:
rno-mir-32
Mapped to miRBase Version:
v22.1
Chromosome Location:
Chr.5: 73934255 - 73934324 [-] on Build Rnor_6.0
Stem-loop Location:
Chr.5: 73934255 - 73934324 [-] on Build Rnor_6.0
Mature miRNA ID:
rno-miR-32-3p
rno-miR-32-5p
Species:
Rat
Product Type:
TaqMan
™
Pri-miRNA Assay
Assay ID
Rn03464551_pri
Size
S: 360 rxns; 20X
M: 750 rxns; 20X
L: 2900 rxns; 60X
Availability
Made To Order
Catalog #
4427012
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Amplicon Length:
95
Chromosome Location:
Chr. 5 - 73934110 - 73934110 [-] on 73934110 Build Rnor_6.0
Species:
Rat
miRBase Details:
Species
miRBase ID
miRBase Accession Number
Rat
rno-miR-32-3p
MIMAT0017103
Rat
rno-miR-32-5p
MIMAT0000811
Stem-loop Details
Rat
Stem-loop ID
rno-mir-32
Stem-loop Accession #
MI0000873
Stem-loop Sequence
GGGGAUAUUGCACAUUACUAAGUUGCAUGUUGUCACGGCCUCAAUGCAAUUUAGUGUGUGUGAUAUUCUC
Chromosome Location
Chr. 5 - 73934255 - 73934324 [-] on Build Rnor_6.0
Mature MicroRNA
miRBase Accession #: MIMAT0017103
miRBase ID: rno-miR-32-3p
Mature miRNA Sequence: GCAAUUUAGUGUGUGUGAUAUU
Chromosome Location: Chr. 5 - 73934255 - 73934324 [-] on Build Rnor_6.0
miRBase Accession #: MIMAT0000811
miRBase ID: rno-miR-32-5p
Mature miRNA Sequence: UAUUGCACAUUACUAAGUUGCA
Chromosome Location: Chr. 5 - 73934255 - 73934324 [-] on Build Rnor_6.0
Gene Family ID
MIPF0000069,
mir-32
Distance to Amplicon
110 bases
Back To Top
More Information
Related Products
Ambion
®
Anti-miR
™
miRNA Inhibitor :
AM20207
Ambion
®
Pre-miR
™
miRNA Precursor :
PM20207
TaqMan
™
MicroRNA Assay :
463582_mat
mir
Vana
®
miRNA inhibitor :
MH20207
mir
Vana
®
miRNA mimic :
MC20207
TaqMan
™
Advanced miRNA Assay :
rno481360_mir
Ambion
®
Anti-miR
™
miRNA Inhibitor :
AM12584
Ambion
®
Pre-miR
™
miRNA Precursor :
PM12584
TaqMan
™
MicroRNA Assay :
002109
mir
Vana
®
miRNA inhibitor :
MH12584
mir
Vana
®
miRNA mimic :
MC12584
TaqMan
™
Advanced miRNA Assay :
mmu482950_mir
TaqMan
™
Advanced miRNA Assay :
rno481448_mir
TaqMan
™
Advanced miRNA Assay :
478026_mir
Back To Top
Related Products
High Capacity RNA-to-cDNA Kit
SuperScript
®
IV VILO
™
Master Mix
TaqMan
®
Universal PCR Master Mix
TaqMan
®
Fast Advanced Master Mix
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.46.0-Offline