Search
Search

| Assay Name: | hsa-miR-32-5p |
|---|---|
| miRBase Accession Number: | MI0000090 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UAUUGCACAUUACUAAGUUGCA |
| Species: | Human, Mouse, Rat, Alligator mississippiensis, Anolis carolinensis, Chrysemys picta, Columba livia, Dasypus novemcinctus, Horse, Rabbit, Tupaia chinensis |
| Product Type: | mirVana® miRNA inhibitor |
Gene Family ID: | MIPF0000069, mir-32 |
Mature miRNA Sequence: | UAUUGCACAUUACUAAGUUGCA |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-32-5p | MIMAT0000090 |
| Mouse | mmu-miR-32-5p | MIMAT0000654 |
| Rat | rno-miR-32-5p | MIMAT0000811 |
| Alligator mississippiensis | ami-miR-32-5p | MIMAT0038090 |
| Anolis carolinensis | aca-miR-32-5p | MIMAT0021932 |
| Chrysemys picta | cpi-miR-32-5p | MIMAT0037687 |
| View More Rows | ||
| Columba livia | cli-miR-32-5p | MIMAT0038462 |
| Dasypus novemcinctus | dno-miR-32-5p | MIMAT0047569 |
| Horse | eca-miR-32 | MIMAT0013180 |
| Rabbit | ocu-miR-32-5p | MIMAT0048163 |
| Tupaia chinensis | tch-miR-32-5p | MIMAT0036485 |