Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › MC12584
          Assay Name: hsa-miR-32-5p
          miRBase Accession Number: MI0000090
          miRBase Version: v22.1
          Chromosome Location: Chr.9: 109046229 - 109046298 [-] on Build GRCh38
          Mature miRNA Sequence: UAUUGCACAUUACUAAGUUGCA
          Species: Human, Mouse, Rat, Alligator mississippiensis, Anolis carolinensis, Chrysemys picta, Columba livia, Dasypus novemcinctus, Horse, Rabbit, Tupaia chinensis
          Product Type: mirVana® miRNA mimic
          Assay ID MC12584

          Purification
          Size
          Availability Made To Order
          Catalog # 4464066
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000069, mir-32

          Mature miRNA Sequence:

          UAUUGCACAUUACUAAGUUGCA

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-32-5p MIMAT0000090
          Mouse mmu-miR-32-5p MIMAT0000654
          Rat rno-miR-32-5p MIMAT0000811
          Alligator mississippiensis ami-miR-32-5p MIMAT0038090
          Anolis carolinensis aca-miR-32-5p MIMAT0021932
          Chrysemys picta cpi-miR-32-5p MIMAT0037687
          View More Rows
          Columba livia cli-miR-32-5p MIMAT0038462
          Dasypus novemcinctus dno-miR-32-5p MIMAT0047569
          Horse eca-miR-32 MIMAT0013180
          Rabbit ocu-miR-32-5p MIMAT0048163
          Tupaia chinensis tch-miR-32-5p MIMAT0036485

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-32
          Stem-loop Accession # MI0000090
          Stem-loop Sequence
          GGAGAUAUUGCACAUUACUAAGUUGCAUGUUGUCACGGCCUCAAUGCAAUUUAGUGUGUGUGAUAUUUUC
          Chromosome Location Chr. 9 - 109046229 - 109046298 [-] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0000090
          miRBase ID: hsa-miR-32-5p
          Mature miRNA Sequence: UAUUGCACAUUACUAAGUUGCA
          Chromosome Location: Chr. 9 - 109046229 - 109046298 [-] on Build GRCh38

          Mouse

          Stem-loop ID mmu-mir-32
          Stem-loop Accession # MI0000691
          Stem-loop Sequence
          GGAGAUAUUGCACAUUACUAAGUUGCAUGUUGUCACGGCCUCAAUGCAAUUUAGUGUGUGUGAUAUUUUC
          Chromosome Location Chr. 4 - 56895229 - 56895298 [-] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0000654
          miRBase ID: mmu-miR-32-5p
          Mature miRNA Sequence: UAUUGCACAUUACUAAGUUGCA
          Chromosome Location: Chr. 4 - 56895229 - 56895298 [-] on Build GRCm38

          Rat

          Stem-loop ID rno-mir-32
          Stem-loop Accession # MI0000873
          Stem-loop Sequence
          GGGGAUAUUGCACAUUACUAAGUUGCAUGUUGUCACGGCCUCAAUGCAAUUUAGUGUGUGUGAUAUUCUC
          Chromosome Location Chr. 5 - 73934255 - 73934324 [-] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0000811
          miRBase ID: rno-miR-32-5p
          Mature miRNA Sequence: UAUUGCACAUUACUAAGUUGCA
          Chromosome Location: Chr. 5 - 73934255 - 73934324 [-] on Build Rnor_6.0

          Alligator mississippiensis

          Stem-loop ID ami-mir-32
          Stem-loop Accession # MI0029697
          Stem-loop Sequence
          UGCUUGCUCUGGUGGAUAUAUUGCACAUUACUAAGUUGCAUGUUGUCACGGCCUCAGUGCAAUUUAGUGUGUGCGAUAUUUUCACAUGAGUGCAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038090
          miRBase ID: ami-miR-32-5p
          Mature miRNA Sequence: UAUUGCACAUUACUAAGUUGCA
          Chromosome Location: NA

          Anolis carolinensis

          Stem-loop ID aca-mir-32
          Stem-loop Accession # MI0018853
          Stem-loop Sequence
          GGGAUAUUGCACAUUACUAAGUUGCAUGUUGUCACGGGCUCAGUGCAAUUUAGUGUGUGCGAUAUUUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0021932
          miRBase ID: aca-miR-32-5p
          Mature miRNA Sequence: UAUUGCACAUUACUAAGUUGCA
          Chromosome Location: NA

          Chrysemys picta

          Stem-loop ID cpi-mir-32
          Stem-loop Accession # MI0029431
          Stem-loop Sequence
          GCUUGCUCUGGUGGAGAUAUUGCACAUUACUAAGUUGCAUGUUGUCACGGCCUCAGUGCAAUUUAGUGUGUGCGAUAUUUUCACAUGAGUGCAUGCAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0037687
          miRBase ID: cpi-miR-32-5p
          Mature miRNA Sequence: UAUUGCACAUUACUAAGUUGCA
          Chromosome Location: NA

          Columba livia

          Stem-loop ID cli-mir-32
          Stem-loop Accession # MI0029936
          Stem-loop Sequence
          UCUGGUGGAGAUAUUGCACAUUACUAAGUUGCAUGUUGUCACGGCCUCAGUGCAAUUUAGUGUGUGCGAUACUUUCACAUGAGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038462
          miRBase ID: cli-miR-32-5p
          Mature miRNA Sequence: UAUUGCACAUUACUAAGUUGCA
          Chromosome Location: NA

          Dasypus novemcinctus

          Stem-loop ID dno-mir-32
          Stem-loop Accession # MI0038941
          Stem-loop Sequence
          UAUUGCACAUUACUAAGUUGCAUGUUGUCACGGCCUCAGUGCAAUUUAGUGUGUGUGAUAUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047569
          miRBase ID: dno-miR-32-5p
          Mature miRNA Sequence: UAUUGCACAUUACUAAGUUGCA
          Chromosome Location: NA

          Horse

          Stem-loop ID eca-mir-32
          Stem-loop Accession # MI0012924
          Stem-loop Sequence
          GGAGAUAUUGCACAUUACUAAGUUGCACGUUGUCACGGCCUCAGUGCAAUUUAGUGUGUGUGAUAUUUUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0013180
          miRBase ID: eca-miR-32
          Mature miRNA Sequence: UAUUGCACAUUACUAAGUUGCA
          Chromosome Location: NA

          Rabbit

          Stem-loop ID ocu-mir-32
          Stem-loop Accession # MI0039254
          Stem-loop Sequence
          UAUUGCACAUUACUAAGUUGCAUGUUGUCACGGCCUCAAUGCAAUUUAGUGUGUGUGAUAUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0048163
          miRBase ID: ocu-miR-32-5p
          Mature miRNA Sequence: UAUUGCACAUUACUAAGUUGCA
          Chromosome Location: NA

          Tupaia chinensis

          Stem-loop ID tch-mir-32
          Stem-loop Accession # MI0031143
          Stem-loop Sequence
          UAUUGCACAUUACUAAGUUGCAUGUUGUCACGGCCUCAAUGCAAUUUAGUGUGUGUGAUAUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036485
          miRBase ID: tch-miR-32-5p
          Mature miRNA Sequence: UAUUGCACAUUACUAAGUUGCA
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Hs03302686_pri
          TaqMan™ Pri-miRNA Assay : Mm03307087_pri
          TaqMan™ Pri-miRNA Assay : Rn03464551_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM12584
          Ambion® Pre-miR™ miRNA Precursor : PM12584
          TaqMan™ MicroRNA Assay : 002109
          mirVana® miRNA inhibitor : MH12584
          TaqMan™ Advanced miRNA Assay : mmu482950_mir
          TaqMan™ Advanced miRNA Assay : rno481448_mir
          TaqMan™ Advanced miRNA Assay : 478026_mir


          miRBase Alias:

          Human: hsa-miR-32 (v17)
          Mouse: mmu-miR-32 (v17)
          Rat: rno-miR-32 (v18)
          Aeromonas caviae Ae398: aca-miR-32 (v18)

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline