Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › 002109
          Assay Name: hsa-miR-32
          miRBase Accession Number: MI0000090
          miRBase Version: v22.1
          Chromosome Location: Chr.9: 109046229 - 109046298 [-] on Build GRCh38
          Mature miRNA Sequence: UAUUGCACAUUACUAAGUUGCA
          Species: Human, Mouse, Rat, Alligator mississippiensis, Anolis carolinensis, Chrysemys picta, Columba livia, Dasypus novemcinctus, Horse, Rabbit, Tupaia chinensis
          Product Type: TaqMan™ MicroRNA Assay
          Assay ID 002109
          Size
          Availability Inventoried
          Catalog # 4427975
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000069, mir-32

          Mature miRNA Sequence:

          UAUUGCACAUUACUAAGUUGCA

          Mature miRNA Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-32-5p MIMAT0000090
          Mouse mmu-miR-32-5p MIMAT0000654
          Rat rno-miR-32-5p MIMAT0000811
          Alligator mississippiensis ami-miR-32-5p MIMAT0038090
          Anolis carolinensis aca-miR-32-5p MIMAT0021932
          Chrysemys picta cpi-miR-32-5p MIMAT0037687
          View More Rows
          Columba livia cli-miR-32-5p MIMAT0038462
          Dasypus novemcinctus dno-miR-32-5p MIMAT0047569
          Horse eca-miR-32 MIMAT0013180
          Rabbit ocu-miR-32-5p MIMAT0048163
          Tupaia chinensis tch-miR-32-5p MIMAT0036485

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-32
          Stem-loop Accession # MI0000090
          Stem-loop Sequence
          GGAGAUAUUGCACAUUACUAAGUUGCAUGUUGUCACGGCCUCAAUGCAAUUUAGUGUGUGUGAUAUUUUC
          Chromosome Location Chr. 9 - 109046229 - 109046298 [-] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0000090
          miRBase ID: hsa-miR-32-5p
          Mature miRNA Sequence: UAUUGCACAUUACUAAGUUGCA
          Chromosome Location: Chr. 9 - 109046229 - 109046298 [-] on Build GRCh38

          Mouse

          Stem-loop ID mmu-mir-32
          Stem-loop Accession # MI0000691
          Stem-loop Sequence
          GGAGAUAUUGCACAUUACUAAGUUGCAUGUUGUCACGGCCUCAAUGCAAUUUAGUGUGUGUGAUAUUUUC
          Chromosome Location Chr. 4 - 56895229 - 56895298 [-] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0000654
          miRBase ID: mmu-miR-32-5p
          Mature miRNA Sequence: UAUUGCACAUUACUAAGUUGCA
          Chromosome Location: Chr. 4 - 56895229 - 56895298 [-] on Build GRCm38

          Rat

          Stem-loop ID rno-mir-32
          Stem-loop Accession # MI0000873
          Stem-loop Sequence
          GGGGAUAUUGCACAUUACUAAGUUGCAUGUUGUCACGGCCUCAAUGCAAUUUAGUGUGUGUGAUAUUCUC
          Chromosome Location Chr. 5 - 73934255 - 73934324 [-] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0000811
          miRBase ID: rno-miR-32-5p
          Mature miRNA Sequence: UAUUGCACAUUACUAAGUUGCA
          Chromosome Location: Chr. 5 - 73934255 - 73934324 [-] on Build Rnor_6.0

          Alligator mississippiensis

          Stem-loop ID ami-mir-32
          Stem-loop Accession # MI0029697
          Stem-loop Sequence
          UGCUUGCUCUGGUGGAUAUAUUGCACAUUACUAAGUUGCAUGUUGUCACGGCCUCAGUGCAAUUUAGUGUGUGCGAUAUUUUCACAUGAGUGCAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038090
          miRBase ID: ami-miR-32-5p
          Mature miRNA Sequence: UAUUGCACAUUACUAAGUUGCA
          Chromosome Location: NA

          Anolis carolinensis

          Stem-loop ID aca-mir-32
          Stem-loop Accession # MI0018853
          Stem-loop Sequence
          GGGAUAUUGCACAUUACUAAGUUGCAUGUUGUCACGGGCUCAGUGCAAUUUAGUGUGUGCGAUAUUUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0021932
          miRBase ID: aca-miR-32-5p
          Mature miRNA Sequence: UAUUGCACAUUACUAAGUUGCA
          Chromosome Location: NA

          Chrysemys picta

          Stem-loop ID cpi-mir-32
          Stem-loop Accession # MI0029431
          Stem-loop Sequence
          GCUUGCUCUGGUGGAGAUAUUGCACAUUACUAAGUUGCAUGUUGUCACGGCCUCAGUGCAAUUUAGUGUGUGCGAUAUUUUCACAUGAGUGCAUGCAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0037687
          miRBase ID: cpi-miR-32-5p
          Mature miRNA Sequence: UAUUGCACAUUACUAAGUUGCA
          Chromosome Location: NA

          Columba livia

          Stem-loop ID cli-mir-32
          Stem-loop Accession # MI0029936
          Stem-loop Sequence
          UCUGGUGGAGAUAUUGCACAUUACUAAGUUGCAUGUUGUCACGGCCUCAGUGCAAUUUAGUGUGUGCGAUACUUUCACAUGAGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038462
          miRBase ID: cli-miR-32-5p
          Mature miRNA Sequence: UAUUGCACAUUACUAAGUUGCA
          Chromosome Location: NA

          Dasypus novemcinctus

          Stem-loop ID dno-mir-32
          Stem-loop Accession # MI0038941
          Stem-loop Sequence
          UAUUGCACAUUACUAAGUUGCAUGUUGUCACGGCCUCAGUGCAAUUUAGUGUGUGUGAUAUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047569
          miRBase ID: dno-miR-32-5p
          Mature miRNA Sequence: UAUUGCACAUUACUAAGUUGCA
          Chromosome Location: NA

          Horse

          Stem-loop ID eca-mir-32
          Stem-loop Accession # MI0012924
          Stem-loop Sequence
          GGAGAUAUUGCACAUUACUAAGUUGCACGUUGUCACGGCCUCAGUGCAAUUUAGUGUGUGUGAUAUUUUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0013180
          miRBase ID: eca-miR-32
          Mature miRNA Sequence: UAUUGCACAUUACUAAGUUGCA
          Chromosome Location: NA

          Rabbit

          Stem-loop ID ocu-mir-32
          Stem-loop Accession # MI0039254
          Stem-loop Sequence
          UAUUGCACAUUACUAAGUUGCAUGUUGUCACGGCCUCAAUGCAAUUUAGUGUGUGUGAUAUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0048163
          miRBase ID: ocu-miR-32-5p
          Mature miRNA Sequence: UAUUGCACAUUACUAAGUUGCA
          Chromosome Location: NA

          Tupaia chinensis

          Stem-loop ID tch-mir-32
          Stem-loop Accession # MI0031143
          Stem-loop Sequence
          UAUUGCACAUUACUAAGUUGCAUGUUGUCACGGCCUCAAUGCAAUUUAGUGUGUGUGAUAUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036485
          miRBase ID: tch-miR-32-5p
          Mature miRNA Sequence: UAUUGCACAUUACUAAGUUGCA
          Chromosome Location: NA


          Back To Top

          More Information


          Associated Megaplex™ RT Primer Pools:

          Megaplex™ RT Primers, Rodent Pool A
          Megaplex™ RT Primers, Human Pool A




          Related Products

          TaqMan™ Pri-miRNA Assay : Hs03302686_pri
          TaqMan™ Pri-miRNA Assay : Mm03307087_pri
          TaqMan™ Pri-miRNA Assay : Rn03464551_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM12584
          Ambion® Pre-miR™ miRNA Precursor : PM12584
          mirVana® miRNA inhibitor : MH12584
          mirVana® miRNA mimic : MC12584
          TaqMan™ Advanced miRNA Assay : mmu482950_mir
          TaqMan™ Advanced miRNA Assay : rno481448_mir
          TaqMan™ Advanced miRNA Assay : 478026_mir
          miRNA Card : TaqMan® Rodent MicroRNA A Cards v2.0
          FGMIR : QS TaqMan OpenArray Hu miRNA Plate
          MIRNA : TaqMan® Array Human MicroRNA A+B Cards Set v3.0
          FGMIR : TaqManTM Human MicroRNA Array A
          FGMIR : TaqManTM Rodent MicroRNA Array A
          miRNA Pool : Megaplex™ RT Primers, Rodent Pool A
          MIRNA : TaqMan® OpenArray Rodent MicroRNA Panel
          miRNA Card : TaqMan® Human MicroRNA A Cards v2.0
          MIRNA : TaqMan® Array Rodent MicroRNA A+B Cards Set v3.0
          MIRNA : TaqMan® Array Human MicroRNA A Cards v2.0
          FGMIR : QS TaqMan OpenArray Rod miRNA Plate
          MIRNA : TaqMan® OpenArray Human MicroRNA Panel
          miRNA Pool : Megaplex™ RT Primers, Human Pool A
          MIRNA : TaqMan® Array Rodent MicroRNA A Card v2.0


          miRBase Alias:

          Human: hsa-miR-32 (v17)
          Mouse: mmu-miR-32 (v17)
          Rat: rno-miR-32 (v18)
          Aeromonas caviae Ae398: aca-miR-32 (v18)

          Back To Top

          Related Products

          • TaqMan® Universal Master Mix II, with UNG
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® MicroRNA Reverse Transcription Kit
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline