Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › AM12584
          Assay Name: hsa-miR-32-5p
          miRBase Accession Number: MI0000090
          miRBase Version: v22.1
          Chromosome Location: Chr.9: 109046229 - 109046298 [-] on Build GRCh38
          Mature miRNA Sequence: UAUUGCACAUUACUAAGUUGCA
          Species: Human, Mouse, Rat, Alligator mississippiensis, Anolis carolinensis, Chrysemys picta, Columba livia, Dasypus novemcinctus, Horse, Rabbit, Tupaia chinensis
          Product Type: Ambion® Anti-miR™ miRNA Inhibitor
          Assay ID AM12584
          Size
          Availability Made To Order
          Catalog # AM17000
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000069, mir-32

          Mature miRNA Sequence:

          UAUUGCACAUUACUAAGUUGCA

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-32-5p MIMAT0000090
          Mouse mmu-miR-32-5p MIMAT0000654
          Rat rno-miR-32-5p MIMAT0000811
          Alligator mississippiensis ami-miR-32-5p MIMAT0038090
          Anolis carolinensis aca-miR-32-5p MIMAT0021932
          Chrysemys picta cpi-miR-32-5p MIMAT0037687
          View More Rows
          Columba livia cli-miR-32-5p MIMAT0038462
          Dasypus novemcinctus dno-miR-32-5p MIMAT0047569
          Horse eca-miR-32 MIMAT0013180
          Rabbit ocu-miR-32-5p MIMAT0048163
          Tupaia chinensis tch-miR-32-5p MIMAT0036485

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-32
          Stem-loop Accession # MI0000090
          Stem-loop Sequence
          GGAGAUAUUGCACAUUACUAAGUUGCAUGUUGUCACGGCCUCAAUGCAAUUUAGUGUGUGUGAUAUUUUC
          Chromosome Location Chr. 9 - 109046229 - 109046298 [-] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0000090
          miRBase ID: hsa-miR-32-5p
          Mature miRNA Sequence: UAUUGCACAUUACUAAGUUGCA
          Chromosome Location: Chr. 9 - 109046229 - 109046298 [-] on Build GRCh38

          Mouse

          Stem-loop ID mmu-mir-32
          Stem-loop Accession # MI0000691
          Stem-loop Sequence
          GGAGAUAUUGCACAUUACUAAGUUGCAUGUUGUCACGGCCUCAAUGCAAUUUAGUGUGUGUGAUAUUUUC
          Chromosome Location Chr. 4 - 56895229 - 56895298 [-] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0000654
          miRBase ID: mmu-miR-32-5p
          Mature miRNA Sequence: UAUUGCACAUUACUAAGUUGCA
          Chromosome Location: Chr. 4 - 56895229 - 56895298 [-] on Build GRCm38

          Rat

          Stem-loop ID rno-mir-32
          Stem-loop Accession # MI0000873
          Stem-loop Sequence
          GGGGAUAUUGCACAUUACUAAGUUGCAUGUUGUCACGGCCUCAAUGCAAUUUAGUGUGUGUGAUAUUCUC
          Chromosome Location Chr. 5 - 73934255 - 73934324 [-] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0000811
          miRBase ID: rno-miR-32-5p
          Mature miRNA Sequence: UAUUGCACAUUACUAAGUUGCA
          Chromosome Location: Chr. 5 - 73934255 - 73934324 [-] on Build Rnor_6.0

          Alligator mississippiensis

          Stem-loop ID ami-mir-32
          Stem-loop Accession # MI0029697
          Stem-loop Sequence
          UGCUUGCUCUGGUGGAUAUAUUGCACAUUACUAAGUUGCAUGUUGUCACGGCCUCAGUGCAAUUUAGUGUGUGCGAUAUUUUCACAUGAGUGCAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038090
          miRBase ID: ami-miR-32-5p
          Mature miRNA Sequence: UAUUGCACAUUACUAAGUUGCA
          Chromosome Location: NA

          Anolis carolinensis

          Stem-loop ID aca-mir-32
          Stem-loop Accession # MI0018853
          Stem-loop Sequence
          GGGAUAUUGCACAUUACUAAGUUGCAUGUUGUCACGGGCUCAGUGCAAUUUAGUGUGUGCGAUAUUUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0021932
          miRBase ID: aca-miR-32-5p
          Mature miRNA Sequence: UAUUGCACAUUACUAAGUUGCA
          Chromosome Location: NA

          Chrysemys picta

          Stem-loop ID cpi-mir-32
          Stem-loop Accession # MI0029431
          Stem-loop Sequence
          GCUUGCUCUGGUGGAGAUAUUGCACAUUACUAAGUUGCAUGUUGUCACGGCCUCAGUGCAAUUUAGUGUGUGCGAUAUUUUCACAUGAGUGCAUGCAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0037687
          miRBase ID: cpi-miR-32-5p
          Mature miRNA Sequence: UAUUGCACAUUACUAAGUUGCA
          Chromosome Location: NA

          Columba livia

          Stem-loop ID cli-mir-32
          Stem-loop Accession # MI0029936
          Stem-loop Sequence
          UCUGGUGGAGAUAUUGCACAUUACUAAGUUGCAUGUUGUCACGGCCUCAGUGCAAUUUAGUGUGUGCGAUACUUUCACAUGAGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038462
          miRBase ID: cli-miR-32-5p
          Mature miRNA Sequence: UAUUGCACAUUACUAAGUUGCA
          Chromosome Location: NA

          Dasypus novemcinctus

          Stem-loop ID dno-mir-32
          Stem-loop Accession # MI0038941
          Stem-loop Sequence
          UAUUGCACAUUACUAAGUUGCAUGUUGUCACGGCCUCAGUGCAAUUUAGUGUGUGUGAUAUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047569
          miRBase ID: dno-miR-32-5p
          Mature miRNA Sequence: UAUUGCACAUUACUAAGUUGCA
          Chromosome Location: NA

          Horse

          Stem-loop ID eca-mir-32
          Stem-loop Accession # MI0012924
          Stem-loop Sequence
          GGAGAUAUUGCACAUUACUAAGUUGCACGUUGUCACGGCCUCAGUGCAAUUUAGUGUGUGUGAUAUUUUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0013180
          miRBase ID: eca-miR-32
          Mature miRNA Sequence: UAUUGCACAUUACUAAGUUGCA
          Chromosome Location: NA

          Rabbit

          Stem-loop ID ocu-mir-32
          Stem-loop Accession # MI0039254
          Stem-loop Sequence
          UAUUGCACAUUACUAAGUUGCAUGUUGUCACGGCCUCAAUGCAAUUUAGUGUGUGUGAUAUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0048163
          miRBase ID: ocu-miR-32-5p
          Mature miRNA Sequence: UAUUGCACAUUACUAAGUUGCA
          Chromosome Location: NA

          Tupaia chinensis

          Stem-loop ID tch-mir-32
          Stem-loop Accession # MI0031143
          Stem-loop Sequence
          UAUUGCACAUUACUAAGUUGCAUGUUGUCACGGCCUCAAUGCAAUUUAGUGUGUGUGAUAUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036485
          miRBase ID: tch-miR-32-5p
          Mature miRNA Sequence: UAUUGCACAUUACUAAGUUGCA
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Hs03302686_pri
          TaqMan™ Pri-miRNA Assay : Mm03307087_pri
          TaqMan™ Pri-miRNA Assay : Rn03464551_pri
          Ambion® Pre-miR™ miRNA Precursor : PM12584
          TaqMan™ MicroRNA Assay : 002109
          mirVana® miRNA inhibitor : MH12584
          mirVana® miRNA mimic : MC12584
          TaqMan™ Advanced miRNA Assay : mmu482950_mir
          TaqMan™ Advanced miRNA Assay : rno481448_mir
          TaqMan™ Advanced miRNA Assay : 478026_mir


          miRBase Alias:

          Human: hsa-miR-32 (v17)
          Mouse: mmu-miR-32 (v17)
          Rat: rno-miR-32 (v18)
          Aeromonas caviae Ae398: aca-miR-32 (v18)

          Back To Top

          Related Products

          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline