Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › AM20414
          Assay Name: rno-miR-212-5p
          miRBase Accession Number: MI0000952
          miRBase Version: v22.1
          Chromosome Location: Chr.10: 62014702 - 62014812 [+] on Build Rnor_6.0
          Mature miRNA Sequence: ACCUUGGCUCUAGACUGCUUACUG
          Species: Rat, Ophiophagus hannah
          Product Type: Ambion® Anti-miR™ miRNA Inhibitor
          Assay ID AM20414
          Size
          Availability Made To Order
          Catalog # AM17000
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000065, mir-132

          Mature miRNA Sequence:

          ACCUUGGCUCUAGACUGCUUACUG

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Rat rno-miR-212-5p MIMAT0017158
          Ophiophagus hannah oha-miR-212-5p MIMAT0036828

          Stem-loop Details



          Rat

          Stem-loop ID rno-mir-212
          Stem-loop Accession # MI0000952
          Stem-loop Sequence
          CGGGAUAUCCCCGCCCGGGCAGCGCGCCGGCACCUUGGCUCUAGACUGCUUACUGCCCGGGCCGCCCUCAGUAACAGUCUCCAGUCACGGCCACCGACGCCUGGCCCCGCC
          Chromosome Location Chr. 10 - 62014702 - 62014812 [+] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0017158
          miRBase ID: rno-miR-212-5p
          Mature miRNA Sequence: ACCUUGGCUCUAGACUGCUUACUG
          Chromosome Location: Chr. 10 - 62014702 - 62014812 [+] on Build Rnor_6.0

          Ophiophagus hannah

          Stem-loop ID oha-mir-212
          Stem-loop Accession # MI0031414
          Stem-loop Sequence
          CGCCCCCGGCCAGCUCGUCAGGACCUUGGCUCUAGACUGCUUACUGCUACAGGUGUGCGAGGCACAGUAACAGUCUACAGUCAUGGCCACUGACGUCUGGCAGCGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036828
          miRBase ID: oha-miR-212-5p
          Mature miRNA Sequence: ACCUUGGCUCUAGACUGCUUACUG
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Rn03465057_pri
          Ambion® Pre-miR™ miRNA Precursor : PM20414
          TaqMan™ MicroRNA Assay : 463930_mat
          mirVana® miRNA inhibitor : MH20414
          mirVana® miRNA mimic : MC20414
          TaqMan™ Advanced miRNA Assay : rno480996_mir


          miRBase Alias:

          Rat: rno-miR-212* (v18)

          Back To Top

          Related Products

          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-6wvps:80/100.66.75.98:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0