Search
Search

| Assay Name: | rno-miR-212-5p |
|---|---|
| miRBase Accession Number: | MI0000952 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | ACCUUGGCUCUAGACUGCUUACUG |
| Species: | Rat, Ophiophagus hannah |
| Product Type: | Ambion® Pre-miR™ miRNA Precursor |
Gene Family ID: | MIPF0000065, mir-132 |
Mature miRNA Sequence: | ACCUUGGCUCUAGACUGCUUACUG |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Rat | rno-miR-212-5p | MIMAT0017158 |
| Ophiophagus hannah | oha-miR-212-5p | MIMAT0036828 |