Search
Search

| Assay Name: | |
|---|---|
| miRBase Accession Number: | MI0018778 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | AACAUUCAUUGCUGUCGGUGGG |
| Species: | Anolis carolinensis, Chicken, Daubentonia madagascariensis, Fugu rubripes, Ictalurus punctatus, Nomascus leucogenys, Oreochromis niloticus, Otolemur garnettii, Papio hamadryas, Salmo salar, Tetraodon nigroviridis, Xenopus tropicalis, Zebrafish |
| Product Type: | Ambion® Pre-miR™ miRNA Precursor |
Gene Family ID: | MIPF0000007, mir-181 |
Mature miRNA Sequence: | AACAUUCAUUGCUGUCGGUGGG |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Anolis carolinensis | aca-miR-181b | MIMAT0021810 |
| Chicken | gga-miR-181b-5p | MIMAT0001151 |
| Daubentonia madagascariensis | dma-miR-181b | MIMAT0049267 |
| Fugu rubripes | fru-miR-181b | MIMAT0003003 |
| Ictalurus punctatus | ipu-miR-181b | MIMAT0029437 |
| Nomascus leucogenys | nle-miR-181b | MIMAT0049430 |
| View More Rows | ||
| Oreochromis niloticus | oni-miR-181d | MIMAT0043525 |
| Otolemur garnettii | oga-miR-181b | MIMAT0049622 |
| Otolemur garnettii | oga-miR-181e | MIMAT0049707 |
| Papio hamadryas | pha-miR-181b | MIMAT0049519 |
| Salmo salar | ssa-miR-181c-5p | MIMAT0032425 |
| Tetraodon nigroviridis | tni-miR-181b | MIMAT0003004 |
| Xenopus tropicalis | xtr-miR-181b | MIMAT0003609 |
| Zebrafish | dre-miR-181b-5p | MIMAT0001270 |