Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › MH10335
          Assay Name: aca-miR-181b
          miRBase Accession Number: MI0018778
          miRBase Version: v22.1
          Chromosome Location: -
          Mature miRNA Sequence: AACAUUCAUUGCUGUCGGUGGG
          Species: Anolis carolinensis, Chicken, Daubentonia madagascariensis, Fugu rubripes, Ictalurus punctatus, Nomascus leucogenys, Oreochromis niloticus, Otolemur garnettii, Papio hamadryas, Salmo salar, Tetraodon nigroviridis, Xenopus tropicalis, Zebrafish
          Product Type: mirVana® miRNA inhibitor
          Assay ID MH10335

          Purification
          Size
          Availability Made To Order
          Catalog # 4464084
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000007, mir-181

          Mature miRNA Sequence:

          AACAUUCAUUGCUGUCGGUGGG

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Anolis carolinensis aca-miR-181b MIMAT0021810
          Chicken gga-miR-181b-5p MIMAT0001151
          Daubentonia madagascariensis dma-miR-181b MIMAT0049267
          Fugu rubripes fru-miR-181b MIMAT0003003
          Ictalurus punctatus ipu-miR-181b MIMAT0029437
          Nomascus leucogenys nle-miR-181b MIMAT0049430
          View More Rows
          Oreochromis niloticus oni-miR-181d MIMAT0043525
          Otolemur garnettii oga-miR-181b MIMAT0049622
          Otolemur garnettii oga-miR-181e MIMAT0049707
          Papio hamadryas pha-miR-181b MIMAT0049519
          Salmo salar ssa-miR-181c-5p MIMAT0032425
          Tetraodon nigroviridis tni-miR-181b MIMAT0003004
          Xenopus tropicalis xtr-miR-181b MIMAT0003609
          Zebrafish dre-miR-181b-5p MIMAT0001270

          Stem-loop Details



          Anolis carolinensis

          Stem-loop ID aca-mir-181b-1
          Stem-loop Accession # MI0018778
          Stem-loop Sequence
          AUAACGGCUGCAAUCAACAUUCAUUGCUGUCGGUGGGUUUUAGGUUCAGUCAGCUCUCUGACCAAUGAAUGCAAACUGCAGACCAGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0021810
          miRBase ID: aca-miR-181b
          Mature miRNA Sequence: AACAUUCAUUGCUGUCGGUGGG
          Chromosome Location: NA

          Stem-loop ID aca-mir-181b-2
          Stem-loop Accession # MI0018779
          Stem-loop Sequence
          CAAAAGGUCACAAUCAACAUUCAUUGCUGUCGGUGGGUUUAACUAUGUGGACAAGCUCACUGAACAAUGAAUGCAACUGUGGCCCCAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0021810
          miRBase ID: aca-miR-181b
          Mature miRNA Sequence: AACAUUCAUUGCUGUCGGUGGG
          Chromosome Location: NA

          Chicken

          Stem-loop ID gga-mir-181b-1
          Stem-loop Accession # MI0001219
          Stem-loop Sequence
          AAAAGGUCACAAUCAACAUUCAUUGCUGUCGGUGGGUUUAACUAUGUGGACAAGCUCACUGAACAAUGAAUGCAACUGUGGCCCCACAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0001151
          miRBase ID: gga-miR-181b-5p
          Mature miRNA Sequence: AACAUUCAUUGCUGUCGGUGGG
          Chromosome Location: NA

          Stem-loop ID gga-mir-181b-2
          Stem-loop Accession # MI0001242
          Stem-loop Sequence
          CUAAUGGCUGCAAUCAACAUUCAUUGCUGUCGGUGGGUUUUCAUUGCUAUCAACUCACUGAUCAAUGAAUGCAAACUGCGGACCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0001151
          miRBase ID: gga-miR-181b-5p
          Mature miRNA Sequence: AACAUUCAUUGCUGUCGGUGGG
          Chromosome Location: NA

          Daubentonia madagascariensis

          Stem-loop ID dma-mir-181b-1
          Stem-loop Accession # MI0039995
          Stem-loop Sequence
          AACAUUCAUUGCUGUCGGUGGGUUGAACUGUGUGGACAAGCUCACUGAACAAUGAAUGCAAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049267
          miRBase ID: dma-miR-181b
          Mature miRNA Sequence: AACAUUCAUUGCUGUCGGUGGG
          Chromosome Location: NA

          Stem-loop ID dma-mir-181b-2
          Stem-loop Accession # MI0040027
          Stem-loop Sequence
          AACAUUCAUUGCUGUCGGUGGGUUUGAGUCUGAAUCAACUCACUGAUCAAUGAAUGCAAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049267
          miRBase ID: dma-miR-181b
          Mature miRNA Sequence: AACAUUCAUUGCUGUCGGUGGG
          Chromosome Location: NA

          Fugu rubripes

          Stem-loop ID fru-mir-181b
          Stem-loop Accession # MI0003331
          Stem-loop Sequence
          AAAGGUCACAAUCAACAUUCAUUGCUGUCGGUGGGUUUAACUGUAUGGAAGAGCUCACUGAACGAUGAAUGCAACUGUGUCCCAGAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0003003
          miRBase ID: fru-miR-181b
          Mature miRNA Sequence: AACAUUCAUUGCUGUCGGUGGG
          Chromosome Location: NA

          Ictalurus punctatus

          Stem-loop ID ipu-mir-181b-1
          Stem-loop Accession # MI0024532
          Stem-loop Sequence
          AACAUUCAUUGCUGUCGGUGGGUUUAGUCUUGUAACAGCUCUCUGAACAAUGAAUGUAAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0029437
          miRBase ID: ipu-miR-181b
          Mature miRNA Sequence: AACAUUCAUUGCUGUCGGUGGG
          Chromosome Location: NA

          Stem-loop ID ipu-mir-181b-2
          Stem-loop Accession # MI0024531
          Stem-loop Sequence
          AACAUUCAUUGCUGUCGGUGGGUUGGAUUGUAAAAGAAAGCUCACUGAACAAUGAAUGCAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0029437
          miRBase ID: ipu-miR-181b
          Mature miRNA Sequence: AACAUUCAUUGCUGUCGGUGGG
          Chromosome Location: NA

          Nomascus leucogenys

          Stem-loop ID nle-mir-181b-1
          Stem-loop Accession # MI0040173
          Stem-loop Sequence
          AACAUUCAUUGCUGUCGGUGGGUUGAACUGUGUGGACAAGCUCACUGAACAAUGAAUGCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049430
          miRBase ID: nle-miR-181b
          Mature miRNA Sequence: AACAUUCAUUGCUGUCGGUGGG
          Chromosome Location: NA

          Stem-loop ID nle-mir-181b-2
          Stem-loop Accession # MI0040175
          Stem-loop Sequence
          AACAUUCAUUGCUGUCGGUGGGUUUGAGUCUGAAUCAACUCACUGAUCGAUGAAUGCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049430
          miRBase ID: nle-miR-181b
          Mature miRNA Sequence: AACAUUCAUUGCUGUCGGUGGG
          Chromosome Location: NA

          Oreochromis niloticus

          Stem-loop ID oni-mir-181d
          Stem-loop Accession # MI0035579
          Stem-loop Sequence
          AACAUUCAUUGCUGUCGGUGGGUUUGCAACAGAAUAACUCACUGAUCAAUGAAUGCAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0043525
          miRBase ID: oni-miR-181d
          Mature miRNA Sequence: AACAUUCAUUGCUGUCGGUGGG
          Chromosome Location: NA

          Otolemur garnettii

          Stem-loop ID oga-mir-181b-1
          Stem-loop Accession # MI0040404
          Stem-loop Sequence
          AACAUUCAUUGCUGUCGGUGGGUUGAAUUGUGUGGACAAGCUCACUGAACAAUGAAUGCAAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049622
          miRBase ID: oga-miR-181b
          Mature miRNA Sequence: AACAUUCAUUGCUGUCGGUGGG
          Chromosome Location: NA

          Stem-loop ID oga-mir-181e
          Stem-loop Accession # MI0040492
          Stem-loop Sequence
          AACAUUCAUUGCUGUCGGUGGGUUUGAGUCUGAACCAACUCACUGAUCAAUGAAUGCAAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049707
          miRBase ID: oga-miR-181e
          Mature miRNA Sequence: AACAUUCAUUGCUGUCGGUGGG
          Chromosome Location: NA

          Papio hamadryas

          Stem-loop ID pha-mir-181b-1
          Stem-loop Accession # MI0040346
          Stem-loop Sequence
          AACAUUCAUUGCUGUCGGUGGGUUGAACUGUGUAGACAAGCUCACUGAACAAUGAAUGCAAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049519
          miRBase ID: pha-miR-181b
          Mature miRNA Sequence: AACAUUCAUUGCUGUCGGUGGG
          Chromosome Location: NA

          Stem-loop ID pha-mir-181b-2
          Stem-loop Accession # MI0040284
          Stem-loop Sequence
          AACAUUCAUUGCUGUCGGUGGGUUUGAGUCUGAAUCAACUCACUGAUCGAUGAAUGCAAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049519
          miRBase ID: pha-miR-181b
          Mature miRNA Sequence: AACAUUCAUUGCUGUCGGUGGG
          Chromosome Location: NA

          Salmo salar

          Stem-loop ID ssa-mir-181c
          Stem-loop Accession # MI0026542
          Stem-loop Sequence
          AACAUUCAUUGCUGUCGGUGGGUUUGAUGUGAAUAUAACUCACUGAUCAAUGAAUGAACA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0032425
          miRBase ID: ssa-miR-181c-5p
          Mature miRNA Sequence: AACAUUCAUUGCUGUCGGUGGG
          Chromosome Location: NA

          Tetraodon nigroviridis

          Stem-loop ID tni-mir-181b-1
          Stem-loop Accession # MI0003332
          Stem-loop Sequence
          AAAGGUCACAAUCAACAUUCAUUGCUGUCGGUGGGUUUAACUGUAUGGAAGAGCUCACUGAACGAUGAAUGCAACUGUGUCCCAGAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0003004
          miRBase ID: tni-miR-181b
          Mature miRNA Sequence: AACAUUCAUUGCUGUCGGUGGG
          Chromosome Location: NA

          Stem-loop ID tni-mir-181b-2
          Stem-loop Accession # MI0003376
          Stem-loop Sequence
          GCUCGCAAUAAACAUUCAUUGCUGUCGGUGGGUUUCCAUGGCAAUAGCUCGCUGACCAAUGAAUGAAGACUGCGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0003004
          miRBase ID: tni-miR-181b
          Mature miRNA Sequence: AACAUUCAUUGCUGUCGGUGGG
          Chromosome Location: NA

          Xenopus tropicalis

          Stem-loop ID xtr-mir-181b-1
          Stem-loop Accession # MI0004849
          Stem-loop Sequence
          AAGGUCACAAUCAACAUUCAUUGCUGUCGGUGGGUUGAGUUAAGAACACAAGCUCGCUGAACGAUGAAUGCAACUGUGUCCCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0003609
          miRBase ID: xtr-miR-181b
          Mature miRNA Sequence: AACAUUCAUUGCUGUCGGUGGG
          Chromosome Location: NA

          Stem-loop ID xtr-mir-181b-2
          Stem-loop Accession # MI0004850
          Stem-loop Sequence
          AAUGGCUGCAAUAAACAUUCAUUGCUGUCGGUGGGUUGUAGUUUAGAAAAGCUCAUUGAUCAAUGAAUGCAAACUGCAGACCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0003609
          miRBase ID: xtr-miR-181b
          Mature miRNA Sequence: AACAUUCAUUGCUGUCGGUGGG
          Chromosome Location: NA

          Zebrafish

          Stem-loop ID dre-mir-181b-1
          Stem-loop Accession # MI0001366
          Stem-loop Sequence
          CAUGUACGCACCUUCAGUUCUUCAAAGGUCAUAAUCAACAUUCAUUGCUGUCGGUGGGUUUAGUCUUGUAACAGCUCUCUGAACAAUGAAUGUAACUGUGGCCCAGAUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0001270
          miRBase ID: dre-miR-181b-5p
          Mature miRNA Sequence: AACAUUCAUUGCUGUCGGUGGG
          Chromosome Location: NA

          Stem-loop ID dre-mir-181b-2
          Stem-loop Accession # MI0001367
          Stem-loop Sequence
          CUAAUGACUGCAAUAAACAUUCAUUGCUGUCGGUGGGUUUCUAAUAGACACAACUCACUGAUCAAUGAAUGCAAACUGCGGUGCAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0001270
          miRBase ID: dre-miR-181b-5p
          Mature miRNA Sequence: AACAUUCAUUGCUGUCGGUGGG
          Chromosome Location: NA

          Stem-loop ID dre-mir-181b-3
          Stem-loop Accession # MI0039514
          Stem-loop Sequence
          AAAGGUCACAAUCAACAUUCAUUGCUGUCGGUGGGUUGGAUUGUAAAAGAAAGCUCACUGAACAAUGAAUGCAACUGUGUCCCAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0001270
          miRBase ID: dre-miR-181b-5p
          Mature miRNA Sequence: AACAUUCAUUGCUGUCGGUGGG
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          Ambion® Anti-miR™ miRNA Inhibitor : AM10335
          Ambion® Pre-miR™ miRNA Precursor : PM10335
          TaqMan™ MicroRNA Assay : 001098
          mirVana® miRNA mimic : MC10335


          miRBase Alias:

          Danio rerio: dre-miR-181b (v20)
          Gallus gallus: gga-miR-181b (v19)
          Monodelphis domestica: mdo-miR-181b (v19)

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline