Search
Search

| Assay Name: | hsa-miR-134-5p |
|---|---|
| miRBase Accession Number: | MI0000474 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UGUGACUGGUUGACCAGAGGGG |
| Species: | Human, Mouse, Rat, Callithrix jacchus, Cricetulus griseus, Dog, Goat, Guinea pig, Horse, Pan troglodytes, Rhesus monkey |
| Product Type: | Ambion® Pre-miR™ miRNA Precursor |
Gene Family ID: | MIPF0000112, mir-134 |
Mature miRNA Sequence: | UGUGACUGGUUGACCAGAGGGG |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-134-5p | MIMAT0000447 |
| Mouse | mmu-miR-134-5p | MIMAT0000146 |
| Rat | rno-miR-134-5p | MIMAT0000840 |
| Callithrix jacchus | cja-miR-134 | MIMAT0039508 |
| Cricetulus griseus | cgr-miR-134 | MIMAT0023760 |
| Dog | cfa-miR-134 | MIMAT0009883 |
| View More Rows | ||
| Goat | chi-miR-134 | MIMAT0035951 |
| Guinea pig | cpo-miR-134-5p | MIMAT0046990 |
| Horse | eca-miR-134 | MIMAT0013127 |
| Pan troglodytes | ptr-miR-134 | MIMAT0008031 |
| Rhesus monkey | mml-miR-134-5p | MIMAT0006190 |