Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › rno480922_mir

          Specificity testing was performed using rat anti-targets.

          Assay Name: rno-miR-134-5p
          Stem-loop Accession Number: MI0000474
          miRBase Version: v22.1
          Chromosome Location: Chr.14: 101054687 - 101054759 [+] on Build GRCh38
          Mature miRNA Sequence: UGUGACUGGUUGACCAGAGGGG
          Species: Human, Mouse, Rat, Callithrix jacchus, Cricetulus griseus, Dog, Goat, Guinea pig, Horse, Pan troglodytes, Rhesus monkey
          Product Type: TaqMan™ Advanced miRNA Assay
          Assay ID rno480922_mir
          Size S: 250 rxns
          Availability Inventoried
          Catalog # A25576
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000112, mir-134

          Mature miRNA Sequence:

          UGUGACUGGUUGACCAGAGGGG

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-134-5p MIMAT0000447
          Mouse mmu-miR-134-5p MIMAT0000146
          Rat rno-miR-134-5p MIMAT0000840
          Callithrix jacchus cja-miR-134 MIMAT0039508
          Cricetulus griseus cgr-miR-134 MIMAT0023760
          Dog cfa-miR-134 MIMAT0009883
          View More Rows
          Goat chi-miR-134 MIMAT0035951
          Guinea pig cpo-miR-134-5p MIMAT0046990
          Horse eca-miR-134 MIMAT0013127
          Pan troglodytes ptr-miR-134 MIMAT0008031
          Rhesus monkey mml-miR-134-5p MIMAT0006190

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-134
          Stem-loop Accession # MI0000474
          Stem-loop Sequence
          CAGGGUGUGUGACUGGUUGACCAGAGGGGCAUGCACUGUGUUCACCCUGUGGGCCACCUAGUCACCAACCCUC
          Chromosome Location Chr. 14 - 101054687 - 101054759 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0000447
          miRBase ID: hsa-miR-134-5p
          Mature miRNA Sequence: UGUGACUGGUUGACCAGAGGGG
          Chromosome Location: Chr. 14 - 101054687 - 101054759 [+] on Build GRCh38

          Mouse

          Stem-loop ID mmu-mir-134
          Stem-loop Accession # MI0000160
          Stem-loop Sequence
          AGGGUGUGUGACUGGUUGACCAGAGGGGCGUGCACUCUGUUCACCCUGUGGGCCACCUAGUCACCAACCCU
          Chromosome Location Chr. 12 - 109734139 - 109734209 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0000146
          miRBase ID: mmu-miR-134-5p
          Mature miRNA Sequence: UGUGACUGGUUGACCAGAGGGG
          Chromosome Location: Chr. 12 - 109734139 - 109734209 [+] on Build GRCm38

          Rat

          Stem-loop ID rno-mir-134
          Stem-loop Accession # MI0000907
          Stem-loop Sequence
          CAGGGUGUGUGACUGGUUGACCAGAGGGGCGUGCACUUUGUUCACCCUGUGGGCCACCUAGUCACCAACCCUC
          Chromosome Location Chr. 6 - 133884547 - 133884619 [+] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0000840
          miRBase ID: rno-miR-134-5p
          Mature miRNA Sequence: UGUGACUGGUUGACCAGAGGGG
          Chromosome Location: Chr. 6 - 133884547 - 133884619 [+] on Build Rnor_6.0

          Callithrix jacchus

          Stem-loop ID cja-mir-134
          Stem-loop Accession # MI0032034
          Stem-loop Sequence
          GUCCAGGGUGUGUGACUGGUUGACCAGAGGGGCGUGCACUGUGUUCACCCUGUGGGCCACCUAGUCACCAACCCUCAGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0039508
          miRBase ID: cja-miR-134
          Mature miRNA Sequence: UGUGACUGGUUGACCAGAGGGG
          Chromosome Location: NA

          Cricetulus griseus

          Stem-loop ID cgr-mir-134
          Stem-loop Accession # MI0020397
          Stem-loop Sequence
          UUCAGGGUGUGUGACUGGUUGACCAGAGGGGCAUGCACUCUGUUUACCCUGUGGGCCACCUAGUCACCAACCCUCAGCAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0023760
          miRBase ID: cgr-miR-134
          Mature miRNA Sequence: UGUGACUGGUUGACCAGAGGGG
          Chromosome Location: NA

          Dog

          Stem-loop ID cfa-mir-134
          Stem-loop Accession # MI0010382
          Stem-loop Sequence
          CAGGGUGUGUGACUGGUUGACCAGAGGGGCAAGCACUCUGUUCACCCUGUGGGCCACCUAGUCACCAACCCUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0009883
          miRBase ID: cfa-miR-134
          Mature miRNA Sequence: UGUGACUGGUUGACCAGAGGGG
          Chromosome Location: NA

          Goat

          Stem-loop ID chi-mir-134
          Stem-loop Accession # MI0030621
          Stem-loop Sequence
          UGGUGUCAGCACCGUUCAGGGUGUGUGACUGGUUGACCAGAGGGGCAAGCACUCCGUUCACCCUCUGGGCUGCCUCGUCACCAACCCUCAGCACCAACCGCACCUGCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0035951
          miRBase ID: chi-miR-134
          Mature miRNA Sequence: UGUGACUGGUUGACCAGAGGGG
          Chromosome Location: NA

          Guinea pig

          Stem-loop ID cpo-mir-134
          Stem-loop Accession # MI0038639
          Stem-loop Sequence
          UGUGACUGGUUGACCAGAGGGGCAUGCACUGUGUUCACCCUGUGGGCCACCUAGUCACCAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0046990
          miRBase ID: cpo-miR-134-5p
          Mature miRNA Sequence: UGUGACUGGUUGACCAGAGGGG
          Chromosome Location: NA

          Horse

          Stem-loop ID eca-mir-134
          Stem-loop Accession # MI0012877
          Stem-loop Sequence
          AGGGUGUGUGACUGGUUGACCAGAGGGGCAAGCACUCUGUUCACCCUGUGGGCCACCUAGUCACCAACCCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0013127
          miRBase ID: eca-miR-134
          Mature miRNA Sequence: UGUGACUGGUUGACCAGAGGGG
          Chromosome Location: NA

          Pan troglodytes

          Stem-loop ID ptr-mir-134
          Stem-loop Accession # MI0008535
          Stem-loop Sequence
          AGGGUGUGUGACUGGUUGACCAGAGGGGCAUGCACUGUGUUCACCCUGUGGGCCACCUAGUCACCAACCCUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0008031
          miRBase ID: ptr-miR-134
          Mature miRNA Sequence: UGUGACUGGUUGACCAGAGGGG
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-134
          Stem-loop Accession # MI0007623
          Stem-loop Sequence
          CAGGGUGUGUGACUGGUUGACCAGAGGGGCGUGCACUGUGUUCACCCUGUGGGCCACCUAGUCACCAACCCUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006190
          miRBase ID: mml-miR-134-5p
          Mature miRNA Sequence: UGUGACUGGUUGACCAGAGGGG
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Rn03465298_pri
          TaqMan™ Pri-miRNA Assay : Hs03303562_pri
          TaqMan™ Pri-miRNA Assay : Mm03307385_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM10341
          Ambion® Pre-miR™ miRNA Precursor : PM10341
          TaqMan™ MicroRNA Assay : 001186
          mirVana® miRNA inhibitor : MH10341
          mirVana® miRNA mimic : MC10341
          TaqMan™ Advanced miRNA Assay : 477901_mir
          TaqMan™ Advanced miRNA Assay : mmu480922_mir

          Back To Top

          Related Products

          • TaqMan® Advanced miRNA cDNA Synthesis Kit
          • TaqMan® Fast Advanced Master Mix
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline