Search
Search

| Assay Name: | hsa-miR-329-3p |
|---|---|
| miRBase Accession Number: | MI0001725 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | AACACACCUGGUUAACCUCUUU |
| Species: | Human, Callithrix jacchus, Dog, Pan troglodytes, Pongo pygmaeus, Pteropus alecto, Rhesus monkey, Sheep |
| Product Type: | Ambion® Pre-miR™ miRNA Precursor |
Gene Family ID: | MIPF0000110, mir-329 |
Mature miRNA Sequence: | AACACACCUGGUUAACCUCUUU |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-329-3p | MIMAT0001629 |
| Callithrix jacchus | cja-miR-329 | MIMAT0039343 |
| Dog | cfa-miR-329b | MIMAT0009899 |
| Pan troglodytes | ptr-miR-329 | MIMAT0008101 |
| Pongo pygmaeus | ppy-miR-329 | MIMAT0015832 |
| Pteropus alecto | pal-miR-329b-3p | MIMAT0040060 |
| View More Rows | ||
| Rhesus monkey | mml-miR-329-3p | MIMAT0006269 |
| Sheep | oar-miR-329b-3p | MIMAT0019264 |