Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › MC10453
          Assay Name: hsa-miR-329-3p
          miRBase Accession Number: MI0001725
          miRBase Version: v22.1
          Chromosome Location: Chr.14: 101026785 - 101026864 [+] on Build GRCh38
          Mature miRNA Sequence: AACACACCUGGUUAACCUCUUU
          Species: Human, Callithrix jacchus, Dog, Pan troglodytes, Pongo pygmaeus, Pteropus alecto, Rhesus monkey, Sheep
          Product Type: mirVana® miRNA mimic
          Assay ID MC10453

          Purification
          Size
          Availability Made To Order
          Catalog # 4464066
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000110, mir-329

          Mature miRNA Sequence:

          AACACACCUGGUUAACCUCUUU

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-329-3p MIMAT0001629
          Callithrix jacchus cja-miR-329 MIMAT0039343
          Dog cfa-miR-329b MIMAT0009899
          Pan troglodytes ptr-miR-329 MIMAT0008101
          Pongo pygmaeus ppy-miR-329 MIMAT0015832
          Pteropus alecto pal-miR-329b-3p MIMAT0040060
          View More Rows
          Rhesus monkey mml-miR-329-3p MIMAT0006269
          Sheep oar-miR-329b-3p MIMAT0019264

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-329-1
          Stem-loop Accession # MI0001725
          Stem-loop Sequence
          GGUACCUGAAGAGAGGUUUUCUGGGUUUCUGUUUCUUUAAUGAGGACGAAACACACCUGGUUAACCUCUUUUCCAGUAUC
          Chromosome Location Chr. 14 - 101026785 - 101026864 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0001629
          miRBase ID: hsa-miR-329-3p
          Mature miRNA Sequence: AACACACCUGGUUAACCUCUUU
          Chromosome Location: Chr. 14 - 101027100 - 101027183 [+] on Build GRCh38

          Stem-loop ID hsa-mir-329-2
          Stem-loop Accession # MI0001726
          Stem-loop Sequence
          GUGGUACCUGAAGAGAGGUUUUCUGGGUUUCUGUUUCUUUAUUGAGGACGAAACACACCUGGUUAACCUCUUUUCCAGUAUCAA
          Chromosome Location Chr. 14 - 101027100 - 101027183 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0001629
          miRBase ID: hsa-miR-329-3p
          Mature miRNA Sequence: AACACACCUGGUUAACCUCUUU
          Chromosome Location: Chr. 14 - 101027100 - 101027183 [+] on Build GRCh38

          Callithrix jacchus

          Stem-loop ID cja-mir-329
          Stem-loop Accession # MI0031856
          Stem-loop Sequence
          UGGUACCUGAAGAGAGGUUUUCUGGGGUUCUGUUUCUUUAAUGAGGAUGAAACACACCUGGUUAACCUCUUUUCCAGUAUCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0039343
          miRBase ID: cja-miR-329
          Mature miRNA Sequence: AACACACCUGGUUAACCUCUUU
          Chromosome Location: NA

          Dog

          Stem-loop ID cfa-mir-329b
          Stem-loop Accession # MI0010398
          Stem-loop Sequence
          GGUACCUGAAGAGAGGUUUUCUGGGUUUCUGUUUCUUUAAUGAGGAUGAAACACACCUGGUUAACCUCUUUUCCAGUAUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0009899
          miRBase ID: cfa-miR-329b
          Mature miRNA Sequence: AACACACCUGGUUAACCUCUUU
          Chromosome Location: NA

          Pan troglodytes

          Stem-loop ID ptr-mir-329-1
          Stem-loop Accession # MI0008619
          Stem-loop Sequence
          GUACCUGAAGAGAGGUUUUCUGGGUUUCUGUUUCUUUAAUGAGGACGAAACACACCUGGUUAACCUCUUUUCCAGUAUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0008101
          miRBase ID: ptr-miR-329
          Mature miRNA Sequence: AACACACCUGGUUAACCUCUUU
          Chromosome Location: NA

          Stem-loop ID ptr-mir-329-2
          Stem-loop Accession # MI0008620
          Stem-loop Sequence
          GUACCUGAAGAGAGGUUUUCUGGGUUUCUGUUUCUUUAUUGAGGAUGAAACACACCUGGUUAACCUCUUUUCCAGUAUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0008101
          miRBase ID: ptr-miR-329
          Mature miRNA Sequence: AACACACCUGGUUAACCUCUUU
          Chromosome Location: NA

          Pongo pygmaeus

          Stem-loop ID ppy-mir-329-1
          Stem-loop Accession # MI0014896
          Stem-loop Sequence
          GGUACCUGAAGAGAGGUUUUCUGGGUUUCUGUUUCUUUAAUGAGGAUGAAACACACCUGGUUAACCUCUUUUCCAGUAUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0015832
          miRBase ID: ppy-miR-329
          Mature miRNA Sequence: AACACACCUGGUUAACCUCUUU
          Chromosome Location: NA

          Stem-loop ID ppy-mir-329-2
          Stem-loop Accession # MI0014897
          Stem-loop Sequence
          GUGGUACCUGAAGAGAGGUUUUCUGGGUUUCUGUUUCUUUAUUGAGGAUGAAACACACCUGGUUAACCUCUUUUCCAGUAUCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0015832
          miRBase ID: ppy-miR-329
          Mature miRNA Sequence: AACACACCUGGUUAACCUCUUU
          Chromosome Location: NA

          Pteropus alecto

          Stem-loop ID pal-mir-329b
          Stem-loop Accession # MI0032472
          Stem-loop Sequence
          AGAGGUUUUCUGGGUUUCUGUUUCUUUAAUGAGGAUGAAACACACCUGGUUAACCUCUUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0040060
          miRBase ID: pal-miR-329b-3p
          Mature miRNA Sequence: AACACACCUGGUUAACCUCUUU
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-329-1
          Stem-loop Accession # MI0007695
          Stem-loop Sequence
          GUGGUACCUGAAGGGAGGUUUUCUGGGUUUCUGUUUCUUUAAUGAGGAUGAAACACACCUGGUUAACCUCUUUUCCAGUAUCAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006269
          miRBase ID: mml-miR-329-3p
          Mature miRNA Sequence: AACACACCUGGUUAACCUCUUU
          Chromosome Location: NA

          Stem-loop ID mml-mir-329-2
          Stem-loop Accession # MI0007696
          Stem-loop Sequence
          GGUACCUGAAGGGAGGUUUUCUGGGUCUCUGUUUCUUUACUGAGGAUGAAACACACCUGGUUAACCUCUUUUCCAGUAUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006269
          miRBase ID: mml-miR-329-3p
          Mature miRNA Sequence: AACACACCUGGUUAACCUCUUU
          Chromosome Location: NA

          Sheep

          Stem-loop ID oar-mir-329b
          Stem-loop Accession # MI0016925
          Stem-loop Sequence
          CACUGGUGUUCUGGUCAGUGUUCACCCAUGGUACCUGAAGAGAGGUUUUCUGGGUUUCUGUUUCUUUAAUGAGGAUGAAACACACCUGGUUAACCUCUUUUCCAGUAUCAAAUCCCAUCUUGAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0019264
          miRBase ID: oar-miR-329b-3p
          Mature miRNA Sequence: AACACACCUGGUUAACCUCUUU
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Hs03303752_pri
          TaqMan™ Pri-miRNA Assay : Hs04225947_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM10453
          Ambion® Pre-miR™ miRNA Precursor : PM10453
          TaqMan™ MicroRNA Assay : 001101
          mirVana® miRNA inhibitor : MH10453
          TaqMan™ Advanced miRNA Assay : 478029_mir


          miRBase Alias:

          Human: hsa-miR-329 (v19)
          Macaca mulatta: mml-miR-329 (v19)

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline