Search
Search

| Assay Name: | hsa-miR-30e-3p |
|---|---|
| miRBase Accession Number: | MI0000749 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | CUUUCAGUCGGAUGUUUACAGC |
| Species: | Human, Mouse, Rat, Alligator mississippiensis, Anolis carolinensis, Chrysemys picta, Cricetulus griseus, Dasypus novemcinctus, Daubentonia madagascariensis, Dog, Guinea pig, Ophiophagus hannah, Ornithorhynchus anatinus, Pig, Pteropus alecto, Python bivittatus, Rabbit, Taeniopygia guttata |
| Product Type: | Ambion® Pre-miR™ miRNA Precursor |
Gene Family ID: | MIPF0000005, mir-30 |
Mature miRNA Sequence: | CUUUCAGUCGGAUGUUUACAGC |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-30e-3p | MIMAT0000693 |
| Mouse | mmu-miR-30e-3p | MIMAT0000249 |
| Rat | rno-miR-30e-3p | MIMAT0004720 |
| Alligator mississippiensis | ami-miR-30e-3p | MIMAT0038087 |
| Anolis carolinensis | aca-miR-30e-3p | MIMAT0021929 |
| Chrysemys picta | cpi-miR-30e-3p | MIMAT0037684 |
| View More Rows | ||
| Cricetulus griseus | cgr-miR-30e-3p | MIMAT0023901 |
| Dasypus novemcinctus | dno-miR-30e-3p | MIMAT0047566 |
| Daubentonia madagascariensis | dma-miR-30e | MIMAT0049282 |
| Dog | cfa-miR-30e | MIMAT0006627 |
| Guinea pig | cpo-miR-30e-3p | MIMAT0046913 |
| Ophiophagus hannah | oha-miR-30e-3p | MIMAT0036887 |
| Ornithorhynchus anatinus | oan-miR-30e-3p | MIMAT0007204 |
| Pig | ssc-miR-30e-3p | MIMAT0013873 |
| Pteropus alecto | pal-miR-30e-3p | MIMAT0039833 |
| Python bivittatus | pbv-miR-30e-3p | MIMAT0038879 |
| Rabbit | ocu-miR-30e-3p | MIMAT0048160 |
| Taeniopygia guttata | tgu-miR-30a-3p | MIMAT0014535 |