Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › rno478388_mir

          Specificity testing was performed using rat anti-targets.

          Assay Name: rno-miR-30e-3p
          Stem-loop Accession Number: MI0000749
          miRBase Version: v22.1
          Chromosome Location: Chr.1: 40754355 - 40754446 [+] on Build GRCh38
          Mature miRNA Sequence: CUUUCAGUCGGAUGUUUACAGC
          Species: Human, Mouse, Rat, Alligator mississippiensis, Anolis carolinensis, Chrysemys picta, Cricetulus griseus, Dasypus novemcinctus, Daubentonia madagascariensis, Dog, Guinea pig, Ophiophagus hannah, Ornithorhynchus anatinus, Pig, Pteropus alecto, Python bivittatus, Rabbit, Taeniopygia guttata
          Product Type: TaqMan™ Advanced miRNA Assay
          Assay ID rno478388_mir
          Size S: 250 rxns
          Availability Inventoried
          Catalog # A25576
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000005, mir-30

          Mature miRNA Sequence:

          CUUUCAGUCGGAUGUUUACAGC

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-30e-3p MIMAT0000693
          Mouse mmu-miR-30e-3p MIMAT0000249
          Rat rno-miR-30e-3p MIMAT0004720
          Alligator mississippiensis ami-miR-30e-3p MIMAT0038087
          Anolis carolinensis aca-miR-30e-3p MIMAT0021929
          Chrysemys picta cpi-miR-30e-3p MIMAT0037684
          View More Rows
          Cricetulus griseus cgr-miR-30e-3p MIMAT0023901
          Dasypus novemcinctus dno-miR-30e-3p MIMAT0047566
          Daubentonia madagascariensis dma-miR-30e MIMAT0049282
          Dog cfa-miR-30e MIMAT0006627
          Guinea pig cpo-miR-30e-3p MIMAT0046913
          Ophiophagus hannah oha-miR-30e-3p MIMAT0036887
          Ornithorhynchus anatinus oan-miR-30e-3p MIMAT0007204
          Pig ssc-miR-30e-3p MIMAT0013873
          Pteropus alecto pal-miR-30e-3p MIMAT0039833
          Python bivittatus pbv-miR-30e-3p MIMAT0038879
          Rabbit ocu-miR-30e-3p MIMAT0048160
          Taeniopygia guttata tgu-miR-30a-3p MIMAT0014535

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-30e
          Stem-loop Accession # MI0000749
          Stem-loop Sequence
          GGGCAGUCUUUGCUACUGUAAACAUCCUUGACUGGAAGCUGUAAGGUGUUCAGAGGAGCUUUCAGUCGGAUGUUUACAGCGGCAGGCUGCCA
          Chromosome Location Chr. 1 - 40754355 - 40754446 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0000693
          miRBase ID: hsa-miR-30e-3p
          Mature miRNA Sequence: CUUUCAGUCGGAUGUUUACAGC
          Chromosome Location: Chr. 1 - 40754355 - 40754446 [+] on Build GRCh38

          Mouse

          Stem-loop ID mmu-mir-30e
          Stem-loop Accession # MI0000259
          Stem-loop Sequence
          GGGCAGUCUUUGCUACUGUAAACAUCCUUGACUGGAAGCUGUAAGGUGUUGAGAGGAGCUUUCAGUCGGAUGUUUACAGCGGCAGGCUGCCA
          Chromosome Location Chr. 4 - 120772606 - 120772697 [-] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0000249
          miRBase ID: mmu-miR-30e-3p
          Mature miRNA Sequence: CUUUCAGUCGGAUGUUUACAGC
          Chromosome Location: Chr. 4 - 120772606 - 120772697 [-] on Build GRCm38

          Rat

          Stem-loop ID rno-mir-30e
          Stem-loop Accession # MI0000867
          Stem-loop Sequence
          GGGCAGUCUUUGCUACUGUAAACAUCCUUGACUGGAAGCUGUAAGGUGUUGAGAGGAGCUUUCAGUCGGAUGUUUACAGCGGCAGGCUGCCA
          Chromosome Location Chr. 5 - 139702872 - 139702963 [-] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0004720
          miRBase ID: rno-miR-30e-3p
          Mature miRNA Sequence: CUUUCAGUCGGAUGUUUACAGC
          Chromosome Location: Chr. 5 - 139702872 - 139702963 [-] on Build Rnor_6.0

          Alligator mississippiensis

          Stem-loop ID ami-mir-30e
          Stem-loop Accession # MI0029695
          Stem-loop Sequence
          CCUGGGCAGUCGAUGCUACUGUAAACAUCCUUGACUGGAAGCUGUAAGGUGCUAGAAGGAGCUUUCAGUCGGAUGUUUACAGCGGCAGGCUGCCACAGUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038087
          miRBase ID: ami-miR-30e-3p
          Mature miRNA Sequence: CUUUCAGUCGGAUGUUUACAGC
          Chromosome Location: NA

          Anolis carolinensis

          Stem-loop ID aca-mir-30e
          Stem-loop Accession # MI0018851
          Stem-loop Sequence
          UAGGCAGUUGUGCUCCUGUAAACAUCCUUGACUGGAAGCUGUGCAAGGCUCAAAGGGGCUUUCAGUCGGAUGUUUACAGCGGCAGGCUGCUGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0021929
          miRBase ID: aca-miR-30e-3p
          Mature miRNA Sequence: CUUUCAGUCGGAUGUUUACAGC
          Chromosome Location: NA

          Chrysemys picta

          Stem-loop ID cpi-mir-30e
          Stem-loop Accession # MI0029429
          Stem-loop Sequence
          GGGCAGUCGUUGCUACUGUAAACAUCCUUGACUGGAAGCUGUAAGGUGCUAGAAGGGAGCUUUCAGUCGGAUGUUUACAGCGGCAGGCUGCCACAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0037684
          miRBase ID: cpi-miR-30e-3p
          Mature miRNA Sequence: CUUUCAGUCGGAUGUUUACAGC
          Chromosome Location: NA

          Cricetulus griseus

          Stem-loop ID cgr-mir-30e
          Stem-loop Accession # MI0020488
          Stem-loop Sequence
          CUUUGCUACUGUAAACAUCCUUGACUGGAAGCUGUAAGGUGUUGAGAAGAGCUUUCAGUCGGAUGUUUACAGCGGCAGGCUGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0023901
          miRBase ID: cgr-miR-30e-3p
          Mature miRNA Sequence: CUUUCAGUCGGAUGUUUACAGC
          Chromosome Location: NA

          Dasypus novemcinctus

          Stem-loop ID dno-mir-30e
          Stem-loop Accession # MI0038939
          Stem-loop Sequence
          UGUAAACAUCCUUGACUGGAAGCUGUAAAGUGUUGAGAGGAGCUUUCAGUCGGAUGUUUACAGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047566
          miRBase ID: dno-miR-30e-3p
          Mature miRNA Sequence: CUUUCAGUCGGAUGUUUACAGC
          Chromosome Location: NA

          Daubentonia madagascariensis

          Stem-loop ID dma-mir-30e
          Stem-loop Accession # MI0040013
          Stem-loop Sequence
          UGUAAACAUCCUUGACUGGAAGCUGUAAGGUGUUCAGAGGAGCUUUCAGUCGGAUGUUUACAGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049282
          miRBase ID: dma-miR-30e
          Mature miRNA Sequence: CUUUCAGUCGGAUGUUUACAGC
          Chromosome Location: NA

          Dog

          Stem-loop ID cfa-mir-30e
          Stem-loop Accession # MI0008023
          Stem-loop Sequence
          UGUAAACAUCCUUGACUGGAAGCUGUAAGGUGCUCAGAGGGGCUUUCAGUCGGAUGUUUACAGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006627
          miRBase ID: cfa-miR-30e
          Mature miRNA Sequence: CUUUCAGUCGGAUGUUUACAGC
          Chromosome Location: NA

          Guinea pig

          Stem-loop ID cpo-mir-30e
          Stem-loop Accession # MI0038595
          Stem-loop Sequence
          UGUAAACAUCCUUGACUGGAAGCUGUAAAGUGUUCAGAGGAGCUUUCAGUCGGAUGUUUACAGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0046913
          miRBase ID: cpo-miR-30e-3p
          Mature miRNA Sequence: CUUUCAGUCGGAUGUUUACAGC
          Chromosome Location: NA

          Ophiophagus hannah

          Stem-loop ID oha-mir-30e
          Stem-loop Accession # MI0031449
          Stem-loop Sequence
          UGCCCCUGUAAACAUCCUUGACUGGAAGCUGUGCAGGCUCAAAGAGGCUUUCAGUCGGAUGUUUACAGCGGCAGGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036887
          miRBase ID: oha-miR-30e-3p
          Mature miRNA Sequence: CUUUCAGUCGGAUGUUUACAGC
          Chromosome Location: NA

          Ornithorhynchus anatinus

          Stem-loop ID oan-mir-30e
          Stem-loop Accession # MI0006911
          Stem-loop Sequence
          GACAGUCUUUGCUACUGUAAACAUCCUUGACUGGAAGCUGGAAGGUGCCGAAGGAGCUUUCAGUCGGAUGUUUACAGCGGCAGGCUGCCACAGUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0007204
          miRBase ID: oan-miR-30e-3p
          Mature miRNA Sequence: CUUUCAGUCGGAUGUUUACAGC
          Chromosome Location: NA

          Pig

          Stem-loop ID ssc-mir-30e
          Stem-loop Accession # MI0013092
          Stem-loop Sequence
          UUUGCCACUGUAAACAUCCUUGACUGGAAGCUGUGAGGCGUUCCCAGGAGCUUUCAGUCGGAUGUUUACAGCGGCAGGCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0013873
          miRBase ID: ssc-miR-30e-3p
          Mature miRNA Sequence: CUUUCAGUCGGAUGUUUACAGC
          Chromosome Location: NA

          Pteropus alecto

          Stem-loop ID pal-mir-30e
          Stem-loop Accession # MI0032352
          Stem-loop Sequence
          UGUAAACAUCCUUGACUGGAAGCUGUAAGGUGUUCAGAGGAGCUUUCAGUCGGAUGUUUACAGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0039833
          miRBase ID: pal-miR-30e-3p
          Mature miRNA Sequence: CUUUCAGUCGGAUGUUUACAGC
          Chromosome Location: NA

          Python bivittatus

          Stem-loop ID pbv-mir-30e
          Stem-loop Accession # MI0030188
          Stem-loop Sequence
          UGGGCAGUUGUUGCUCCUGUAAACAUCCUUGACUGGAAGCUGUACAGGCUCAAAGAGGCUUUCAGUCGGAUGUUUACAGCGGCAGGCUGCCACAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038879
          miRBase ID: pbv-miR-30e-3p
          Mature miRNA Sequence: CUUUCAGUCGGAUGUUUACAGC
          Chromosome Location: NA

          Rabbit

          Stem-loop ID ocu-mir-30e
          Stem-loop Accession # MI0039252
          Stem-loop Sequence
          UGUAAACAUCCUUGACUGGAAGCUGUAAGGUGUUAAGAGGAGCUUUCAGUCGGAUGUUUACAGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0048160
          miRBase ID: ocu-miR-30e-3p
          Mature miRNA Sequence: CUUUCAGUCGGAUGUUUACAGC
          Chromosome Location: NA

          Taeniopygia guttata

          Stem-loop ID tgu-mir-30a
          Stem-loop Accession # MI0013749
          Stem-loop Sequence
          UUGGGGCAGUCAGUGCCUCUGUAAACAUCCUUGACUGGAAGCUGGAAGGGCUGGAAGGAGCUUUCAGUCGGAUGUUUACAGCAGCAGGCUGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0014535
          miRBase ID: tgu-miR-30a-3p
          Mature miRNA Sequence: CUUUCAGUCGGAUGUUUACAGC
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Hs03303424_pri
          TaqMan™ Pri-miRNA Assay : Rn03464782_pri
          TaqMan™ Pri-miRNA Assay : Mm03306569_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM10491
          Ambion® Pre-miR™ miRNA Precursor : PM10491
          TaqMan™ MicroRNA Assay : 000422
          mirVana® miRNA inhibitor : MH10491
          mirVana® miRNA mimic : MC10491
          TaqMan™ Advanced miRNA Assay : 478388_mir
          TaqMan™ Advanced miRNA Assay : mmu478388_mir

          Back To Top

          Related Products

          • TaqMan® Advanced miRNA cDNA Synthesis Kit
          • TaqMan® Fast Advanced Master Mix
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline