Search
Search

| Assay Name: | |
|---|---|
| miRBase Accession Number: | MI0000003 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UGGAAUGUAAAGAAGUAUGUA |
| Species: | Caenorhabditis elegans, Ascaris suum, Caenorhabditis brenneri, Caenorhabditis briggsae, Chicken, Dog, Gorilla gorilla, Heligmosomoides polygyrus, Oryzias latipes, Pan paniscus, Pig, Python bivittatus, Xenopus tropicalis |
| Product Type: | Ambion® Pre-miR™ miRNA Precursor |
Gene Family ID: | MIPF0000038, mir-1 |
Mature miRNA Sequence: | UGGAAUGUAAAGAAGUAUGUA |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Caenorhabditis elegans | cel-miR-1-3p | MIMAT0000003 |
| Ascaris suum | asu-miR-1-3p | MIMAT0021424 |
| Caenorhabditis brenneri | cbn-miR-1 | MIMAT0029710 |
| Caenorhabditis briggsae | cbr-miR-1 | MIMAT0000465 |
| Chicken | gga-miR-1a-3p | MIMAT0001127 |
| Dog | cfa-miR-1 | MIMAT0006656 |
| View More Rows | ||
| Gorilla gorilla | ggo-miR-1 | MIMAT0024078 |
| Heligmosomoides polygyrus | hpo-miR-1-3p | MIMAT0040465 |
| Oryzias latipes | ola-miR-1-3p | MIMAT0022558 |
| Pan paniscus | ppa-miR-1 | MIMAT0002621 |
| Pig | ssc-miR-1 | MIMAT0010187 |
| Python bivittatus | pbv-miR-1a-3p | MIMAT0038813 |
| Xenopus tropicalis | xtr-miR-1a | MIMAT0003551 |