Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › MH10660
          Assay Name: cel-miR-1-3p
          miRBase Accession Number: MI0000003
          miRBase Version: v22.1
          Chromosome Location: -
          Mature miRNA Sequence: UGGAAUGUAAAGAAGUAUGUA
          Species: Caenorhabditis elegans, Ascaris suum, Caenorhabditis brenneri, Caenorhabditis briggsae, Chicken, Dog, Gorilla gorilla, Heligmosomoides polygyrus, Oryzias latipes, Pan paniscus, Pig, Python bivittatus, Xenopus tropicalis
          Product Type: mirVana® miRNA inhibitor
          Assay ID MH10660

          Purification
          Size
          Availability Made To Order
          Catalog # 4464084
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000038, mir-1

          Mature miRNA Sequence:

          UGGAAUGUAAAGAAGUAUGUA

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Caenorhabditis elegans cel-miR-1-3p MIMAT0000003
          Ascaris suum asu-miR-1-3p MIMAT0021424
          Caenorhabditis brenneri cbn-miR-1 MIMAT0029710
          Caenorhabditis briggsae cbr-miR-1 MIMAT0000465
          Chicken gga-miR-1a-3p MIMAT0001127
          Dog cfa-miR-1 MIMAT0006656
          View More Rows
          Gorilla gorilla ggo-miR-1 MIMAT0024078
          Heligmosomoides polygyrus hpo-miR-1-3p MIMAT0040465
          Oryzias latipes ola-miR-1-3p MIMAT0022558
          Pan paniscus ppa-miR-1 MIMAT0002621
          Pig ssc-miR-1 MIMAT0010187
          Python bivittatus pbv-miR-1a-3p MIMAT0038813
          Xenopus tropicalis xtr-miR-1a MIMAT0003551

          Stem-loop Details



          Caenorhabditis elegans

          Stem-loop ID cel-mir-1
          Stem-loop Accession # MI0000003
          Stem-loop Sequence
          AAAGUGACCGUACCGAGCUGCAUACUUCCUUACAUGCCCAUACUAUAUCAUAAAUGGAUAUGGAAUGUAAAGAAGUAUGUAGAACGGGGUGGUAGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0000003
          miRBase ID: cel-miR-1-3p
          Mature miRNA Sequence: UGGAAUGUAAAGAAGUAUGUA
          Chromosome Location: NA

          Ascaris suum

          Stem-loop ID asu-mir-1
          Stem-loop Accession # MI0018528
          Stem-loop Sequence
          UUCUGUUGCAUCGUACUUCUUUACAUCGCCAUACUGGUAUUUCAAAGAGCUAUGGAAUGUAAAGAAGUAUGUAGUUACUGAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0021424
          miRBase ID: asu-miR-1-3p
          Mature miRNA Sequence: UGGAAUGUAAAGAAGUAUGUA
          Chromosome Location: NA

          Caenorhabditis brenneri

          Stem-loop ID cbn-mir-1-1
          Stem-loop Accession # MI0024872
          Stem-loop Sequence
          CAUGCUUCUUCUACAUGCCCAUACUAUAUCGUAAAUGGAUAUGGAAUGUAAAGAAGUAUGUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0029710
          miRBase ID: cbn-miR-1
          Mature miRNA Sequence: UGGAAUGUAAAGAAGUAUGUA
          Chromosome Location: NA

          Stem-loop ID cbn-mir-1-2
          Stem-loop Accession # MI0024873
          Stem-loop Sequence
          CAUGCUUCUUCUACAUGCCCAUACUAUAUCGUAAAUGGAUAUGGAAUGUAAAGAAGUAUGUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0029710
          miRBase ID: cbn-miR-1
          Mature miRNA Sequence: UGGAAUGUAAAGAAGUAUGUA
          Chromosome Location: NA

          Caenorhabditis briggsae

          Stem-loop ID cbr-mir-1
          Stem-loop Accession # MI0000493
          Stem-loop Sequence
          UGCUGUGCCGAGCUGCAUACUUCCUUACAUGCCCAUACUGUACUGUGAAUGGAUAUGGAAUGUAAAGAAGUAUGUAGAACGGAAUCUAACGGCG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0000465
          miRBase ID: cbr-miR-1
          Mature miRNA Sequence: UGGAAUGUAAAGAAGUAUGUA
          Chromosome Location: NA

          Chicken

          Stem-loop ID gga-mir-1a-1
          Stem-loop Accession # MI0001247
          Stem-loop Sequence
          UGAGAGACAUACUUCUUUAUAUGCCCAUAUGAACCUGGCAAUCUAUGGAAUGUAAAGAAGUAUGUAUUUCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0001127
          miRBase ID: gga-miR-1a-3p
          Mature miRNA Sequence: UGGAAUGUAAAGAAGUAUGUA
          Chromosome Location: NA

          Stem-loop ID gga-mir-1a-2
          Stem-loop Accession # MI0001196
          Stem-loop Sequence
          ACCUGCUCAGAGCACAUACUUCUUUAUGUACCCAUAUGAACAUACAAUGCUAUGGAAUGUAAAGAAGUAUGUAUUUUUGGCAGGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0001127
          miRBase ID: gga-miR-1a-3p
          Mature miRNA Sequence: UGGAAUGUAAAGAAGUAUGUA
          Chromosome Location: NA

          Dog

          Stem-loop ID cfa-mir-1-1
          Stem-loop Accession # MI0008060
          Stem-loop Sequence
          CAUACUUCUUUAUAUGCCCAUAUGGACCUGCUAAGCUAUGGAAUGUAAAGAAGUAUGUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006656
          miRBase ID: cfa-miR-1
          Mature miRNA Sequence: UGGAAUGUAAAGAAGUAUGUA
          Chromosome Location: NA

          Stem-loop ID cfa-mir-1-2
          Stem-loop Accession # MI0008118
          Stem-loop Sequence
          CAUACUUCUUUAUGUACCCAUAUGAACAUACAAUGCUAUGGAAUGUAAAGAAGUAUGUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006656
          miRBase ID: cfa-miR-1
          Mature miRNA Sequence: UGGAAUGUAAAGAAGUAUGUA
          Chromosome Location: NA

          Gorilla gorilla

          Stem-loop ID ggo-mir-1
          Stem-loop Accession # MI0020625
          Stem-loop Sequence
          AGCUAACAACUUAGUAAUACCUACUCAGAGUACAUACUUCUUUAUGUACCCAUAUGAACAUACAAUGCUAUGGAAUGUAAAGAAGUAUGUAUUUUUGGUAGGCAAUAAACC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0024078
          miRBase ID: ggo-miR-1
          Mature miRNA Sequence: UGGAAUGUAAAGAAGUAUGUA
          Chromosome Location: NA

          Heligmosomoides polygyrus

          Stem-loop ID hpo-mir-1
          Stem-loop Accession # MI0032689
          Stem-loop Sequence
          CGUGCUUCUUUGCAUGCCCAUACUGUAUUUUGAUAUGGAAUGUAAAGAAGUAUGUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0040465
          miRBase ID: hpo-miR-1-3p
          Mature miRNA Sequence: UGGAAUGUAAAGAAGUAUGUA
          Chromosome Location: NA

          Oryzias latipes

          Stem-loop ID ola-mir-1-1
          Stem-loop Accession # MI0019437
          Stem-loop Sequence
          GACAGCGGCCUGCUUGGGGGACAUACUUCUUUAUAUGCCCAUAUGAACACGAGCAACUAUGGAAUGUAAAGAAGUAUGUAUCCCAGGUUGGGAGAAAUGAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0022558
          miRBase ID: ola-miR-1-3p
          Mature miRNA Sequence: UGGAAUGUAAAGAAGUAUGUA
          Chromosome Location: NA

          Stem-loop ID ola-mir-1-2
          Stem-loop Accession # MI0019525
          Stem-loop Sequence
          ACAAUUACUUCCCUGGGGUACAUACUUCUUUAUGUACCCAUAUGAAGAUAUGAAAGCUAUGGAAUGUAAAGAAGUAUGUAUUCUCGGUGAGGUGCGAGUCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0022558
          miRBase ID: ola-miR-1-3p
          Mature miRNA Sequence: UGGAAUGUAAAGAAGUAUGUA
          Chromosome Location: NA

          Pan paniscus

          Stem-loop ID ppa-mir-1
          Stem-loop Accession # MI0002930
          Stem-loop Sequence
          ACCUACUCAGAGUACAUACUUCUUUAUGUACCCAUAUGAACAUACAAUGCUAUGGAAUGUAAAGAAGUAUGUAUUUUUGGUAGGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002621
          miRBase ID: ppa-miR-1
          Mature miRNA Sequence: UGGAAUGUAAAGAAGUAUGUA
          Chromosome Location: NA

          Pig

          Stem-loop ID ssc-mir-1
          Stem-loop Accession # MI0010682
          Stem-loop Sequence
          CCGGUUGACGUACCUGCUUGGGGGACAUACUUCUUUAUGUGCCCAUAUGGACCUGCUAAGCUAUGGAAUGUAAAGAAGUAUGUAUUUCAGGCUGGGAACUCUCACCGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0010187
          miRBase ID: ssc-miR-1
          Mature miRNA Sequence: UGGAAUGUAAAGAAGUAUGUA
          Chromosome Location: NA

          Python bivittatus

          Stem-loop ID pbv-mir-1a-1
          Stem-loop Accession # MI0030143
          Stem-loop Sequence
          ACCUGCUUGAGUGACAUACUUCUUUAUAUGCCCAUAUGAACCUGGCAAACUAUGGAAUGUAAAGAAGUAUGUAUUUCAGGCAUGGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038813
          miRBase ID: pbv-miR-1a-3p
          Mature miRNA Sequence: UGGAAUGUAAAGAAGUAUGUA
          Chromosome Location: NA

          Stem-loop ID pbv-mir-1a-2
          Stem-loop Accession # MI0030144
          Stem-loop Sequence
          ACCUGCCCAGAGCACAUACUUCUUUAUGUACCCAUAUGAAGAUAUAAUGUUAUGGAAUGUAAAGAAGUAUGUACUUUUGGCAGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038813
          miRBase ID: pbv-miR-1a-3p
          Mature miRNA Sequence: UGGAAUGUAAAGAAGUAUGUA
          Chromosome Location: NA

          Xenopus tropicalis

          Stem-loop ID xtr-mir-1a-1
          Stem-loop Accession # MI0004789
          Stem-loop Sequence
          ACCUGUUUGGAGUACAUACUUCUUUAUGUACCCAUAUGAACAUACAAUGCUAUGGAAUGUAAAGAAGUAUGUAUUUUUAUCAGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0003551
          miRBase ID: xtr-miR-1a
          Mature miRNA Sequence: UGGAAUGUAAAGAAGUAUGUA
          Chromosome Location: NA

          Stem-loop ID xtr-mir-1a-2
          Stem-loop Accession # MI0004788
          Stem-loop Sequence
          UGGGAGACAUACUUCUUUAUAUGUCCAUAUGGAAGUGCCAAUGCUAUGGAAUGUAAAGAAGUAUGUAUCUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0003551
          miRBase ID: xtr-miR-1a
          Mature miRNA Sequence: UGGAAUGUAAAGAAGUAUGUA
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          Ambion® Anti-miR™ miRNA Inhibitor : AM10660
          Ambion® Pre-miR™ miRNA Precursor : PM10660
          TaqMan™ MicroRNA Assay : 000385
          mirVana® miRNA mimic : MC10660


          miRBase Alias:

          Caenorhabditis elegans: cel-miR-1 (v17)
          Atlantibacter subterranea: asu-miR-1 (v18)
          Gallus gallus: gga-miR-1a (v19)
          Monodelphis domestica: mdo-miR-1 (v19)

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline