Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › PM11073
          Assay Name: rno-miR-200b-3p
          miRBase Accession Number: MI0000944
          miRBase Version: v22.1
          Chromosome Location: Chr.5: 173490144 - 173490238 [-] on Build Rnor_6.0
          Mature miRNA Sequence: UAAUACUGCCUGGUAAUGAUGAC
          Species: Rat, Cricetulus griseus, Gorilla gorilla, Guinea pig, Pig, Pteropus alecto
          Product Type: Ambion® Pre-miR™ miRNA Precursor
          Assay ID PM11073
          Size
          Availability Made To Order
          Catalog # AM17100
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000019, mir-8

          Mature miRNA Sequence:

          UAAUACUGCCUGGUAAUGAUGAC

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Rat rno-miR-200b-3p MIMAT0000875
          Cricetulus griseus cgr-miR-200b MIMAT0023842
          Gorilla gorilla ggo-miR-200b MIMAT0024299
          Guinea pig cpo-miR-200b-3p MIMAT0047092
          Pig ssc-miR-200b MIMAT0041606
          Pteropus alecto pal-miR-200b-3p MIMAT0039865

          Stem-loop Details



          Rat

          Stem-loop ID rno-mir-200b
          Stem-loop Accession # MI0000944
          Stem-loop Sequence
          CCAACUUGGGCAGCCGUGGCCAUCUUACUGGGCAGCAUUGGAUAGUGUCUGAUCUCUAAUACUGCCUGGUAAUGAUGACGGCGGAGCCCUGCACG
          Chromosome Location Chr. 5 - 173490144 - 173490238 [-] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0000875
          miRBase ID: rno-miR-200b-3p
          Mature miRNA Sequence: UAAUACUGCCUGGUAAUGAUGAC
          Chromosome Location: Chr. 5 - 173490144 - 173490238 [-] on Build Rnor_6.0

          Cricetulus griseus

          Stem-loop ID cgr-mir-200b
          Stem-loop Accession # MI0020452
          Stem-loop Sequence
          UCCAACUUGGGCAGCCGUGGCCAUCUUACUGGGCAGCAUUGGAUAGUGUCUGAUCUCUAAUACUGCCUGGUAAUGAUGACGGCGGAGCCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0023842
          miRBase ID: cgr-miR-200b
          Mature miRNA Sequence: UAAUACUGCCUGGUAAUGAUGAC
          Chromosome Location: NA

          Gorilla gorilla

          Stem-loop ID ggo-mir-200b
          Stem-loop Accession # MI0020847
          Stem-loop Sequence
          CGGCCCCCGGACCCAGCUCGGGCAGCCGUGGCCAUCUUACUGGGCAGCAUUGGAUGGAGUCAGGUCUCUAAUACUGCCUGGUAAUGAUGACGGCGGAGCCCUGCACGCAGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0024299
          miRBase ID: ggo-miR-200b
          Mature miRNA Sequence: UAAUACUGCCUGGUAAUGAUGAC
          Chromosome Location: NA

          Guinea pig

          Stem-loop ID cpo-mir-200b
          Stem-loop Accession # MI0038693
          Stem-loop Sequence
          CAUCUUACUGGGCAGCAUUGGAUGGUGUCUGGUCUCUAAUACUGCCUGGUAAUGAUGAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047092
          miRBase ID: cpo-miR-200b-3p
          Mature miRNA Sequence: UAAUACUGCCUGGUAAUGAUGAC
          Chromosome Location: NA

          Pig

          Stem-loop ID ssc-mir-200b
          Stem-loop Accession # MI0033407
          Stem-loop Sequence
          CCGCCAUCUUACUGGGCAGCAUUGGAUGGUGUCUGGUCUCUAAUACUGCCUGGUAAUGAUGACGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0041606
          miRBase ID: ssc-miR-200b
          Mature miRNA Sequence: UAAUACUGCCUGGUAAUGAUGAC
          Chromosome Location: NA

          Pteropus alecto

          Stem-loop ID pal-mir-200b
          Stem-loop Accession # MI0032370
          Stem-loop Sequence
          CAUCUUACUGGGCAGCAUUGGAUGGAGUCUGGUCUCUAAUACUGCCUGGUAAUGAUGAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0039865
          miRBase ID: pal-miR-200b-3p
          Mature miRNA Sequence: UAAUACUGCCUGGUAAUGAUGAC
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Rn03464804_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM11073
          TaqMan™ MicroRNA Assay : 001800
          mirVana® miRNA inhibitor : MH11073
          mirVana® miRNA mimic : MC11073
          TaqMan™ Advanced miRNA Assay : rno481286_mir


          miRBase Alias:

          Human: hsa-miR-200b (v9.2)
          Mouse: mmu-miR-200b (v9.2)
          Rat: rno-miR-200b (v18)

          Back To Top

          Related Products

          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline