Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › MC11073
          Assay Name: rno-miR-200b-3p
          miRBase Accession Number: MI0000944
          miRBase Version: v22.1
          Chromosome Location: Chr.5: 173490144 - 173490238 [-] on Build Rnor_6.0
          Mature miRNA Sequence: UAAUACUGCCUGGUAAUGAUGAC
          Species: Rat, Cricetulus griseus, Gorilla gorilla, Guinea pig, Pig, Pteropus alecto
          Product Type: mirVana® miRNA mimic
          Assay ID MC11073

          Purification
          Size
          Availability Made To Order
          Catalog # 4464066
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000019, mir-8

          Mature miRNA Sequence:

          UAAUACUGCCUGGUAAUGAUGAC

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Rat rno-miR-200b-3p MIMAT0000875
          Cricetulus griseus cgr-miR-200b MIMAT0023842
          Gorilla gorilla ggo-miR-200b MIMAT0024299
          Guinea pig cpo-miR-200b-3p MIMAT0047092
          Pig ssc-miR-200b MIMAT0041606
          Pteropus alecto pal-miR-200b-3p MIMAT0039865

          Stem-loop Details



          Rat

          Stem-loop ID rno-mir-200b
          Stem-loop Accession # MI0000944
          Stem-loop Sequence
          CCAACUUGGGCAGCCGUGGCCAUCUUACUGGGCAGCAUUGGAUAGUGUCUGAUCUCUAAUACUGCCUGGUAAUGAUGACGGCGGAGCCCUGCACG
          Chromosome Location Chr. 5 - 173490144 - 173490238 [-] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0000875
          miRBase ID: rno-miR-200b-3p
          Mature miRNA Sequence: UAAUACUGCCUGGUAAUGAUGAC
          Chromosome Location: Chr. 5 - 173490144 - 173490238 [-] on Build Rnor_6.0

          Cricetulus griseus

          Stem-loop ID cgr-mir-200b
          Stem-loop Accession # MI0020452
          Stem-loop Sequence
          UCCAACUUGGGCAGCCGUGGCCAUCUUACUGGGCAGCAUUGGAUAGUGUCUGAUCUCUAAUACUGCCUGGUAAUGAUGACGGCGGAGCCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0023842
          miRBase ID: cgr-miR-200b
          Mature miRNA Sequence: UAAUACUGCCUGGUAAUGAUGAC
          Chromosome Location: NA

          Gorilla gorilla

          Stem-loop ID ggo-mir-200b
          Stem-loop Accession # MI0020847
          Stem-loop Sequence
          CGGCCCCCGGACCCAGCUCGGGCAGCCGUGGCCAUCUUACUGGGCAGCAUUGGAUGGAGUCAGGUCUCUAAUACUGCCUGGUAAUGAUGACGGCGGAGCCCUGCACGCAGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0024299
          miRBase ID: ggo-miR-200b
          Mature miRNA Sequence: UAAUACUGCCUGGUAAUGAUGAC
          Chromosome Location: NA

          Guinea pig

          Stem-loop ID cpo-mir-200b
          Stem-loop Accession # MI0038693
          Stem-loop Sequence
          CAUCUUACUGGGCAGCAUUGGAUGGUGUCUGGUCUCUAAUACUGCCUGGUAAUGAUGAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047092
          miRBase ID: cpo-miR-200b-3p
          Mature miRNA Sequence: UAAUACUGCCUGGUAAUGAUGAC
          Chromosome Location: NA

          Pig

          Stem-loop ID ssc-mir-200b
          Stem-loop Accession # MI0033407
          Stem-loop Sequence
          CCGCCAUCUUACUGGGCAGCAUUGGAUGGUGUCUGGUCUCUAAUACUGCCUGGUAAUGAUGACGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0041606
          miRBase ID: ssc-miR-200b
          Mature miRNA Sequence: UAAUACUGCCUGGUAAUGAUGAC
          Chromosome Location: NA

          Pteropus alecto

          Stem-loop ID pal-mir-200b
          Stem-loop Accession # MI0032370
          Stem-loop Sequence
          CAUCUUACUGGGCAGCAUUGGAUGGAGUCUGGUCUCUAAUACUGCCUGGUAAUGAUGAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0039865
          miRBase ID: pal-miR-200b-3p
          Mature miRNA Sequence: UAAUACUGCCUGGUAAUGAUGAC
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Rn03464804_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM11073
          Ambion® Pre-miR™ miRNA Precursor : PM11073
          TaqMan™ MicroRNA Assay : 001800
          mirVana® miRNA inhibitor : MH11073
          TaqMan™ Advanced miRNA Assay : rno481286_mir


          miRBase Alias:

          Rat: rno-miR-200b (v18)

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline