Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › PM11154
          Assay Name: hsa-miR-124-5p
          miRBase Accession Number: MI0000443
          miRBase Version: v22.1
          Chromosome Location: Chr.8: 9903388 - 9903472 [-] on Build GRCh38
          Mature miRNA Sequence: CGUGUUCACAGCGGACCUUGAU
          Species: Human, Mouse, Rat, Alligator mississippiensis, Columba livia, Dasypus novemcinctus, Gadus morhua, Guinea pig, Ophiophagus hannah, Ornithorhynchus anatinus, Paralichthys olivaceus, Petromyzon marinus, Python bivittatus, Rabbit, Xenopus laevis, Zebrafish
          Product Type: Ambion® Pre-miR™ miRNA Precursor
          Assay ID PM11154
          Size
          Availability Made To Order
          Catalog # AM17100
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000021, mir-124

          Mature miRNA Sequence:

          CGUGUUCACAGCGGACCUUGAU

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-124-5p MIMAT0004591
          Mouse mmu-miR-124-5p MIMAT0004527
          Rat rno-miR-124-5p MIMAT0004728
          Alligator mississippiensis ami-miR-124-5p MIMAT0038125
          Columba livia cli-miR-124-2-5p MIMAT0038495
          Dasypus novemcinctus dno-miR-124-5p MIMAT0047612
          View More Rows
          Gadus morhua gmo-miR-124-5p MIMAT0044327
          Guinea pig cpo-miR-124-5p MIMAT0046963
          Ophiophagus hannah oha-miR-124-5p MIMAT0036669
          Ornithorhynchus anatinus oan-miR-124a-5p MIMAT0007113
          Paralichthys olivaceus pol-miR-124-5p MIMAT0025430
          Petromyzon marinus pma-miR-124-5p MIMAT0019444
          Python bivittatus pbv-miR-124a-5p MIMAT0038913
          Python bivittatus pbv-miR-124b-5p MIMAT0038911
          Rabbit ocu-miR-124-5p MIMAT0048203
          Xenopus laevis xla-miR-124-5p MIMAT0046430
          Zebrafish dre-miR-124-5p MIMAT0031960

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-124-1
          Stem-loop Accession # MI0000443
          Stem-loop Sequence
          AGGCCUCUCUCUCCGUGUUCACAGCGGACCUUGAUUUAAAUGUCCAUACAAUUAAGGCACGCGGUGAAUGCCAAGAAUGGGGCUG
          Chromosome Location Chr. 8 - 9903388 - 9903472 [-] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0004591
          miRBase ID: hsa-miR-124-5p
          Mature miRNA Sequence: CGUGUUCACAGCGGACCUUGAU
          Chromosome Location: Chr. 8 - 9903388 - 9903472 [-] on Build GRCh38

          Stem-loop ID hsa-mir-124-2
          Stem-loop Accession # MI0000444
          Stem-loop Sequence
          AUCAAGAUUAGAGGCUCUGCUCUCCGUGUUCACAGCGGACCUUGAUUUAAUGUCAUACAAUUAAGGCACGCGGUGAAUGCCAAGAGCGGAGCCUACGGCUGCACUUGAA
          Chromosome Location Chr. 8 - 64379149 - 64379257 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0004591
          miRBase ID: hsa-miR-124-5p
          Mature miRNA Sequence: CGUGUUCACAGCGGACCUUGAU
          Chromosome Location: Chr. 8 - 9903388 - 9903472 [-] on Build GRCh38

          Stem-loop ID hsa-mir-124-3
          Stem-loop Accession # MI0000445
          Stem-loop Sequence
          UGAGGGCCCCUCUGCGUGUUCACAGCGGACCUUGAUUUAAUGUCUAUACAAUUAAGGCACGCGGUGAAUGCCAAGAGAGGCGCCUCC
          Chromosome Location Chr. 20 - 63178500 - 63178586 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0004591
          miRBase ID: hsa-miR-124-5p
          Mature miRNA Sequence: CGUGUUCACAGCGGACCUUGAU
          Chromosome Location: Chr. 8 - 9903388 - 9903472 [-] on Build GRCh38

          Mouse

          Stem-loop ID mmu-mir-124-1
          Stem-loop Accession # MI0000716
          Stem-loop Sequence
          AGGCCUCUCUCUCCGUGUUCACAGCGGACCUUGAUUUAAAUGUCCAUACAAUUAAGGCACGCGGUGAAUGCCAAGAAUGGGGCUG
          Chromosome Location Chr. 14 - 64590657 - 64590741 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0004527
          miRBase ID: mmu-miR-124-5p
          Mature miRNA Sequence: CGUGUUCACAGCGGACCUUGAU
          Chromosome Location: Chr. 14 - 64590657 - 64590741 [+] on Build GRCm38

          Stem-loop ID mmu-mir-124-2
          Stem-loop Accession # MI0000717
          Stem-loop Sequence
          AUCAAGAUCAGAGACUCUGCUCUCCGUGUUCACAGCGGACCUUGAUUUAAUGUCAUACAAUUAAGGCACGCGGUGAAUGCCAAGAGCGGAGCCUACGGCUGCACUUGAA
          Chromosome Location Chr. 3 - 17795662 - 17795770 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0004527
          miRBase ID: mmu-miR-124-5p
          Mature miRNA Sequence: CGUGUUCACAGCGGACCUUGAU
          Chromosome Location: Chr. 14 - 64590657 - 64590741 [+] on Build GRCm38

          Stem-loop ID mmu-mir-124-3
          Stem-loop Accession # MI0000150
          Stem-loop Sequence
          CUCUGCGUGUUCACAGCGGACCUUGAUUUAAUGUCUAUACAAUUAAGGCACGCGGUGAAUGCCAAGAG
          Chromosome Location Chr. 2 - 180894040 - 180894107 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0004527
          miRBase ID: mmu-miR-124-5p
          Mature miRNA Sequence: CGUGUUCACAGCGGACCUUGAU
          Chromosome Location: Chr. 14 - 64590657 - 64590741 [+] on Build GRCm38

          Rat

          Stem-loop ID rno-mir-124-1
          Stem-loop Accession # MI0000893
          Stem-loop Sequence
          AGGCCUCUCUCUCCGUGUUCACAGCGGACCUUGAUUUAAAUGUCCAUACAAUUAAGGCACGCGGUGAAUGCCAAGAAUGGGGCUG
          Chromosome Location Chr. 15 - 47964298 - 47964382 [+] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0004728
          miRBase ID: rno-miR-124-5p
          Mature miRNA Sequence: CGUGUUCACAGCGGACCUUGAU
          Chromosome Location: Chr. 2 - 102311603 - 102311711 [+] on Build Rnor_6.0

          Stem-loop ID rno-mir-124-2
          Stem-loop Accession # MI0000894
          Stem-loop Sequence
          AUCAAGAUCAGAGACUCUGCUCUCCGUGUUCACAGCGGACCUUGAUUUAAUGUCAUACAAUUAAGGCACGCGGUGAAUGCCAAGAGCGGAGCCUACGGCUGCACUUGAA
          Chromosome Location Chr. 2 - 102311603 - 102311711 [+] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0004728
          miRBase ID: rno-miR-124-5p
          Mature miRNA Sequence: CGUGUUCACAGCGGACCUUGAU
          Chromosome Location: Chr. 2 - 102311603 - 102311711 [+] on Build Rnor_6.0

          Stem-loop ID rno-mir-124-3
          Stem-loop Accession # MI0000892
          Stem-loop Sequence
          UGAGGGCCCCUCUGCGUGUUCACAGCGGACCUUGAUUUAAUGUCUAUACAAUUAAGGCACGCGGUGAAUGCCAAGAGAGGCGCCUCC
          Chromosome Location Chr. 3 - 176406205 - 176406291 [+] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0004728
          miRBase ID: rno-miR-124-5p
          Mature miRNA Sequence: CGUGUUCACAGCGGACCUUGAU
          Chromosome Location: Chr. 2 - 102311603 - 102311711 [+] on Build Rnor_6.0

          Alligator mississippiensis

          Stem-loop ID ami-mir-124-1
          Stem-loop Accession # MI0029717
          Stem-loop Sequence
          GGCCCCGCGGCCCCUCUCUGCGUGUUCACAGCGGACCUUGAUUUAAAUGUCCAUACAAUUAAGGCACGCGGUGAAUGCCAAGAACGGGGCCGCCUCGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038125
          miRBase ID: ami-miR-124-5p
          Mature miRNA Sequence: CGUGUUCACAGCGGACCUUGAU
          Chromosome Location: NA

          Stem-loop ID ami-mir-124-2
          Stem-loop Accession # MI0029718
          Stem-loop Sequence
          UUCCCGGGGCUCUCGCCUCUCCGUGUUCACAGCGGACCUUGAUUUAAUGUCAUACAAUUAAGGCACGCGGUGAAUGCCAAGAGCGGAGCCGGAAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038125
          miRBase ID: ami-miR-124-5p
          Mature miRNA Sequence: CGUGUUCACAGCGGACCUUGAU
          Chromosome Location: NA

          Stem-loop ID ami-mir-124-3
          Stem-loop Accession # MI0029719
          Stem-loop Sequence
          CCAGGCGAGCCCCUCUCUGCGUGUUCACAGCGGACCUUGAUUUAAUGUCCAUACAAUUAAGGCACGCGGUGAAUGCCAAGAGAGGAGCCUCACAGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038125
          miRBase ID: ami-miR-124-5p
          Mature miRNA Sequence: CGUGUUCACAGCGGACCUUGAU
          Chromosome Location: NA

          Stem-loop ID ami-mir-124-4
          Stem-loop Accession # MI0029720
          Stem-loop Sequence
          UCCGGACCGCGGCCUCGGCUCUCCGUGUUCACAGCGGACCUUGAUUUAAUGUCACACAAUUAAGGCACGCGGUGAAUGCCAAGAGAGGGGCCGCCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038125
          miRBase ID: ami-miR-124-5p
          Mature miRNA Sequence: CGUGUUCACAGCGGACCUUGAU
          Chromosome Location: NA

          Columba livia

          Stem-loop ID cli-mir-124-2
          Stem-loop Accession # MI0029955
          Stem-loop Sequence
          CCCGAGCCCCUCUCUGCGUGUUCACAGCGGACCUUGAUUUAAUGUCCAUACAAUUAAGGCACGCGGUGAAUGCCAAGAGAGGAGCCUCACA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038495
          miRBase ID: cli-miR-124-2-5p
          Mature miRNA Sequence: CGUGUUCACAGCGGACCUUGAU
          Chromosome Location: NA

          Dasypus novemcinctus

          Stem-loop ID dno-mir-124-1
          Stem-loop Accession # MI0038964
          Stem-loop Sequence
          CGUGUUCACAGCGGACCUUGAUUUAAUGUCAUACAAUUAAGGCACGCGGUGAAUGCCAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047612
          miRBase ID: dno-miR-124-5p
          Mature miRNA Sequence: CGUGUUCACAGCGGACCUUGAU
          Chromosome Location: NA

          Stem-loop ID dno-mir-124-2
          Stem-loop Accession # MI0038965
          Stem-loop Sequence
          CGUGUUCACAGCGGACCUUGAUUUAAUGUCUACACAAUUAAGGCACGCGGUGAAUGCCAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047612
          miRBase ID: dno-miR-124-5p
          Mature miRNA Sequence: CGUGUUCACAGCGGACCUUGAU
          Chromosome Location: NA

          Gadus morhua

          Stem-loop ID gmo-mir-124-2
          Stem-loop Accession # MI0036222
          Stem-loop Sequence
          CGUGUUCACAGCGGACCUUGAUUUAAUGUCUUACAAUUAAGGCACGCGGUGAAUGCCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0044327
          miRBase ID: gmo-miR-124-5p
          Mature miRNA Sequence: CGUGUUCACAGCGGACCUUGAU
          Chromosome Location: NA

          Guinea pig

          Stem-loop ID cpo-mir-124-1
          Stem-loop Accession # MI0038622
          Stem-loop Sequence
          CGUGUUCACAGCGGACCUUGAUUUAAAUGUCCAUACAAUUAAGGCACGCGGUGAAUGCCAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0046963
          miRBase ID: cpo-miR-124-5p
          Mature miRNA Sequence: CGUGUUCACAGCGGACCUUGAU
          Chromosome Location: NA

          Stem-loop ID cpo-mir-124-2
          Stem-loop Accession # MI0038623
          Stem-loop Sequence
          CGUGUUCACAGCGGACCUUGAUUUAAUGUCAUACAAUUAAGGCACGCGGUGAAUGCCAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0046963
          miRBase ID: cpo-miR-124-5p
          Mature miRNA Sequence: CGUGUUCACAGCGGACCUUGAU
          Chromosome Location: NA

          Ophiophagus hannah

          Stem-loop ID oha-mir-124-2
          Stem-loop Accession # MI0031324
          Stem-loop Sequence
          CCUUCUCUCCGUGUUCACAGCGGACCUUGAUUUAAUGUCCAUACAAUUAAGGCACGCGGUGAAUGCCAAGAGAUCGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036669
          miRBase ID: oha-miR-124-5p
          Mature miRNA Sequence: CGUGUUCACAGCGGACCUUGAU
          Chromosome Location: NA

          Stem-loop ID oha-mir-124-3
          Stem-loop Accession # MI0031326
          Stem-loop Sequence
          AAGCCCUUCGUGUUCACAGCGGACCUUGAUUUAAUUUCGAUACAAUUAAGGCACGCGGUGAAUGCCAAGAGAGAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036669
          miRBase ID: oha-miR-124-5p
          Mature miRNA Sequence: CGUGUUCACAGCGGACCUUGAU
          Chromosome Location: NA

          Stem-loop ID oha-mir-124-4
          Stem-loop Accession # MI0031325
          Stem-loop Sequence
          CCCUCUCUGCGUGUUCACAGCGGACCUUGAUUUAAAUGUCCAUACAAUUAAGGCACGCGGUGAAUGCUAAGAACGAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036669
          miRBase ID: oha-miR-124-5p
          Mature miRNA Sequence: CGUGUUCACAGCGGACCUUGAU
          Chromosome Location: NA

          Ornithorhynchus anatinus

          Stem-loop ID oan-mir-124a-1
          Stem-loop Accession # MI0006854
          Stem-loop Sequence
          CACUAGGCCUGUCUCUCCGUGUUCACAGCGGACCUUGAUUUAAAUUUGUACACAAUUAAGGCACGCGGUGAAUGCCAAGAGCCCAGCCAAAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0007113
          miRBase ID: oan-miR-124a-5p
          Mature miRNA Sequence: CGUGUUCACAGCGGACCUUGAU
          Chromosome Location: NA

          Stem-loop ID oan-mir-124a-2
          Stem-loop Accession # MI0024076
          Stem-loop Sequence
          CGUGUUCACAGCGGACCUUGAUUUAAUGUCAUACAAUUAAGGGCACUCGGUGAAUGCCAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0007113
          miRBase ID: oan-miR-124a-5p
          Mature miRNA Sequence: CGUGUUCACAGCGGACCUUGAU
          Chromosome Location: NA

          Paralichthys olivaceus

          Stem-loop ID pol-mir-124
          Stem-loop Accession # MI0022198
          Stem-loop Sequence
          CGUGUUCACAGCGGACCUUGAUUUAAUGUCCAUACAAUUAAGGCACGCGGUGAAUGCCAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0025430
          miRBase ID: pol-miR-124-5p
          Mature miRNA Sequence: CGUGUUCACAGCGGACCUUGAU
          Chromosome Location: NA

          Petromyzon marinus

          Stem-loop ID pma-mir-124
          Stem-loop Accession # MI0017057
          Stem-loop Sequence
          CGUCCGCUCUUCGUGUUCACAGCGGACCUUGAUUUAAUGUGCUACAAUUAAGGCACGCGGUGAAUGCCAAGAGCGGUGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0019444
          miRBase ID: pma-miR-124-5p
          Mature miRNA Sequence: CGUGUUCACAGCGGACCUUGAU
          Chromosome Location: NA

          Python bivittatus

          Stem-loop ID pbv-mir-124a-1
          Stem-loop Accession # MI0030212
          Stem-loop Sequence
          CGAGGCUCUGUCUCUCCGUGUUCACAGCGGACCUUGAUUUAAUGUCAUACAAUUAAGGCACGCGGUGAAUGCCAAGAGCAGGGCUUGAAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038913
          miRBase ID: pbv-miR-124a-5p
          Mature miRNA Sequence: CGUGUUCACAGCGGACCUUGAU
          Chromosome Location: NA

          Stem-loop ID pbv-mir-124a-2
          Stem-loop Accession # MI0030213
          Stem-loop Sequence
          CCUGGGCCCUUCUCUCCGUGUUCACAGCGGACCUUGAUUUAAUGUCCAUACAAUUAAGGCACGCGGUGAAUGCCAAGAGACCGGUUCCCCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038913
          miRBase ID: pbv-miR-124a-5p
          Mature miRNA Sequence: CGUGUUCACAGCGGACCUUGAU
          Chromosome Location: NA

          Stem-loop ID pbv-mir-124a-3
          Stem-loop Accession # MI0030214
          Stem-loop Sequence
          CUUGGCUUAAGCCCUUCGUGUUCACAGCGGACCUUGAUUUAAUUUCGAUACAAUUAAGGCACGCGGUGAAUGCCAAGAGAGAAGCCACGUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038913
          miRBase ID: pbv-miR-124a-5p
          Mature miRNA Sequence: CGUGUUCACAGCGGACCUUGAU
          Chromosome Location: NA

          Stem-loop ID pbv-mir-124b
          Stem-loop Accession # MI0030211
          Stem-loop Sequence
          CAAAGGCCCCUCUCUGCGUGUUCACAGCGGACCUUGAUUUAAAUGUCCAUACAAUUAAGGCACGCGGUGAAUGCUAAGAACGAGGCUGGAAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038911
          miRBase ID: pbv-miR-124b-5p
          Mature miRNA Sequence: CGUGUUCACAGCGGACCUUGAU
          Chromosome Location: NA

          Rabbit

          Stem-loop ID ocu-mir-124-1
          Stem-loop Accession # MI0039276
          Stem-loop Sequence
          CGUGUUCACAGCGGACCUUGAUUUAAAUGUCCAUACAAUUAAGGCACGCGGUGAAUGCCAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0048203
          miRBase ID: ocu-miR-124-5p
          Mature miRNA Sequence: CGUGUUCACAGCGGACCUUGAU
          Chromosome Location: NA

          Stem-loop ID ocu-mir-124-2
          Stem-loop Accession # MI0039277
          Stem-loop Sequence
          CGUGUUCACAGCGGACCUUGAUUUAAUGUCAUACAAUUAAGGCACGCGGUGAAUGCCAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0048203
          miRBase ID: ocu-miR-124-5p
          Mature miRNA Sequence: CGUGUUCACAGCGGACCUUGAU
          Chromosome Location: NA

          Xenopus laevis

          Stem-loop ID xla-mir-124-1
          Stem-loop Accession # MI0038237
          Stem-loop Sequence
          CGUGUUCACAGCGGACCUUGAUUUAAAUGUCCAUACAAUUAAGGCACGCGGUGAAUGCCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0046430
          miRBase ID: xla-miR-124-5p
          Mature miRNA Sequence: CGUGUUCACAGCGGACCUUGAU
          Chromosome Location: NA

          Zebrafish

          Stem-loop ID dre-mir-124-2
          Stem-loop Accession # MI0001967
          Stem-loop Sequence
          CCUGCUUUUCUUCGUGUUCACAGCGGACCUUGAUUUAAAUGUCCAUACAAUUAAGGCACGCGGUGAAUGCCAAGAGAGAUGGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0031960
          miRBase ID: dre-miR-124-5p
          Mature miRNA Sequence: CGUGUUCACAGCGGACCUUGAU
          Chromosome Location: NA

          Stem-loop ID dre-mir-124-3
          Stem-loop Accession # MI0001968
          Stem-loop Sequence
          GGCUCUGUGGGAUUUCAGACUCUGGCUUUCCGUGUUCACAGCGGACCUUGAUUUAAUGUCUUACAAUUAAGGCACGCGGUGAAUGCCAAGAGCGGAGCCUUUUAACAUCAGCAGGCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0031960
          miRBase ID: dre-miR-124-5p
          Mature miRNA Sequence: CGUGUUCACAGCGGACCUUGAU
          Chromosome Location: NA

          Stem-loop ID dre-mir-124-5
          Stem-loop Accession # MI0001970
          Stem-loop Sequence
          GGGUUUUGCUCGUGCGUUCUUUUUGAGUUCUCGCUCUGCGUGUUCACAGCGGACCUUGAUUUAAUGUCCAUACAAUUAAGGCACGCGGUGAAUGCCAAGAGAAGAAUCUCUCCAGCAACGAGUUUGCGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0031960
          miRBase ID: dre-miR-124-5p
          Mature miRNA Sequence: CGUGUUCACAGCGGACCUUGAU
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Mm03306225_pri
          TaqMan™ Pri-miRNA Assay : Rn03465782_pri
          TaqMan™ Pri-miRNA Assay : Mm03307226_pri
          TaqMan™ Pri-miRNA Assay : Hs03303081_pri
          TaqMan™ Pri-miRNA Assay : Rn03464574_pri
          TaqMan™ Pri-miRNA Assay : Rn03464567_pri
          TaqMan™ Pri-miRNA Assay : Mm03307218_pri
          TaqMan™ Pri-miRNA Assay : Hs03303077_pri
          TaqMan™ Pri-miRNA Assay : Hs03303090_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM11154
          TaqMan™ MicroRNA Assay : 002197
          mirVana® miRNA inhibitor : MH11154
          mirVana® miRNA mimic : MC11154
          TaqMan™ Advanced miRNA Assay : rno480902_mir
          TaqMan™ Advanced miRNA Assay : mmu480902_mir
          TaqMan™ Advanced miRNA Assay : 477880_mir


          miRBase Alias:

          Human: hsa-miR-124* (v17)
          Mouse: mmu-miR-124* (v17)
          Rat: rno-miR-124* (v19)
          Ochrobactrum anthropi: oan-miR-124-5p (v19)
          Ochrobactrum anthropi: oan-miR-124* (v18)
          Phocea massiliensis: pma-miR-124* (v18)

          Back To Top

          Related Products

          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline