Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › mmu480902_mir

          Specificity testing was performed using mouse anti-targets.

          Assay Name: mmu-miR-124-5p
          Stem-loop Accession Number: MI0000443
          miRBase Version: v22.1
          Chromosome Location: Chr.8: 9903388 - 9903472 [-] on Build GRCh38
          Mature miRNA Sequence: CGUGUUCACAGCGGACCUUGAU
          Species: Human, Mouse, Rat, Alligator mississippiensis, Columba livia, Dasypus novemcinctus, Gadus morhua, Guinea pig, Ophiophagus hannah, Ornithorhynchus anatinus, Paralichthys olivaceus, Petromyzon marinus, Python bivittatus, Rabbit, Xenopus laevis, Zebrafish
          Product Type: TaqMan™ Advanced miRNA Assay
          Assay ID mmu480902_mir
          Size S: 250 rxns
          Availability Inventoried
          Catalog # A25576
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000021, mir-124

          Mature miRNA Sequence:

          CGUGUUCACAGCGGACCUUGAU

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-124-5p MIMAT0004591
          Mouse mmu-miR-124-5p MIMAT0004527
          Rat rno-miR-124-5p MIMAT0004728
          Alligator mississippiensis ami-miR-124-5p MIMAT0038125
          Columba livia cli-miR-124-2-5p MIMAT0038495
          Dasypus novemcinctus dno-miR-124-5p MIMAT0047612
          View More Rows
          Gadus morhua gmo-miR-124-5p MIMAT0044327
          Guinea pig cpo-miR-124-5p MIMAT0046963
          Ophiophagus hannah oha-miR-124-5p MIMAT0036669
          Ornithorhynchus anatinus oan-miR-124a-5p MIMAT0007113
          Paralichthys olivaceus pol-miR-124-5p MIMAT0025430
          Petromyzon marinus pma-miR-124-5p MIMAT0019444
          Python bivittatus pbv-miR-124a-5p MIMAT0038913
          Python bivittatus pbv-miR-124b-5p MIMAT0038911
          Rabbit ocu-miR-124-5p MIMAT0048203
          Xenopus laevis xla-miR-124-5p MIMAT0046430
          Zebrafish dre-miR-124-5p MIMAT0031960

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-124-1
          Stem-loop Accession # MI0000443
          Stem-loop Sequence
          AGGCCUCUCUCUCCGUGUUCACAGCGGACCUUGAUUUAAAUGUCCAUACAAUUAAGGCACGCGGUGAAUGCCAAGAAUGGGGCUG
          Chromosome Location Chr. 8 - 9903388 - 9903472 [-] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0004591
          miRBase ID: hsa-miR-124-5p
          Mature miRNA Sequence: CGUGUUCACAGCGGACCUUGAU
          Chromosome Location: Chr. 8 - 9903388 - 9903472 [-] on Build GRCh38

          Stem-loop ID hsa-mir-124-2
          Stem-loop Accession # MI0000444
          Stem-loop Sequence
          AUCAAGAUUAGAGGCUCUGCUCUCCGUGUUCACAGCGGACCUUGAUUUAAUGUCAUACAAUUAAGGCACGCGGUGAAUGCCAAGAGCGGAGCCUACGGCUGCACUUGAA
          Chromosome Location Chr. 8 - 64379149 - 64379257 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0004591
          miRBase ID: hsa-miR-124-5p
          Mature miRNA Sequence: CGUGUUCACAGCGGACCUUGAU
          Chromosome Location: Chr. 8 - 9903388 - 9903472 [-] on Build GRCh38

          Stem-loop ID hsa-mir-124-3
          Stem-loop Accession # MI0000445
          Stem-loop Sequence
          UGAGGGCCCCUCUGCGUGUUCACAGCGGACCUUGAUUUAAUGUCUAUACAAUUAAGGCACGCGGUGAAUGCCAAGAGAGGCGCCUCC
          Chromosome Location Chr. 20 - 63178500 - 63178586 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0004591
          miRBase ID: hsa-miR-124-5p
          Mature miRNA Sequence: CGUGUUCACAGCGGACCUUGAU
          Chromosome Location: Chr. 8 - 9903388 - 9903472 [-] on Build GRCh38

          Mouse

          Stem-loop ID mmu-mir-124-1
          Stem-loop Accession # MI0000716
          Stem-loop Sequence
          AGGCCUCUCUCUCCGUGUUCACAGCGGACCUUGAUUUAAAUGUCCAUACAAUUAAGGCACGCGGUGAAUGCCAAGAAUGGGGCUG
          Chromosome Location Chr. 14 - 64590657 - 64590741 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0004527
          miRBase ID: mmu-miR-124-5p
          Mature miRNA Sequence: CGUGUUCACAGCGGACCUUGAU
          Chromosome Location: Chr. 14 - 64590657 - 64590741 [+] on Build GRCm38

          Stem-loop ID mmu-mir-124-2
          Stem-loop Accession # MI0000717
          Stem-loop Sequence
          AUCAAGAUCAGAGACUCUGCUCUCCGUGUUCACAGCGGACCUUGAUUUAAUGUCAUACAAUUAAGGCACGCGGUGAAUGCCAAGAGCGGAGCCUACGGCUGCACUUGAA
          Chromosome Location Chr. 3 - 17795662 - 17795770 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0004527
          miRBase ID: mmu-miR-124-5p
          Mature miRNA Sequence: CGUGUUCACAGCGGACCUUGAU
          Chromosome Location: Chr. 14 - 64590657 - 64590741 [+] on Build GRCm38

          Stem-loop ID mmu-mir-124-3
          Stem-loop Accession # MI0000150
          Stem-loop Sequence
          CUCUGCGUGUUCACAGCGGACCUUGAUUUAAUGUCUAUACAAUUAAGGCACGCGGUGAAUGCCAAGAG
          Chromosome Location Chr. 2 - 180894040 - 180894107 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0004527
          miRBase ID: mmu-miR-124-5p
          Mature miRNA Sequence: CGUGUUCACAGCGGACCUUGAU
          Chromosome Location: Chr. 14 - 64590657 - 64590741 [+] on Build GRCm38

          Rat

          Stem-loop ID rno-mir-124-1
          Stem-loop Accession # MI0000893
          Stem-loop Sequence
          AGGCCUCUCUCUCCGUGUUCACAGCGGACCUUGAUUUAAAUGUCCAUACAAUUAAGGCACGCGGUGAAUGCCAAGAAUGGGGCUG
          Chromosome Location Chr. 15 - 47964298 - 47964382 [+] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0004728
          miRBase ID: rno-miR-124-5p
          Mature miRNA Sequence: CGUGUUCACAGCGGACCUUGAU
          Chromosome Location: Chr. 2 - 102311603 - 102311711 [+] on Build Rnor_6.0

          Stem-loop ID rno-mir-124-2
          Stem-loop Accession # MI0000894
          Stem-loop Sequence
          AUCAAGAUCAGAGACUCUGCUCUCCGUGUUCACAGCGGACCUUGAUUUAAUGUCAUACAAUUAAGGCACGCGGUGAAUGCCAAGAGCGGAGCCUACGGCUGCACUUGAA
          Chromosome Location Chr. 2 - 102311603 - 102311711 [+] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0004728
          miRBase ID: rno-miR-124-5p
          Mature miRNA Sequence: CGUGUUCACAGCGGACCUUGAU
          Chromosome Location: Chr. 2 - 102311603 - 102311711 [+] on Build Rnor_6.0

          Stem-loop ID rno-mir-124-3
          Stem-loop Accession # MI0000892
          Stem-loop Sequence
          UGAGGGCCCCUCUGCGUGUUCACAGCGGACCUUGAUUUAAUGUCUAUACAAUUAAGGCACGCGGUGAAUGCCAAGAGAGGCGCCUCC
          Chromosome Location Chr. 3 - 176406205 - 176406291 [+] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0004728
          miRBase ID: rno-miR-124-5p
          Mature miRNA Sequence: CGUGUUCACAGCGGACCUUGAU
          Chromosome Location: Chr. 2 - 102311603 - 102311711 [+] on Build Rnor_6.0

          Alligator mississippiensis

          Stem-loop ID ami-mir-124-1
          Stem-loop Accession # MI0029717
          Stem-loop Sequence
          GGCCCCGCGGCCCCUCUCUGCGUGUUCACAGCGGACCUUGAUUUAAAUGUCCAUACAAUUAAGGCACGCGGUGAAUGCCAAGAACGGGGCCGCCUCGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038125
          miRBase ID: ami-miR-124-5p
          Mature miRNA Sequence: CGUGUUCACAGCGGACCUUGAU
          Chromosome Location: NA

          Stem-loop ID ami-mir-124-2
          Stem-loop Accession # MI0029718
          Stem-loop Sequence
          UUCCCGGGGCUCUCGCCUCUCCGUGUUCACAGCGGACCUUGAUUUAAUGUCAUACAAUUAAGGCACGCGGUGAAUGCCAAGAGCGGAGCCGGAAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038125
          miRBase ID: ami-miR-124-5p
          Mature miRNA Sequence: CGUGUUCACAGCGGACCUUGAU
          Chromosome Location: NA

          Stem-loop ID ami-mir-124-3
          Stem-loop Accession # MI0029719
          Stem-loop Sequence
          CCAGGCGAGCCCCUCUCUGCGUGUUCACAGCGGACCUUGAUUUAAUGUCCAUACAAUUAAGGCACGCGGUGAAUGCCAAGAGAGGAGCCUCACAGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038125
          miRBase ID: ami-miR-124-5p
          Mature miRNA Sequence: CGUGUUCACAGCGGACCUUGAU
          Chromosome Location: NA

          Stem-loop ID ami-mir-124-4
          Stem-loop Accession # MI0029720
          Stem-loop Sequence
          UCCGGACCGCGGCCUCGGCUCUCCGUGUUCACAGCGGACCUUGAUUUAAUGUCACACAAUUAAGGCACGCGGUGAAUGCCAAGAGAGGGGCCGCCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038125
          miRBase ID: ami-miR-124-5p
          Mature miRNA Sequence: CGUGUUCACAGCGGACCUUGAU
          Chromosome Location: NA

          Columba livia

          Stem-loop ID cli-mir-124-2
          Stem-loop Accession # MI0029955
          Stem-loop Sequence
          CCCGAGCCCCUCUCUGCGUGUUCACAGCGGACCUUGAUUUAAUGUCCAUACAAUUAAGGCACGCGGUGAAUGCCAAGAGAGGAGCCUCACA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038495
          miRBase ID: cli-miR-124-2-5p
          Mature miRNA Sequence: CGUGUUCACAGCGGACCUUGAU
          Chromosome Location: NA

          Dasypus novemcinctus

          Stem-loop ID dno-mir-124-1
          Stem-loop Accession # MI0038964
          Stem-loop Sequence
          CGUGUUCACAGCGGACCUUGAUUUAAUGUCAUACAAUUAAGGCACGCGGUGAAUGCCAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047612
          miRBase ID: dno-miR-124-5p
          Mature miRNA Sequence: CGUGUUCACAGCGGACCUUGAU
          Chromosome Location: NA

          Stem-loop ID dno-mir-124-2
          Stem-loop Accession # MI0038965
          Stem-loop Sequence
          CGUGUUCACAGCGGACCUUGAUUUAAUGUCUACACAAUUAAGGCACGCGGUGAAUGCCAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047612
          miRBase ID: dno-miR-124-5p
          Mature miRNA Sequence: CGUGUUCACAGCGGACCUUGAU
          Chromosome Location: NA

          Gadus morhua

          Stem-loop ID gmo-mir-124-2
          Stem-loop Accession # MI0036222
          Stem-loop Sequence
          CGUGUUCACAGCGGACCUUGAUUUAAUGUCUUACAAUUAAGGCACGCGGUGAAUGCCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0044327
          miRBase ID: gmo-miR-124-5p
          Mature miRNA Sequence: CGUGUUCACAGCGGACCUUGAU
          Chromosome Location: NA

          Guinea pig

          Stem-loop ID cpo-mir-124-1
          Stem-loop Accession # MI0038622
          Stem-loop Sequence
          CGUGUUCACAGCGGACCUUGAUUUAAAUGUCCAUACAAUUAAGGCACGCGGUGAAUGCCAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0046963
          miRBase ID: cpo-miR-124-5p
          Mature miRNA Sequence: CGUGUUCACAGCGGACCUUGAU
          Chromosome Location: NA

          Stem-loop ID cpo-mir-124-2
          Stem-loop Accession # MI0038623
          Stem-loop Sequence
          CGUGUUCACAGCGGACCUUGAUUUAAUGUCAUACAAUUAAGGCACGCGGUGAAUGCCAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0046963
          miRBase ID: cpo-miR-124-5p
          Mature miRNA Sequence: CGUGUUCACAGCGGACCUUGAU
          Chromosome Location: NA

          Ophiophagus hannah

          Stem-loop ID oha-mir-124-2
          Stem-loop Accession # MI0031324
          Stem-loop Sequence
          CCUUCUCUCCGUGUUCACAGCGGACCUUGAUUUAAUGUCCAUACAAUUAAGGCACGCGGUGAAUGCCAAGAGAUCGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036669
          miRBase ID: oha-miR-124-5p
          Mature miRNA Sequence: CGUGUUCACAGCGGACCUUGAU
          Chromosome Location: NA

          Stem-loop ID oha-mir-124-3
          Stem-loop Accession # MI0031326
          Stem-loop Sequence
          AAGCCCUUCGUGUUCACAGCGGACCUUGAUUUAAUUUCGAUACAAUUAAGGCACGCGGUGAAUGCCAAGAGAGAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036669
          miRBase ID: oha-miR-124-5p
          Mature miRNA Sequence: CGUGUUCACAGCGGACCUUGAU
          Chromosome Location: NA

          Stem-loop ID oha-mir-124-4
          Stem-loop Accession # MI0031325
          Stem-loop Sequence
          CCCUCUCUGCGUGUUCACAGCGGACCUUGAUUUAAAUGUCCAUACAAUUAAGGCACGCGGUGAAUGCUAAGAACGAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036669
          miRBase ID: oha-miR-124-5p
          Mature miRNA Sequence: CGUGUUCACAGCGGACCUUGAU
          Chromosome Location: NA

          Ornithorhynchus anatinus

          Stem-loop ID oan-mir-124a-1
          Stem-loop Accession # MI0006854
          Stem-loop Sequence
          CACUAGGCCUGUCUCUCCGUGUUCACAGCGGACCUUGAUUUAAAUUUGUACACAAUUAAGGCACGCGGUGAAUGCCAAGAGCCCAGCCAAAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0007113
          miRBase ID: oan-miR-124a-5p
          Mature miRNA Sequence: CGUGUUCACAGCGGACCUUGAU
          Chromosome Location: NA

          Stem-loop ID oan-mir-124a-2
          Stem-loop Accession # MI0024076
          Stem-loop Sequence
          CGUGUUCACAGCGGACCUUGAUUUAAUGUCAUACAAUUAAGGGCACUCGGUGAAUGCCAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0007113
          miRBase ID: oan-miR-124a-5p
          Mature miRNA Sequence: CGUGUUCACAGCGGACCUUGAU
          Chromosome Location: NA

          Paralichthys olivaceus

          Stem-loop ID pol-mir-124
          Stem-loop Accession # MI0022198
          Stem-loop Sequence
          CGUGUUCACAGCGGACCUUGAUUUAAUGUCCAUACAAUUAAGGCACGCGGUGAAUGCCAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0025430
          miRBase ID: pol-miR-124-5p
          Mature miRNA Sequence: CGUGUUCACAGCGGACCUUGAU
          Chromosome Location: NA

          Petromyzon marinus

          Stem-loop ID pma-mir-124
          Stem-loop Accession # MI0017057
          Stem-loop Sequence
          CGUCCGCUCUUCGUGUUCACAGCGGACCUUGAUUUAAUGUGCUACAAUUAAGGCACGCGGUGAAUGCCAAGAGCGGUGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0019444
          miRBase ID: pma-miR-124-5p
          Mature miRNA Sequence: CGUGUUCACAGCGGACCUUGAU
          Chromosome Location: NA

          Python bivittatus

          Stem-loop ID pbv-mir-124a-1
          Stem-loop Accession # MI0030212
          Stem-loop Sequence
          CGAGGCUCUGUCUCUCCGUGUUCACAGCGGACCUUGAUUUAAUGUCAUACAAUUAAGGCACGCGGUGAAUGCCAAGAGCAGGGCUUGAAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038913
          miRBase ID: pbv-miR-124a-5p
          Mature miRNA Sequence: CGUGUUCACAGCGGACCUUGAU
          Chromosome Location: NA

          Stem-loop ID pbv-mir-124a-2
          Stem-loop Accession # MI0030213
          Stem-loop Sequence
          CCUGGGCCCUUCUCUCCGUGUUCACAGCGGACCUUGAUUUAAUGUCCAUACAAUUAAGGCACGCGGUGAAUGCCAAGAGACCGGUUCCCCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038913
          miRBase ID: pbv-miR-124a-5p
          Mature miRNA Sequence: CGUGUUCACAGCGGACCUUGAU
          Chromosome Location: NA

          Stem-loop ID pbv-mir-124a-3
          Stem-loop Accession # MI0030214
          Stem-loop Sequence
          CUUGGCUUAAGCCCUUCGUGUUCACAGCGGACCUUGAUUUAAUUUCGAUACAAUUAAGGCACGCGGUGAAUGCCAAGAGAGAAGCCACGUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038913
          miRBase ID: pbv-miR-124a-5p
          Mature miRNA Sequence: CGUGUUCACAGCGGACCUUGAU
          Chromosome Location: NA

          Stem-loop ID pbv-mir-124b
          Stem-loop Accession # MI0030211
          Stem-loop Sequence
          CAAAGGCCCCUCUCUGCGUGUUCACAGCGGACCUUGAUUUAAAUGUCCAUACAAUUAAGGCACGCGGUGAAUGCUAAGAACGAGGCUGGAAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038911
          miRBase ID: pbv-miR-124b-5p
          Mature miRNA Sequence: CGUGUUCACAGCGGACCUUGAU
          Chromosome Location: NA

          Rabbit

          Stem-loop ID ocu-mir-124-1
          Stem-loop Accession # MI0039276
          Stem-loop Sequence
          CGUGUUCACAGCGGACCUUGAUUUAAAUGUCCAUACAAUUAAGGCACGCGGUGAAUGCCAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0048203
          miRBase ID: ocu-miR-124-5p
          Mature miRNA Sequence: CGUGUUCACAGCGGACCUUGAU
          Chromosome Location: NA

          Stem-loop ID ocu-mir-124-2
          Stem-loop Accession # MI0039277
          Stem-loop Sequence
          CGUGUUCACAGCGGACCUUGAUUUAAUGUCAUACAAUUAAGGCACGCGGUGAAUGCCAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0048203
          miRBase ID: ocu-miR-124-5p
          Mature miRNA Sequence: CGUGUUCACAGCGGACCUUGAU
          Chromosome Location: NA

          Xenopus laevis

          Stem-loop ID xla-mir-124-1
          Stem-loop Accession # MI0038237
          Stem-loop Sequence
          CGUGUUCACAGCGGACCUUGAUUUAAAUGUCCAUACAAUUAAGGCACGCGGUGAAUGCCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0046430
          miRBase ID: xla-miR-124-5p
          Mature miRNA Sequence: CGUGUUCACAGCGGACCUUGAU
          Chromosome Location: NA

          Zebrafish

          Stem-loop ID dre-mir-124-2
          Stem-loop Accession # MI0001967
          Stem-loop Sequence
          CCUGCUUUUCUUCGUGUUCACAGCGGACCUUGAUUUAAAUGUCCAUACAAUUAAGGCACGCGGUGAAUGCCAAGAGAGAUGGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0031960
          miRBase ID: dre-miR-124-5p
          Mature miRNA Sequence: CGUGUUCACAGCGGACCUUGAU
          Chromosome Location: NA

          Stem-loop ID dre-mir-124-3
          Stem-loop Accession # MI0001968
          Stem-loop Sequence
          GGCUCUGUGGGAUUUCAGACUCUGGCUUUCCGUGUUCACAGCGGACCUUGAUUUAAUGUCUUACAAUUAAGGCACGCGGUGAAUGCCAAGAGCGGAGCCUUUUAACAUCAGCAGGCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0031960
          miRBase ID: dre-miR-124-5p
          Mature miRNA Sequence: CGUGUUCACAGCGGACCUUGAU
          Chromosome Location: NA

          Stem-loop ID dre-mir-124-5
          Stem-loop Accession # MI0001970
          Stem-loop Sequence
          GGGUUUUGCUCGUGCGUUCUUUUUGAGUUCUCGCUCUGCGUGUUCACAGCGGACCUUGAUUUAAUGUCCAUACAAUUAAGGCACGCGGUGAAUGCCAAGAGAAGAAUCUCUCCAGCAACGAGUUUGCGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0031960
          miRBase ID: dre-miR-124-5p
          Mature miRNA Sequence: CGUGUUCACAGCGGACCUUGAU
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Mm03306225_pri
          TaqMan™ Pri-miRNA Assay : Rn03465782_pri
          TaqMan™ Pri-miRNA Assay : Mm03307226_pri
          TaqMan™ Pri-miRNA Assay : Hs03303081_pri
          TaqMan™ Pri-miRNA Assay : Rn03464574_pri
          TaqMan™ Pri-miRNA Assay : Rn03464567_pri
          TaqMan™ Pri-miRNA Assay : Mm03307218_pri
          TaqMan™ Pri-miRNA Assay : Hs03303077_pri
          TaqMan™ Pri-miRNA Assay : Hs03303090_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM11154
          Ambion® Pre-miR™ miRNA Precursor : PM11154
          TaqMan™ MicroRNA Assay : 002197
          mirVana® miRNA inhibitor : MH11154
          mirVana® miRNA mimic : MC11154
          TaqMan™ Advanced miRNA Assay : rno480902_mir
          TaqMan™ Advanced miRNA Assay : 477880_mir

          Back To Top

          Related Products

          • TaqMan® Advanced miRNA cDNA Synthesis Kit
          • TaqMan® Fast Advanced Master Mix
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline