Search
Search

| Assay Name: | hsa-miR-589-5p |
|---|---|
| miRBase Accession Number: | MI0003599 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UGAGAACCACGUCUGCUCUGAG |
| Species: | Human, Callithrix jacchus, Pongo pygmaeus, Rhesus monkey |
| Product Type: | Ambion® Pre-miR™ miRNA Precursor |
Gene Family ID: | MIPF0000496, mir-589 |
Mature miRNA Sequence: | UGAGAACCACGUCUGCUCUGAG |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-589-5p | MIMAT0004799 |
| Callithrix jacchus | cja-miR-589 | MIMAT0039627 |
| Pongo pygmaeus | ppy-miR-589 | MIMAT0016031 |
| Rhesus monkey | mml-miR-589 | MIMAT0006449 |