Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › MC11162
          Assay Name: hsa-miR-589-5p
          miRBase Accession Number: MI0003599
          miRBase Version: v22.1
          Chromosome Location: Chr.7: 5495819 - 5495917 [-] on Build GRCh38
          Mature miRNA Sequence: UGAGAACCACGUCUGCUCUGAG
          Species: Human, Callithrix jacchus, Pongo pygmaeus, Rhesus monkey
          Product Type: mirVana® miRNA mimic
          Assay ID MC11162

          Purification
          Size
          Availability Made To Order
          Catalog # 4464066
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000496, mir-589

          Mature miRNA Sequence:

          UGAGAACCACGUCUGCUCUGAG

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-589-5p MIMAT0004799
          Callithrix jacchus cja-miR-589 MIMAT0039627
          Pongo pygmaeus ppy-miR-589 MIMAT0016031
          Rhesus monkey mml-miR-589 MIMAT0006449

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-589
          Stem-loop Accession # MI0003599
          Stem-loop Sequence
          UCCAGCCUGUGCCCAGCAGCCCCUGAGAACCACGUCUGCUCUGAGCUGGGUACUGCCUGUUCAGAACAAAUGCCGGUUCCCAGACGCUGCCAGCUGGCC
          Chromosome Location Chr. 7 - 5495819 - 5495917 [-] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0004799
          miRBase ID: hsa-miR-589-5p
          Mature miRNA Sequence: UGAGAACCACGUCUGCUCUGAG
          Chromosome Location: Chr. 7 - 5495819 - 5495917 [-] on Build GRCh38

          Callithrix jacchus

          Stem-loop ID cja-mir-589
          Stem-loop Accession # MI0032164
          Stem-loop Sequence
          GCAGCCGCUGAGAACCACGUCUGCUCUGAGCCGGGUACUGCCUGUUCAGAACAAACACCGCUUCCCAGACGCUGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0039627
          miRBase ID: cja-miR-589
          Mature miRNA Sequence: UGAGAACCACGUCUGCUCUGAG
          Chromosome Location: NA

          Pongo pygmaeus

          Stem-loop ID ppy-mir-589
          Stem-loop Accession # MI0015076
          Stem-loop Sequence
          UCCAGCCUGUGCUCAUCGGCCCCUGAGAACCACGUCUGCUCUGAGCUGGGUACUGCCUGUUCAGAACAAACGCUGGUUCCCAGACGCUGCCAGCUGGCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0016031
          miRBase ID: ppy-miR-589
          Mature miRNA Sequence: UGAGAACCACGUCUGCUCUGAG
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-589
          Stem-loop Accession # MI0007854
          Stem-loop Sequence
          UCCAGCCUGUGCCCAGCAGCCCCUGAGAACCACGUCUGCUCUGAGCUGGGUACUGCCUGUUCAGAACAGACGCUGCUUCCCAGACGCUGCCAGCUGGCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006449
          miRBase ID: mml-miR-589
          Mature miRNA Sequence: UGAGAACCACGUCUGCUCUGAG
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Hs03304476_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM11162
          Ambion® Pre-miR™ miRNA Precursor : PM11162
          TaqMan™ MicroRNA Assay : 002409
          mirVana® miRNA inhibitor : MH11162
          TaqMan™ Advanced miRNA Assay : 479073_mir


          miRBase Alias:

          Human: hsa-miR-589 (v17)

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline