Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › PM12067
          Assay Name: hsa-miR-675-5p
          miRBase Accession Number: MI0005416
          miRBase Version: v22.1
          Chromosome Location: Chr.11: 1996759 - 1996831 [-] on Build GRCh38
          Mature miRNA Sequence: UGGUGCGGAGAGGGCCCACAGUG
          Species: Human, Gorilla gorilla, Pongo pygmaeus, Rhesus monkey
          Product Type: Ambion® Pre-miR™ miRNA Precursor
          Assay ID PM12067
          Size
          Availability Made To Order
          Catalog # AM17100
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000365, mir-675

          Mature miRNA Sequence:

          UGGUGCGGAGAGGGCCCACAGUG

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-675-5p MIMAT0004284
          Gorilla gorilla ggo-miR-675b MIMAT0024305
          Pongo pygmaeus ppy-miR-675a MIMAT0016098
          Rhesus monkey mml-miR-675-5p MIMAT0006507

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-675
          Stem-loop Accession # MI0005416
          Stem-loop Sequence
          CCCAGGGUCUGGUGCGGAGAGGGCCCACAGUGGACUUGGUGACGCUGUAUGCCCUCACCGCUCAGCCCCUGGG
          Chromosome Location Chr. 11 - 1996759 - 1996831 [-] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0004284
          miRBase ID: hsa-miR-675-5p
          Mature miRNA Sequence: UGGUGCGGAGAGGGCCCACAGUG
          Chromosome Location: Chr. 11 - 1996759 - 1996831 [-] on Build GRCh38

          Gorilla gorilla

          Stem-loop ID ggo-mir-675b
          Stem-loop Accession # MI0020853
          Stem-loop Sequence
          AGGCACUGCGGCCCAGGGUCUGGUGCGGAGAGGGCCCACAGUGGACUUGGUGACGCUGUAUGCCCUCACCGCUCAGCCCCUGGGGCUGGCUUGGCAGACAGUACAGCAUCCAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0024305
          miRBase ID: ggo-miR-675b
          Mature miRNA Sequence: UGGUGCGGAGAGGGCCCACAGUG
          Chromosome Location: NA

          Pongo pygmaeus

          Stem-loop ID ppy-mir-675a
          Stem-loop Accession # MI0015138
          Stem-loop Sequence
          CCCAGGGUCUGGUGCGGAGAGGGCCCACAGUGGACUUGGUGACGCUGUAUGCCCUCACCGCUCAGCCCCUGGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0016098
          miRBase ID: ppy-miR-675a
          Mature miRNA Sequence: UGGUGCGGAGAGGGCCCACAGUG
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-675
          Stem-loop Accession # MI0007908
          Stem-loop Sequence
          CCCAGGGUCUGGUGCGGAGAGGGCCCACAGUGGACUUGGUGACACUGUAUGCCCUCACCGCUCAGCCCCUGGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006507
          miRBase ID: mml-miR-675-5p
          Mature miRNA Sequence: UGGUGCGGAGAGGGCCCACAGUG
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Hs03295696_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM12067
          TaqMan™ MicroRNA Assay : 002005
          mirVana® miRNA inhibitor : MH12067
          mirVana® miRNA mimic : MC12067
          TaqMan™ Advanced miRNA Assay : 478196_mir


          miRBase Alias:

          Human: hsa-miR-675 (v17)
          Macaca mulatta: mml-miR-675 (v19)

          Back To Top

          Related Products

          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline