Search
Search

| Assay Name: | hsa-miR-675-5p |
|---|---|
| miRBase Accession Number: | MI0005416 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UGGUGCGGAGAGGGCCCACAGUG |
| Species: | Human, Gorilla gorilla, Pongo pygmaeus, Rhesus monkey |
| Product Type: | mirVana® miRNA inhibitor |
Gene Family ID: | MIPF0000365, mir-675 |
Mature miRNA Sequence: | UGGUGCGGAGAGGGCCCACAGUG |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-675-5p | MIMAT0004284 |
| Gorilla gorilla | ggo-miR-675b | MIMAT0024305 |
| Pongo pygmaeus | ppy-miR-675a | MIMAT0016098 |
| Rhesus monkey | mml-miR-675-5p | MIMAT0006507 |