Search
Search

| Assay Name: | hsa-miR-99b-3p |
|---|---|
| miRBase Accession Number: | MI0000746 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | CAAGCUCGUGUCUGUGGGUCCG |
| Species: | Human, Mouse, Rat, Cricetulus griseus, Guinea pig |
| Product Type: | Ambion® Pre-miR™ miRNA Precursor |
Gene Family ID: | MIPF0000033, mir-10 |
Mature miRNA Sequence: | CAAGCUCGUGUCUGUGGGUCCG |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-99b-3p | MIMAT0004678 |
| Mouse | mmu-miR-99b-3p | MIMAT0004525 |
| Rat | rno-miR-99b-3p | MIMAT0004725 |
| Cricetulus griseus | cgr-miR-99b | MIMAT0041167 |
| Guinea pig | cpo-miR-99b-3p | MIMAT0046944 |