Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn03465992_pri
          Assay Name:
          rno-mir-99b
          Stem-loop ID:
          rno-mir-3596c rno-mir-99b
          Mapped to miRBase Version:
          v22.1
          Chromosome Location:
          Chr.1: 59704372 - 59704488 [-] on Build Rnor_6.0
          Stem-loop Location:
          Chr.1: 59704372 - 59704488 [-] on Build Rnor_6.0 Chr.1: 59704216 - 59704285 [+] on Build Rnor_6.0
          Mature miRNA ID:
          rno-miR-3596c rno-miR-99b-5p rno-miR-99b-3p
          Species:
          Rat
          Product Type:
          TaqMan™ Pri-miRNA Assay
          Assay ID Rn03465992_pri
          Size
          Availability Made To Order
          Catalog # 4427012
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Amplicon Length:

          62

          Chromosome Location:

          Chr. 1 - 59703963 - 59703963 [-] on 59703963 Build Rnor_6.0

          Species:

          Rat

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Rat rno-miR-3596c MIMAT0017877
          Rat rno-miR-99b-5p MIMAT0000821
          Rat rno-miR-99b-3p MIMAT0004725

          Stem-loop Details



          Rat

          Stem-loop ID rno-mir-3596c
          Stem-loop Accession # MI0015459
          Stem-loop Sequence
          CCCCUUCCAGUGCAGGGCGCAGCCUGGGGAAAGCUAGGAGGCCGUAUAGUGAUCUCCUCGGGUGUCUUCCUCAACUAUACAACCUCCUACCUCAGCCCGGGGGGCGCGGCAGGUGGG
          Chromosome Location Chr. 1 - 59704372 - 59704488 [-] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0017877
          miRBase ID: rno-miR-3596c
          Mature miRNA Sequence: ACUAUACAACCUCCUACCUCA
          Chromosome Location: Chr. 1 - 59704372 - 59704488 [-] on Build Rnor_6.0

          Gene Family ID MIPF0001194, mir-3596
          Distance to Amplicon 375 bases
          Stem-loop ID rno-mir-99b
          Stem-loop Accession # MI0000884
          Stem-loop Sequence
          GGCACCCACCCGUAGAACCGACCUUGCGGGGCCUUCGCCGCACACAAGCUCGUGUCUGUGGGUCCGUGUC
          Chromosome Location Chr. 1 - 59704216 - 59704285 [+] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0000821
          miRBase ID: rno-miR-99b-5p
          Mature miRNA Sequence: CACCCGUAGAACCGACCUUGCG
          Chromosome Location: Chr. 1 - 59704216 - 59704285 [+] on Build Rnor_6.0

          miRBase Accession #: MIMAT0004725
          miRBase ID: rno-miR-99b-3p
          Mature miRNA Sequence: CAAGCUCGUGUCUGUGGGUCCG
          Chromosome Location: Chr. 1 - 59704216 - 59704285 [+] on Build Rnor_6.0

          Gene Family ID MIPF0000033, mir-10
          Distance to Amplicon 219 bases

          Back To Top

          More Information




          Related Products

          Ambion® Anti-miR™ miRNA Inhibitor : AM11021
          Ambion® Pre-miR™ miRNA Precursor : PM11021
          TaqMan™ MicroRNA Assay : 000436
          mirVana® miRNA inhibitor : MH11021
          mirVana® miRNA mimic : MC11021
          TaqMan™ Advanced miRNA Assay : rno481284_mir
          TaqMan™ Advanced miRNA Assay : 478343_mir
          TaqMan™ Advanced miRNA Assay : mmu481284_mir
          Ambion® Anti-miR™ miRNA Inhibitor : AM12932
          Ambion® Pre-miR™ miRNA Precursor : PM12932
          TaqMan™ MicroRNA Assay : 002196
          mirVana® miRNA inhibitor : MH12932
          mirVana® miRNA mimic : MC12932
          TaqMan™ Advanced miRNA Assay : 478216_mir
          TaqMan™ Advanced miRNA Assay : rno481283_mir
          TaqMan™ Advanced miRNA Assay : mmu481283_mir
          Ambion® Anti-miR™ miRNA Inhibitor : AM19665
          Ambion® Pre-miR™ miRNA Precursor : PM19665
          TaqMan™ MicroRNA Assay : 464581_mat
          mirVana® miRNA inhibitor : MH19665
          mirVana® miRNA mimic : MC19665
          TaqMan™ Advanced miRNA Assay : rno480883_mir

          Back To Top

          Related Products

          • High Capacity RNA-to-cDNA Kit
          • SuperScript® IV VILO™ Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Fast Advanced Master Mix
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline