Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › 000436
          Assay Name: hsa-miR-99b
          miRBase Accession Number: MI0000746
          miRBase Version: v22.1
          Chromosome Location: Chr.19: 51692612 - 51692681 [+] on Build GRCh38
          Mature miRNA Sequence: CACCCGUAGAACCGACCUUGCG
          Species: Human, Mouse, Rat, Bovine, Dog, Goat, Guinea pig, Horse, Otolemur garnettii, Pan troglodytes, Pig, Pteropus alecto, Rhesus monkey, Tupaia chinensis
          Product Type: TaqMan™ MicroRNA Assay
          Assay ID 000436
          Size
          Availability Inventoried
          Catalog # 4427975
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000033, mir-10

          Mature miRNA Sequence:

          CACCCGUAGAACCGACCUUGCG

          Mature miRNA Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-99b-5p MIMAT0000689
          Mouse mmu-miR-99b-5p MIMAT0000132
          Rat rno-miR-99b-5p MIMAT0000821
          Bovine bta-miR-99b MIMAT0004345
          Dog cfa-miR-99b MIMAT0006607
          Goat chi-miR-99b-5p MIMAT0036314
          View More Rows
          Guinea pig cpo-miR-99b-5p MIMAT0046943
          Horse eca-miR-99b MIMAT0013018
          Otolemur garnettii oga-miR-99b MIMAT0049653
          Pan troglodytes ptr-miR-99b MIMAT0008351
          Pig ssc-miR-99b MIMAT0006018
          Pteropus alecto pal-miR-99b-5p MIMAT0039834
          Rhesus monkey mml-miR-99b-5p MIMAT0006179
          Tupaia chinensis tch-miR-99b-5p MIMAT0036582

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-99b
          Stem-loop Accession # MI0000746
          Stem-loop Sequence
          GGCACCCACCCGUAGAACCGACCUUGCGGGGCCUUCGCCGCACACAAGCUCGUGUCUGUGGGUCCGUGUC
          Chromosome Location Chr. 19 - 51692612 - 51692681 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0000689
          miRBase ID: hsa-miR-99b-5p
          Mature miRNA Sequence: CACCCGUAGAACCGACCUUGCG
          Chromosome Location: Chr. 19 - 51692612 - 51692681 [+] on Build GRCh38

          Mouse

          Stem-loop ID mmu-mir-99b
          Stem-loop Accession # MI0000147
          Stem-loop Sequence
          GGCACCCACCCGUAGAACCGACCUUGCGGGGCCUUCGCCGCACACAAGCUCGUGUCUGUGGGUCCGUGUC
          Chromosome Location Chr. 17 - 17830188 - 17830257 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0000132
          miRBase ID: mmu-miR-99b-5p
          Mature miRNA Sequence: CACCCGUAGAACCGACCUUGCG
          Chromosome Location: Chr. 17 - 17830188 - 17830257 [+] on Build GRCm38

          Rat

          Stem-loop ID rno-mir-99b
          Stem-loop Accession # MI0000884
          Stem-loop Sequence
          GGCACCCACCCGUAGAACCGACCUUGCGGGGCCUUCGCCGCACACAAGCUCGUGUCUGUGGGUCCGUGUC
          Chromosome Location Chr. 1 - 59704216 - 59704285 [+] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0000821
          miRBase ID: rno-miR-99b-5p
          Mature miRNA Sequence: CACCCGUAGAACCGACCUUGCG
          Chromosome Location: Chr. 1 - 59704216 - 59704285 [+] on Build Rnor_6.0

          Bovine

          Stem-loop ID bta-mir-99b
          Stem-loop Accession # MI0005469
          Stem-loop Sequence
          GGCACCCACCCGUAGAACCGACCUUGCGGGGCCUUCGCCGCACACAAGCUCGUGUCUGUGGGUCCGUGUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0004345
          miRBase ID: bta-miR-99b
          Mature miRNA Sequence: CACCCGUAGAACCGACCUUGCG
          Chromosome Location: NA

          Dog

          Stem-loop ID cfa-mir-99b
          Stem-loop Accession # MI0008003
          Stem-loop Sequence
          CACCCGUAGAACCGACCUUGCGGGGCCUUCGCCGCACACAAGCUCGAGUCUGUGGGUCUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006607
          miRBase ID: cfa-miR-99b
          Mature miRNA Sequence: CACCCGUAGAACCGACCUUGCG
          Chromosome Location: NA

          Goat

          Stem-loop ID chi-mir-99b
          Stem-loop Accession # MI0030846
          Stem-loop Sequence
          GGCACCCACCCGUAGAACCGACCUUGCGGGGCCUUCGCCGCACACAAGCUCGUGUCUGUGGGUCCGUGUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036314
          miRBase ID: chi-miR-99b-5p
          Mature miRNA Sequence: CACCCGUAGAACCGACCUUGCG
          Chromosome Location: NA

          Guinea pig

          Stem-loop ID cpo-mir-99b
          Stem-loop Accession # MI0038611
          Stem-loop Sequence
          CACCCGUAGAACCGACCUUGCGGGGCCUUCGCCACACACAAGCUCGUGUCUGUGGGUCCG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0046943
          miRBase ID: cpo-miR-99b-5p
          Mature miRNA Sequence: CACCCGUAGAACCGACCUUGCG
          Chromosome Location: NA

          Horse

          Stem-loop ID eca-mir-99b
          Stem-loop Accession # MI0012767
          Stem-loop Sequence
          GGCACCCACCCGUAGAACCGACCUUGCGGGGCCUUCGCCGCACACAAGCUCGUGUCUGUGGGUCCGUGUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0013018
          miRBase ID: eca-miR-99b
          Mature miRNA Sequence: CACCCGUAGAACCGACCUUGCG
          Chromosome Location: NA

          Otolemur garnettii

          Stem-loop ID oga-mir-99b
          Stem-loop Accession # MI0040436
          Stem-loop Sequence
          CACCCGUAGAACCGACCUUGCGGGGCCUUCGCCGCACACAAGCUCGUGUCUGUGGGUCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049653
          miRBase ID: oga-miR-99b
          Mature miRNA Sequence: CACCCGUAGAACCGACCUUGCG
          Chromosome Location: NA

          Pan troglodytes

          Stem-loop ID ptr-mir-99b
          Stem-loop Accession # MI0008891
          Stem-loop Sequence
          GCACCCACCCGUAGAACCGACCUUGCGGGGCCUUCGCCGCACACAAGCUCGUGUCUGUGGGUCCGUGUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0008351
          miRBase ID: ptr-miR-99b
          Mature miRNA Sequence: CACCCGUAGAACCGACCUUGCG
          Chromosome Location: NA

          Pig

          Stem-loop ID ssc-mir-99b
          Stem-loop Accession # MI0007077
          Stem-loop Sequence
          GGCACCCACCCGUAGAACCGACCUUGCGGGGCCUUCGCCGCACACAAGCUCGUGUCUGUGGGUCCGUGUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006018
          miRBase ID: ssc-miR-99b
          Mature miRNA Sequence: CACCCGUAGAACCGACCUUGCG
          Chromosome Location: NA

          Pteropus alecto

          Stem-loop ID pal-mir-99b
          Stem-loop Accession # MI0032353
          Stem-loop Sequence
          CACCCGUAGAACCGACCUUGCGGGGCCUUCGCCGCACACAAGCUCGUGUCUGUGGGUCCGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0039834
          miRBase ID: pal-miR-99b-5p
          Mature miRNA Sequence: CACCCGUAGAACCGACCUUGCG
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-99b
          Stem-loop Accession # MI0007609
          Stem-loop Sequence
          GGCACCCACCCGUAGAACCGACCUUGCGGGGCCUUCGCCGCACACAAGCUCGUGUCUGUGGGUCCGUGUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006179
          miRBase ID: mml-miR-99b-5p
          Mature miRNA Sequence: CACCCGUAGAACCGACCUUGCG
          Chromosome Location: NA

          Tupaia chinensis

          Stem-loop ID tch-mir-99b
          Stem-loop Accession # MI0031246
          Stem-loop Sequence
          CACCCGUAGAACCGACCUUGCGGGGCCUUCGCCGCACACAAGCUCGUGUCUGUGGGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036582
          miRBase ID: tch-miR-99b-5p
          Mature miRNA Sequence: CACCCGUAGAACCGACCUUGCG
          Chromosome Location: NA


          Back To Top

          More Information


          Associated Megaplex™ RT Primer Pools:

          Megaplex™ RT Primers, Rodent Pool A
          Megaplex™ RT Primers, Human Pool A




          Related Products

          TaqMan™ Pri-miRNA Assay : Mm03306774_pri
          TaqMan™ Pri-miRNA Assay : Rn03465992_pri
          TaqMan™ Pri-miRNA Assay : Hs03295173_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM11021
          Ambion® Pre-miR™ miRNA Precursor : PM11021
          mirVana® miRNA inhibitor : MH11021
          mirVana® miRNA mimic : MC11021
          TaqMan™ Advanced miRNA Assay : rno481284_mir
          TaqMan™ Advanced miRNA Assay : 478343_mir
          TaqMan™ Advanced miRNA Assay : mmu481284_mir
          MIRNA : TaqMan® Array Rodent MicroRNA A Card v2.0
          FGMIR : QS TaqMan OpenArray Rod miRNA Plate
          FGMIR : QS TaqMan OpenArray Hu miRNA Plate
          MIRNA : TaqMan® OpenArray Human MicroRNA Panel
          miRNA Card : TaqMan® Rodent MicroRNA A Cards v2.0
          miRNA Card : TaqMan® Human MicroRNA A Cards v2.0
          MIRNA : TaqMan® OpenArray Rodent MicroRNA Panel
          miRNA Pool : Megaplex™ RT Primers, Rodent Pool A
          MIRNA : TaqMan® Array Rodent MicroRNA A+B Cards Set v3.0
          FGMIR : TaqManTM Rodent MicroRNA Array A
          FGMIR : TaqManTM Human MicroRNA Array A
          MIRNA : TaqMan® Array Human MicroRNA A+B Cards Set v3.0
          MIRNA : TaqMan® Array Human MicroRNA A Cards v2.0
          miRNA Pool : Megaplex™ RT Primers, Human Pool A


          miRBase Alias:

          Human: hsa-miR-99b (v17)
          Mouse: mmu-miR-99b (v17)
          Rat: rno-miR-99b (v18)
          Macaca mulatta: mml-miR-99b (v19)

          Back To Top

          Related Products

          • TaqMan® Universal Master Mix II, with UNG
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® MicroRNA Reverse Transcription Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline