Search
Search

| Assay Name: | rno-miR-3588 |
|---|---|
| miRBase Accession Number: | MI0015466 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UCACAAGUUAGGGUCUCAGGGA |
| Species: | Rat, Branchiostoma belcheri, Branchiostoma floridae |
| Product Type: | Ambion® Pre-miR™ miRNA Precursor |
Gene Family ID: | MIPF0000033, mir-10 |
Mature miRNA Sequence: | UCACAAGUUAGGGUCUCAGGGA |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Rat | rno-miR-3588 | MIMAT0017887 |
| Branchiostoma belcheri | bbe-miR-125b-3p | MIMAT0031581 |
| Branchiostoma floridae | bfl-miR-125b | MIMAT0010007 |