Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › 007655_mat
          Assay Name: rno-miR-3588
          miRBase Accession Number: MI0015466
          miRBase Version: v22.1
          Chromosome Location: Chr.11: 16097337 - 16097442 [-] on Build Rnor_6.0
          Mature miRNA Sequence: UCACAAGUUAGGGUCUCAGGGA
          Species: Rat, Branchiostoma belcheri, Branchiostoma floridae
          Product Type: TaqMan™ MicroRNA Assay
          Assay ID 007655_mat
          Size
          Availability Made To Order
          Catalog # 4440886
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000033, mir-10

          Mature miRNA Sequence:

          UCACAAGUUAGGGUCUCAGGGA

          Mature miRNA Details:



          Species miRBase ID miRBase Accession Number
          Rat rno-miR-3588 MIMAT0017887
          Branchiostoma belcheri bbe-miR-125b-3p MIMAT0031581
          Branchiostoma floridae bfl-miR-125b MIMAT0010007

          Stem-loop Details



          Rat

          Stem-loop ID rno-mir-3588
          Stem-loop Accession # MI0015466
          Stem-loop Sequence
          CUGCUGGUACCUCUCCGCCUAGGUCCCAAGAGCCUGACUUGUGAUGUUACUAAAAUACCUCACAAGUUAGGGUCUCAGGGACUAGGAAAAGUCUGGUUUGAUGCAG
          Chromosome Location Chr. 11 - 16097337 - 16097442 [-] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0017887
          miRBase ID: rno-miR-3588
          Mature miRNA Sequence: UCACAAGUUAGGGUCUCAGGGA
          Chromosome Location: Chr. 11 - 16097337 - 16097442 [-] on Build Rnor_6.0

          Branchiostoma belcheri

          Stem-loop ID bbe-mir-125b
          Stem-loop Accession # MI0026178
          Stem-loop Sequence
          GUGGAGGGCGUAUGUCACCCAGCUCCCAAGAUCCUAACCUGUGAGCCAUGGACAUCACAAGUUAGGGUCUCAGGGAAUGGGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0031581
          miRBase ID: bbe-miR-125b-3p
          Mature miRNA Sequence: UCACAAGUUAGGGUCUCAGGGA
          Chromosome Location: NA

          Branchiostoma floridae

          Stem-loop ID bfl-mir-125b
          Stem-loop Accession # MI0010518
          Stem-loop Sequence
          GUGGAGGGCGUAUGUCACCCAGCUCCCAAGAUCCUAACCUGUGAGCCAUGGACAUCACAAGUUAGGGUCUCAGGGAAUGGGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0010007
          miRBase ID: bfl-miR-125b
          Mature miRNA Sequence: UCACAAGUUAGGGUCUCAGGGA
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Rn03465042_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM15414
          Ambion® Pre-miR™ miRNA Precursor : PM15414
          mirVana® miRNA inhibitor : MH15414
          mirVana® miRNA mimic : MC15414
          TaqMan™ Advanced miRNA Assay : rno481119_mir

          Back To Top

          Related Products

          • TaqMan® Universal Master Mix II, with UNG
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® MicroRNA Reverse Transcription Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline