Search
Search

| Assay Name: | mmu-miR-221-5p |
|---|---|
| miRBase Accession Number: | MI0000709 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | ACCUGGCAUACAAUGUAGAUUUCUGU |
| Species: | Mouse, Cricetulus griseus, Dasypus novemcinctus, Guinea pig, Pig, Rabbit, Rhesus monkey, Taeniopygia guttata, Zebrafish |
| Product Type: | Ambion® Pre-miR™ miRNA Precursor |
Gene Family ID: | MIPF0000051, mir-221 |
Mature miRNA Sequence: | ACCUGGCAUACAAUGUAGAUUUCUGU |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-221-5p | MIMAT0017060 |
| Cricetulus griseus | cgr-miR-221-5p | MIMAT0023858 |
| Dasypus novemcinctus | dno-miR-221-5p | MIMAT0047775 |
| Guinea pig | cpo-miR-221-5p | MIMAT0047130 |
| Pig | ssc-miR-221-5p | MIMAT0022949 |
| Rabbit | ocu-miR-221-5p | MIMAT0048354 |
| View More Rows | ||
| Rhesus monkey | mml-miR-221-5p | MIMAT0026590 |
| Taeniopygia guttata | tgu-miR-221-5p | MIMAT0026976 |
| Zebrafish | dre-miR-221-5p | MIMAT0031926 |