Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › MC19413
          Assay Name: mmu-miR-221-5p
          miRBase Accession Number: MI0000709
          miRBase Version: v22.1
          Chromosome Location: Chr.X: 19146294 - 19146388 [-] on Build GRCm38
          Mature miRNA Sequence: ACCUGGCAUACAAUGUAGAUUUCUGU
          Species: Mouse, Cricetulus griseus, Dasypus novemcinctus, Guinea pig, Pig, Rabbit, Rhesus monkey, Taeniopygia guttata, Zebrafish
          Product Type: mirVana® miRNA mimic
          Assay ID MC19413

          Purification
          Size
          Availability Made To Order
          Catalog # 4464066
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000051, mir-221

          Mature miRNA Sequence:

          ACCUGGCAUACAAUGUAGAUUUCUGU

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Mouse mmu-miR-221-5p MIMAT0017060
          Cricetulus griseus cgr-miR-221-5p MIMAT0023858
          Dasypus novemcinctus dno-miR-221-5p MIMAT0047775
          Guinea pig cpo-miR-221-5p MIMAT0047130
          Pig ssc-miR-221-5p MIMAT0022949
          Rabbit ocu-miR-221-5p MIMAT0048354
          View More Rows
          Rhesus monkey mml-miR-221-5p MIMAT0026590
          Taeniopygia guttata tgu-miR-221-5p MIMAT0026976
          Zebrafish dre-miR-221-5p MIMAT0031926

          Stem-loop Details



          Mouse

          Stem-loop ID mmu-mir-221
          Stem-loop Accession # MI0000709
          Stem-loop Sequence
          AUCCAGGUCUGGGGCAUGAACCUGGCAUACAAUGUAGAUUUCUGUGUUUGUUAGGCAACAGCUACAUUGUCUGCUGGGUUUCAGGCUACCUGGAA
          Chromosome Location Chr. X - 19146294 - 19146388 [-] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0017060
          miRBase ID: mmu-miR-221-5p
          Mature miRNA Sequence: ACCUGGCAUACAAUGUAGAUUUCUGU
          Chromosome Location: Chr. X - 19146294 - 19146388 [-] on Build GRCm38

          Cricetulus griseus

          Stem-loop ID cgr-mir-221
          Stem-loop Accession # MI0020464
          Stem-loop Sequence
          GGGGCAUGAACCUGGCAUACAAUGUAGAUUUCUGUGUUUAUUAGGCAACAGCUACAUUGUCUGCUGGGUUUCAGGCUACCUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0023858
          miRBase ID: cgr-miR-221-5p
          Mature miRNA Sequence: ACCUGGCAUACAAUGUAGAUUUCUGU
          Chromosome Location: NA

          Dasypus novemcinctus

          Stem-loop ID dno-mir-221
          Stem-loop Accession # MI0039052
          Stem-loop Sequence
          ACCUGGCAUACAAUGUAGAUUUCUGUGUUUGUUGGACAACAGCUACAUUGUCUGCUGGGUUUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047775
          miRBase ID: dno-miR-221-5p
          Mature miRNA Sequence: ACCUGGCAUACAAUGUAGAUUUCUGU
          Chromosome Location: NA

          Guinea pig

          Stem-loop ID cpo-mir-221
          Stem-loop Accession # MI0038713
          Stem-loop Sequence
          ACCUGGCAUACAAUGUAGAUUUCUGUGUUUGUUGAACAACAGCUACAUUGUCUGCUGGGUUUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047130
          miRBase ID: cpo-miR-221-5p
          Mature miRNA Sequence: ACCUGGCAUACAAUGUAGAUUUCUGU
          Chromosome Location: NA

          Pig

          Stem-loop ID ssc-mir-221
          Stem-loop Accession # MI0008221
          Stem-loop Sequence
          UGAACCUGGCAUACAAUGUAGAUUUCUGUGUUUGUUGGGCAACAGCUACAUUGUCUGCUGGGUUUCAGGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0022949
          miRBase ID: ssc-miR-221-5p
          Mature miRNA Sequence: ACCUGGCAUACAAUGUAGAUUUCUGU
          Chromosome Location: NA

          Rabbit

          Stem-loop ID ocu-mir-221
          Stem-loop Accession # MI0039357
          Stem-loop Sequence
          ACCUGGCAUACAAUGUAGAUUUCUGUGUUUUUUGGGCAACAGCUACAUUGUCUGCUGGGUUUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0048354
          miRBase ID: ocu-miR-221-5p
          Mature miRNA Sequence: ACCUGGCAUACAAUGUAGAUUUCUGU
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-221
          Stem-loop Accession # MI0002892
          Stem-loop Sequence
          UGAACAUCCAGGUCUGGGGCAUGAACCUGGCAUACAAUGUAGAUUUCUGUGUUCGUUAGGCAACAGCUACAUUGUCUGCUGGGUUUCAGGCUACCUGGAAACAUGUUCUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0026590
          miRBase ID: mml-miR-221-5p
          Mature miRNA Sequence: ACCUGGCAUACAAUGUAGAUUUCUGU
          Chromosome Location: NA

          Taeniopygia guttata

          Stem-loop ID tgu-mir-221
          Stem-loop Accession # MI0013710
          Stem-loop Sequence
          ACCUGGCAUACAAUGUAGAUUUCUGUGUUUGUUAAGCAACAGCUACAUUGUCUGCUGGGUUUCCAGCUGCCUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0026976
          miRBase ID: tgu-miR-221-5p
          Mature miRNA Sequence: ACCUGGCAUACAAUGUAGAUUUCUGU
          Chromosome Location: NA

          Zebrafish

          Stem-loop ID dre-mir-221
          Stem-loop Accession # MI0001387
          Stem-loop Sequence
          GUCGUGAACCUGGCAUACAAUGUAGAUUUCUGUGUGGUACUAUCUACAGCUACAUUGUCUGCUGGGUUUCAGGCCAGCAGAAUAAUUCUGCUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0031926
          miRBase ID: dre-miR-221-5p
          Mature miRNA Sequence: ACCUGGCAUACAAUGUAGAUUUCUGU
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Mm03307181_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM19413
          Ambion® Pre-miR™ miRNA Precursor : PM19413
          TaqMan™ MicroRNA Assay : 464489_mat
          mirVana® miRNA inhibitor : MH19413
          TaqMan™ Advanced miRNA Assay : mmu481659_mir


          miRBase Alias:

          Mouse: mmu-miR-221* (v17)

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline