Search
Search

| Assay Name: | mmu-miR-155-3p |
|---|---|
| miRBase Accession Number: | MI0000177 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | CUCCUACCUGUUAGCAUUAAC |
| Species: | Mouse, Rat |
| Product Type: | Ambion® Pre-miR™ miRNA Precursor |
Gene Family ID: | MIPF0000157, mir-155 |
Mature miRNA Sequence: | CUCCUACCUGUUAGCAUUAAC |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-155-3p | MIMAT0016993 |
| Rat | rno-miR-155-3p | MIMAT0030410 |