Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Mm03306395_pri
Assay Name:
mmu-mir-155
Stem-loop ID:
mmu-mir-155
Mapped to miRBase Version:
v22.1
Chromosome Location:
Chr.16: 84714140 - 84714204 [+] on Build GRCm38
Stem-loop Location:
Chr.16: 84714140 - 84714204 [+] on Build GRCm38
Mature miRNA ID:
mmu-miR-155-3p
mmu-miR-155-5p
Species:
Mouse
Product Type:
TaqMan
™
Pri-miRNA Assay
Assay ID
Mm03306395_pri
Size
S: 360 rxns; 20X
M: 750 rxns; 20X
L: 2900 rxns; 60X
Availability
Made To Order
Catalog #
4427012
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Amplicon Length:
85
Chromosome Location:
Chr. 16 - 84714532 - 84714532 [-] on 84714532 Build GRCm38
Species:
Mouse
miRBase Details:
Species
miRBase ID
miRBase Accession Number
Mouse
mmu-miR-155-3p
MIMAT0016993
Mouse
mmu-miR-155-5p
MIMAT0000165
Stem-loop Details
Mouse
Stem-loop ID
mmu-mir-155
Stem-loop Accession #
MI0000177
Stem-loop Sequence
CUGUUAAUGCUAAUUGUGAUAGGGGUUUUGGCCUCUGACUGACUCCUACCUGUUAGCAUUAACAG
Chromosome Location
Chr. 16 - 84714140 - 84714204 [+] on Build GRCm38
Mature MicroRNA
miRBase Accession #: MIMAT0016993
miRBase ID: mmu-miR-155-3p
Mature miRNA Sequence: CUCCUACCUGUUAGCAUUAAC
Chromosome Location: Chr. 16 - 84714140 - 84714204 [+] on Build GRCm38
miRBase Accession #: MIMAT0000165
miRBase ID: mmu-miR-155-5p
Mature miRNA Sequence: UUAAUGCUAAUUGUGAUAGGGGU
Chromosome Location: Chr. 16 - 84714140 - 84714204 [+] on Build GRCm38
Gene Family ID
MIPF0000157,
mir-155
Distance to Amplicon
292 bases
Back To Top
More Information
Related Products
Ambion
®
Anti-miR
™
miRNA Inhibitor :
AM19607
Ambion
®
Pre-miR
™
miRNA Precursor :
PM19607
TaqMan
™
MicroRNA Assay :
464539_mat
mir
Vana
®
miRNA inhibitor :
MH19607
mir
Vana
®
miRNA mimic :
MC19607
TaqMan
™
Advanced miRNA Assay :
mmu481328_mir
TaqMan
™
Advanced miRNA Assay :
rno481328_mir
Ambion
®
Anti-miR
™
miRNA Inhibitor :
AM13058
Ambion
®
Pre-miR
™
miRNA Precursor :
PM13058
TaqMan
™
MicroRNA Assay :
002571
mir
Vana
®
miRNA inhibitor :
MH13058
mir
Vana
®
miRNA mimic :
MC13058
TaqMan
™
Advanced miRNA Assay :
mmu480953_mir
TaqMan
™
Advanced miRNA Assay :
rno480953_mir
Back To Top
Related Products
High Capacity RNA-to-cDNA Kit
SuperScript
®
IV VILO
™
Master Mix
TaqMan
®
Universal PCR Master Mix
TaqMan
®
Fast Advanced Master Mix
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.46.0-Offline