Search
Search

| Assay Name: | hsa-miR-152-5p |
|---|---|
| miRBase Accession Number: | MI0000462 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | AGGUUCUGUGAUACACUCCGACU |
| Species: | Human, Rat, Cricetulus griseus, Monodelphis domestica, Pteropus alecto, Rabbit, Rhesus monkey |
| Product Type: | Ambion® Pre-miR™ miRNA Precursor |
Gene Family ID: | MIPF0000056, mir-148 |
Mature miRNA Sequence: | AGGUUCUGUGAUACACUCCGACU |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-152-5p | MIMAT0026479 |
| Rat | rno-miR-152-5p | MIMAT0017134 |
| Cricetulus griseus | cgr-miR-152-5p | MIMAT0023784 |
| Monodelphis domestica | mdo-miR-152-5p | MIMAT0026671 |
| Pteropus alecto | pal-miR-152-5p | MIMAT0040106 |
| Rabbit | ocu-miR-152-5p | MIMAT0048267 |
| View More Rows | ||
| Rhesus monkey | mml-miR-152-5p | MIMAT0026829 |