Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › MC19680
          Assay Name: hsa-miR-152-5p
          miRBase Accession Number: MI0000462
          miRBase Version: v22.1
          Chromosome Location: Chr.17: 48037161 - 48037247 [-] on Build GRCh38
          Mature miRNA Sequence: AGGUUCUGUGAUACACUCCGACU
          Species: Human, Rat, Cricetulus griseus, Monodelphis domestica, Pteropus alecto, Rabbit, Rhesus monkey
          Product Type: mirVana® miRNA mimic
          Assay ID MC19680

          Purification
          Size
          Availability Made To Order
          Catalog # 4464066
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000056, mir-148

          Mature miRNA Sequence:

          AGGUUCUGUGAUACACUCCGACU

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-152-5p MIMAT0026479
          Rat rno-miR-152-5p MIMAT0017134
          Cricetulus griseus cgr-miR-152-5p MIMAT0023784
          Monodelphis domestica mdo-miR-152-5p MIMAT0026671
          Pteropus alecto pal-miR-152-5p MIMAT0040106
          Rabbit ocu-miR-152-5p MIMAT0048267
          View More Rows
          Rhesus monkey mml-miR-152-5p MIMAT0026829

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-152
          Stem-loop Accession # MI0000462
          Stem-loop Sequence
          UGUCCCCCCCGGCCCAGGUUCUGUGAUACACUCCGACUCGGGCUCUGGAGCAGUCAGUGCAUGACAGAACUUGGGCCCGGAAGGACC
          Chromosome Location Chr. 17 - 48037161 - 48037247 [-] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0026479
          miRBase ID: hsa-miR-152-5p
          Mature miRNA Sequence: AGGUUCUGUGAUACACUCCGACU
          Chromosome Location: Chr. 17 - 48037161 - 48037247 [-] on Build GRCh38

          Rat

          Stem-loop ID rno-mir-152
          Stem-loop Accession # MI0000921
          Stem-loop Sequence
          UGUUCCCCGGGCCCAGGUUCUGUGAUACACUCCGACUCGGGCUCUGGAGCAGUCAGUGCAUGACAGAACUUGGGCCCGGUAGGAC
          Chromosome Location Chr. 10 - 84719319 - 84719403 [+] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0017134
          miRBase ID: rno-miR-152-5p
          Mature miRNA Sequence: AGGUUCUGUGAUACACUCCGACU
          Chromosome Location: Chr. 10 - 84719319 - 84719403 [+] on Build Rnor_6.0

          Cricetulus griseus

          Stem-loop ID cgr-mir-152
          Stem-loop Accession # MI0020413
          Stem-loop Sequence
          CCCGGGCCCAGGUUCUGUGAUACACUCCGACUCGGGCUCUGGAGCAGUCAGUGCAUGACAGAACUUGGGCCCGGUAGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0023784
          miRBase ID: cgr-miR-152-5p
          Mature miRNA Sequence: AGGUUCUGUGAUACACUCCGACU
          Chromosome Location: NA

          Monodelphis domestica

          Stem-loop ID mdo-mir-152
          Stem-loop Accession # MI0005306
          Stem-loop Sequence
          CCCGGCCCAGGUUCUGUGAUACACUCCGACUUAGACUCUGGAGCAGUCAGUGCAUGACAGAACUUGGGCUUGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0026671
          miRBase ID: mdo-miR-152-5p
          Mature miRNA Sequence: AGGUUCUGUGAUACACUCCGACU
          Chromosome Location: NA

          Pteropus alecto

          Stem-loop ID pal-mir-152
          Stem-loop Accession # MI0032497
          Stem-loop Sequence
          AGGUUCUGUGAUACACUCCGACUCGAGCUCUGGAGCAGUCAGUGCAUGACAGAACUUGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0040106
          miRBase ID: pal-miR-152-5p
          Mature miRNA Sequence: AGGUUCUGUGAUACACUCCGACU
          Chromosome Location: NA

          Rabbit

          Stem-loop ID ocu-mir-152
          Stem-loop Accession # MI0039311
          Stem-loop Sequence
          AGGUUCUGUGAUACACUCCGACUCGGGCUCUGGAGCAGUCAGUGCAUGACAGAACUUGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0048267
          miRBase ID: ocu-miR-152-5p
          Mature miRNA Sequence: AGGUUCUGUGAUACACUCCGACU
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-152
          Stem-loop Accession # MI0007643
          Stem-loop Sequence
          UGUCCCCCCCGGCCCAGGUUCUGUGAUACACUCCGACUCGGGCUCUGGAGCAGUCAGUGCAUGACAGAACUUGGGCCCGGAAGGACC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0026829
          miRBase ID: mml-miR-152-5p
          Mature miRNA Sequence: AGGUUCUGUGAUACACUCCGACU
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Hs03303177_pri
          TaqMan™ Pri-miRNA Assay : Rn03465082_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM19680
          Ambion® Pre-miR™ miRNA Precursor : PM19680
          TaqMan™ MicroRNA Assay : 464242_mat
          mirVana® miRNA inhibitor : MH19680
          TaqMan™ Advanced miRNA Assay : rno480949_mir
          TaqMan™ Advanced miRNA Assay : 478722_mir


          miRBase Alias:

          Rat: rno-miR-152* (v18)

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline