Search
Search

| Assay Name: | hsa-miR-128-1-5p |
|---|---|
| miRBase Accession Number: | MI0000447 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | CGGGGCCGUAGCACUGUCUGAGA |
| Species: | Human, Cricetulus griseus, Dasypus novemcinctus, Goat, Guinea pig, Monodelphis domestica, Pteropus alecto, Rabbit, Rhesus monkey |
| Product Type: | Ambion® Pre-miR™ miRNA Precursor |
Gene Family ID: | MIPF0000048, mir-128 |
Mature miRNA Sequence: | CGGGGCCGUAGCACUGUCUGAGA |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-128-1-5p | MIMAT0026477 |
| Cricetulus griseus | cgr-miR-128-5p | MIMAT0023749 |
| Dasypus novemcinctus | dno-miR-128-5p | MIMAT0047621 |
| Goat | chi-miR-128-5p | MIMAT0035933 |
| Guinea pig | cpo-miR-128-5p | MIMAT0046974 |
| Monodelphis domestica | mdo-miR-128b-5p | MIMAT0028608 |
| View More Rows | ||
| Pteropus alecto | pal-miR-128-5p | MIMAT0039793 |
| Rabbit | ocu-miR-128a-5p | MIMAT0048210 |
| Rhesus monkey | mml-miR-128a-5p | MIMAT0026565 |