Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › AM24586
          Assay Name: hsa-miR-128-1-5p
          miRBase Accession Number: MI0000447
          miRBase Version: v22.1
          Chromosome Location: Chr.2: 135665397 - 135665478 [+] on Build GRCh38
          Mature miRNA Sequence: CGGGGCCGUAGCACUGUCUGAGA
          Species: Human, Cricetulus griseus, Dasypus novemcinctus, Goat, Guinea pig, Monodelphis domestica, Pteropus alecto, Rabbit, Rhesus monkey
          Product Type: Ambion® Anti-miR™ miRNA Inhibitor
          Assay ID AM24586
          Size
          Availability Made To Order
          Catalog # AM17000
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000048, mir-128

          Mature miRNA Sequence:

          CGGGGCCGUAGCACUGUCUGAGA

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-128-1-5p MIMAT0026477
          Cricetulus griseus cgr-miR-128-5p MIMAT0023749
          Dasypus novemcinctus dno-miR-128-5p MIMAT0047621
          Goat chi-miR-128-5p MIMAT0035933
          Guinea pig cpo-miR-128-5p MIMAT0046974
          Monodelphis domestica mdo-miR-128b-5p MIMAT0028608
          View More Rows
          Pteropus alecto pal-miR-128-5p MIMAT0039793
          Rabbit ocu-miR-128a-5p MIMAT0048210
          Rhesus monkey mml-miR-128a-5p MIMAT0026565

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-128-1
          Stem-loop Accession # MI0000447
          Stem-loop Sequence
          UGAGCUGUUGGAUUCGGGGCCGUAGCACUGUCUGAGAGGUUUACAUUUCUCACAGUGAACCGGUCUCUUUUUCAGCUGCUUC
          Chromosome Location Chr. 2 - 135665397 - 135665478 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0026477
          miRBase ID: hsa-miR-128-1-5p
          Mature miRNA Sequence: CGGGGCCGUAGCACUGUCUGAGA
          Chromosome Location: Chr. 2 - 135665397 - 135665478 [+] on Build GRCh38

          Cricetulus griseus

          Stem-loop ID cgr-mir-128
          Stem-loop Accession # MI0020391
          Stem-loop Sequence
          UGUUGGAUUCGGGGCCGUAGCACUGUCUGAGAGGUGUAUAUUUCUCACAGUGAACCGGUCUCUUUUUCAGCUGCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0023749
          miRBase ID: cgr-miR-128-5p
          Mature miRNA Sequence: CGGGGCCGUAGCACUGUCUGAGA
          Chromosome Location: NA

          Dasypus novemcinctus

          Stem-loop ID dno-mir-128-1
          Stem-loop Accession # MI0038970
          Stem-loop Sequence
          CGGGGCCGUAGCACUGUCUGAGAGGUUUACAUUUCUCACAGUGAACCGGUCUCUUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047621
          miRBase ID: dno-miR-128-5p
          Mature miRNA Sequence: CGGGGCCGUAGCACUGUCUGAGA
          Chromosome Location: NA

          Goat

          Stem-loop ID chi-mir-128
          Stem-loop Accession # MI0030610
          Stem-loop Sequence
          UGUUUAAUAUUUUGCAAUAAUUGGCCUUGUUCCUGAGCUGUUGGAUUCGGGGCCGUAGCACUGUCUGAGAGGUUUACAUUUCUCACAGUGAACCGGUCUCUUUUUCAGCUGCUUCCUGGCUUCUUUUUACUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0035933
          miRBase ID: chi-miR-128-5p
          Mature miRNA Sequence: CGGGGCCGUAGCACUGUCUGAGA
          Chromosome Location: NA

          Guinea pig

          Stem-loop ID cpo-mir-128-1
          Stem-loop Accession # MI0038629
          Stem-loop Sequence
          CGGGGCCGUAGCACUGUCUGAGAGGUUUACAUUUCUCACAGUGAACCGGUCUCUUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0046974
          miRBase ID: cpo-miR-128-5p
          Mature miRNA Sequence: CGGGGCCGUAGCACUGUCUGAGA
          Chromosome Location: NA

          Monodelphis domestica

          Stem-loop ID mdo-mir-128b
          Stem-loop Accession # MI0023827
          Stem-loop Sequence
          CGGGGCCGUAGCACUGUCUGAGAGGUUUACAUUUCUCACAGUGAACCGGUCUCUUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0028608
          miRBase ID: mdo-miR-128b-5p
          Mature miRNA Sequence: CGGGGCCGUAGCACUGUCUGAGA
          Chromosome Location: NA

          Pteropus alecto

          Stem-loop ID pal-mir-128
          Stem-loop Accession # MI0032332
          Stem-loop Sequence
          CGGGGCCGUAGCACUGUCUGAGAGGUUUACAUUUCUCACAGUGAACCGGUCUCUUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0039793
          miRBase ID: pal-miR-128-5p
          Mature miRNA Sequence: CGGGGCCGUAGCACUGUCUGAGA
          Chromosome Location: NA

          Rabbit

          Stem-loop ID ocu-mir-128a
          Stem-loop Accession # MI0039281
          Stem-loop Sequence
          CGGGGCCGUAGCACUGUCUGAGAGGUUUACAUUUCUCACAGUGAACCGGUCUCUUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0048210
          miRBase ID: ocu-miR-128a-5p
          Mature miRNA Sequence: CGGGGCCGUAGCACUGUCUGAGA
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-128a
          Stem-loop Accession # MI0002518
          Stem-loop Sequence
          UGAGCUGUUGGAUUCGGGGCCGUAGCACUGUCUGAGAGGUUUACAUUUCUCACAGUGAACCGGUCUCUUUUUCAGCUGCUUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0026565
          miRBase ID: mml-miR-128a-5p
          Mature miRNA Sequence: CGGGGCCGUAGCACUGUCUGAGA
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Hs03303101_pri
          Ambion® Pre-miR™ miRNA Precursor : PM24586
          TaqMan™ MicroRNA Assay : 466976_mat
          mirVana® miRNA inhibitor : MH24586
          mirVana® miRNA mimic : MC24586
          TaqMan™ Advanced miRNA Assay : 478682_mir

          Back To Top

          Related Products

          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline