Search
Search

| Assay Name: | hsa-miR-6133 |
|---|---|
| miRBase Accession Number: | MI0021278 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UGAGGGAGGAGGUUGGGUA |
| Species: | Human, Pan troglodytes, Rhesus monkey |
| Product Type: | Ambion® Pre-miR™ miRNA Precursor |
Gene Family ID: | MIPF0001415, mir-6133 |
Mature miRNA Sequence: | UGAGGGAGGAGGUUGGGUA |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-6133 | MIMAT0024617 |
| Pan troglodytes | ptr-miR-6133 | MIMAT0024607 |
| Rhesus monkey | mml-miR-6133 | MIMAT0024626 |