Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › 471214_mat
          Assay Name: hsa-miR-6133
          miRBase Accession Number: MI0021278
          miRBase Version: v22.1
          Chromosome Location: Chr.7: 133290881 - 133290988 [+] on Build GRCh38
          Mature miRNA Sequence: UGAGGGAGGAGGUUGGGUA
          Species: Human, Pan troglodytes, Rhesus monkey
          Product Type: TaqMan™ MicroRNA Assay
          Assay ID 471214_mat
          Size
          Availability Made To Order
          Catalog # 4440886
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0001415, mir-6133

          Mature miRNA Sequence:

          UGAGGGAGGAGGUUGGGUA

          Mature miRNA Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-6133 MIMAT0024617
          Pan troglodytes ptr-miR-6133 MIMAT0024607
          Rhesus monkey mml-miR-6133 MIMAT0024626

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-6133
          Stem-loop Accession # MI0021278
          Stem-loop Sequence
          GGAAUGUCACCUGUGUGUUUUCUCUGCAUGCCCUCUUCAUUGUUCUGCUGAAGACUGGUCUCUUCAUGUGUGAGGGAGGAGGUUGGGUAUUGAGGGAAAACAGGGGGC
          Chromosome Location Chr. 7 - 133290881 - 133290988 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0024617
          miRBase ID: hsa-miR-6133
          Mature miRNA Sequence: UGAGGGAGGAGGUUGGGUA
          Chromosome Location: Chr. 7 - 133290881 - 133290988 [+] on Build GRCh38

          Pan troglodytes

          Stem-loop ID ptr-mir-6133
          Stem-loop Accession # MI0021268
          Stem-loop Sequence
          GGAAUGUCACCUGUGUGUUUUCUCUGCAUGCCCUCUUCAUUGUUCUGCUGAAGACUGGUCUCUUCAUGUGUGAGGGAGGAGGUUGGGUAUUGAGGGAAAACAGGGGGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0024607
          miRBase ID: ptr-miR-6133
          Mature miRNA Sequence: UGAGGGAGGAGGUUGGGUA
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-6133
          Stem-loop Accession # MI0021287
          Stem-loop Sequence
          GGAAUGUCACCUGUGUGUUUUCUCUGCAUACCCUCUUCAUUGUUCUACUGAAGACUGGUCUCUUUAUGUGUGAGGGAGGAGGUUGGGUAUUGAGAGAAAACAGGGGGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0024626
          miRBase ID: mml-miR-6133
          Mature miRNA Sequence: UGAGGGAGGAGGUUGGGUA
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Hs04349640_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM25769
          Ambion® Pre-miR™ miRNA Precursor : PM25769
          mirVana® miRNA inhibitor : MH25769
          mirVana® miRNA mimic : MC25769
          TaqMan™ Advanced miRNA Assay : 480188_mir

          Back To Top

          Related Products

          • TaqMan® Universal Master Mix II, with UNG
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® MicroRNA Reverse Transcription Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline