Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › mmu480908_mir

          Specificity testing was performed using mouse anti-targets.

          Assay Name: mmu-miR-126a-5p
          Stem-loop Accession Number: MI0000471
          miRBase Version: v22.1
          Chromosome Location: Chr.9: 136670602 - 136670686 [+] on Build GRCh38
          Mature miRNA Sequence: CAUUAUUACUUUUGGUACGCG
          Species: Human, Mouse, Rat, Alligator mississippiensis, Anolis carolinensis, Astatotilapia burtoni, Bovine, Chicken, Chrysemys picta, Cyprinus carpio, Dog, Gadus morhua, Horse, Ictalurus punctatus, Metriaclima zebra, Microcebus murinus, Monodelphis domestica, Neolamprologus brichardi, Nomascus leucogenys, Oreochromis niloticus, Ornithorhynchus anatinus, Oryzias latipes, Pig, Pundamilia nyererei, Python bivittatus, Salmo salar, Xenopus laevis, Xenopus tropicalis, Zebrafish
          Product Type: TaqMan™ Advanced miRNA Assay
          Assay ID mmu480908_mir
          Size S: 250 rxns
          Availability Inventoried
          Catalog # A25576
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000115, mir-126

          Mature miRNA Sequence:

          CAUUAUUACUUUUGGUACGCG

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-126-5p MIMAT0000444
          Mouse mmu-miR-126a-5p MIMAT0000137
          Rat rno-miR-126a-5p MIMAT0000831
          Alligator mississippiensis ami-miR-126-5p MIMAT0038131
          Anolis carolinensis aca-miR-126-5p MIMAT0021730
          Astatotilapia burtoni abu-miR-126-5p MIMAT0042065
          View More Rows
          Bovine bta-miR-126-5p MIMAT0004328
          Chicken gga-miR-126-5p MIMAT0003723
          Chrysemys picta cpi-miR-126-5p MIMAT0037733
          Cyprinus carpio ccr-miR-126-5p MIMAT0026206
          Dog cfa-miR-126 MIMAT0006730
          Gadus morhua gmo-miR-126-5p MIMAT0044216
          Horse eca-miR-126-5p MIMAT0013176
          Ictalurus punctatus ipu-miR-126b MIMAT0029401
          Metriaclima zebra mze-miR-126 MIMAT0042303
          Microcebus murinus mmr-miR-126 MIMAT0049139
          Monodelphis domestica mdo-miR-126-5p MIMAT0012743
          Neolamprologus brichardi nbr-miR-126 MIMAT0042489
          Nomascus leucogenys nle-miR-126 MIMAT0049342
          Oreochromis niloticus oni-miR-126 MIMAT0042673
          Ornithorhynchus anatinus oan-miR-126-5p MIMAT0007125
          Oryzias latipes ola-miR-126-5p MIMAT0022602
          Pig ssc-miR-126-5p MIMAT0018377
          Pundamilia nyererei pny-miR-126 MIMAT0042879
          Python bivittatus pbv-miR-126-5p MIMAT0038918
          Salmo salar ssa-miR-126-5p MIMAT0032311
          Xenopus laevis xla-miR-126-5p MIMAT0046435
          Xenopus tropicalis xtr-miR-126-5p MIMAT0003587
          Zebrafish dre-miR-126a-5p MIMAT0003157
          Zebrafish dre-miR-126b-5p MIMAT0011307

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-126
          Stem-loop Accession # MI0000471
          Stem-loop Sequence
          CGCUGGCGACGGGACAUUAUUACUUUUGGUACGCGCUGUGACACUUCAAACUCGUACCGUGAGUAAUAAUGCGCCGUCCACGGCA
          Chromosome Location Chr. 9 - 136670602 - 136670686 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0000444
          miRBase ID: hsa-miR-126-5p
          Mature miRNA Sequence: CAUUAUUACUUUUGGUACGCG
          Chromosome Location: Chr. 9 - 136670602 - 136670686 [+] on Build GRCh38

          Mouse

          Stem-loop ID mmu-mir-126a
          Stem-loop Accession # MI0000153
          Stem-loop Sequence
          UGACAGCACAUUAUUACUUUUGGUACGCGCUGUGACACUUCAAACUCGUACCGUGAGUAAUAAUGCGCGGUCA
          Chromosome Location Chr. 2 - 26591357 - 26591429 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0000137
          miRBase ID: mmu-miR-126a-5p
          Mature miRNA Sequence: CAUUAUUACUUUUGGUACGCG
          Chromosome Location: Chr. 2 - 26591357 - 26591429 [+] on Build GRCm38

          Rat

          Stem-loop ID rno-mir-126a
          Stem-loop Accession # MI0000898
          Stem-loop Sequence
          UGACAGCACAUUAUUACUUUUGGUACGCGCUGUGACACUUCAAACUCGUACCGUGAGUAAUAAUGCGUGGUCA
          Chromosome Location Chr. 3 - 4042488 - 4042560 [+] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0000831
          miRBase ID: rno-miR-126a-5p
          Mature miRNA Sequence: CAUUAUUACUUUUGGUACGCG
          Chromosome Location: Chr. 3 - 4042488 - 4042560 [+] on Build Rnor_6.0

          Alligator mississippiensis

          Stem-loop ID ami-mir-126
          Stem-loop Accession # MI0029724
          Stem-loop Sequence
          CGGCCCAUUAUUACUUUUGGUACGCGCUGUGACACUUCAAACUCGUACCGUGAGUAAUAAUGCGCUGCGGCCAGCACC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038131
          miRBase ID: ami-miR-126-5p
          Mature miRNA Sequence: CAUUAUUACUUUUGGUACGCG
          Chromosome Location: NA

          Anolis carolinensis

          Stem-loop ID aca-mir-126
          Stem-loop Accession # MI0018727
          Stem-loop Sequence
          CGCUCCCCGCGGUCCAUUAUUACUUUUGGUACGCGCUGUGCCACUUCAAACUCGUACCGUGAGUAAUAAUGCGCUGAGGGACAGCACCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0021730
          miRBase ID: aca-miR-126-5p
          Mature miRNA Sequence: CAUUAUUACUUUUGGUACGCG
          Chromosome Location: NA

          Astatotilapia burtoni

          Stem-loop ID abu-mir-126
          Stem-loop Accession # MI0033760
          Stem-loop Sequence
          CAUUAUUACUUUUGGUACGCGCUAUGCCACUCUCAACUCGUACCGUGAGUAAUAAUGCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0042065
          miRBase ID: abu-miR-126-5p
          Mature miRNA Sequence: CAUUAUUACUUUUGGUACGCG
          Chromosome Location: NA

          Bovine

          Stem-loop ID bta-mir-126
          Stem-loop Accession # MI0004754
          Stem-loop Sequence
          UGACGGGACAUUAUUACUUUUGGUACGCGCUGUGACACUUCAAACUCGUACCGUGAGUAAUAAUGCGCUGUCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0004328
          miRBase ID: bta-miR-126-5p
          Mature miRNA Sequence: CAUUAUUACUUUUGGUACGCG
          Chromosome Location: NA

          Chicken

          Stem-loop ID gga-mir-126
          Stem-loop Accession # MI0001244
          Stem-loop Sequence
          GCUGGUGACGGCCCAUUAUUACUUUUGGUACGCGCUGUGACACUUCAAACUCGUACCGUGAGUAAUAAUGCGCUGUGGUCAGCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0003723
          miRBase ID: gga-miR-126-5p
          Mature miRNA Sequence: CAUUAUUACUUUUGGUACGCG
          Chromosome Location: NA

          Chrysemys picta

          Stem-loop ID cpi-mir-126
          Stem-loop Accession # MI0029462
          Stem-loop Sequence
          GGCGGCGACGCUGGUGCCGGCCCAUUAUUACUUUUGGUACGCGCUGUGACACUUCAAACUCGUACCGUGAGUAAUAAUGCGCUGCGGCCAGCCCUGGCGAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0037733
          miRBase ID: cpi-miR-126-5p
          Mature miRNA Sequence: CAUUAUUACUUUUGGUACGCG
          Chromosome Location: NA

          Cyprinus carpio

          Stem-loop ID ccr-mir-126
          Stem-loop Accession # MI0023314
          Stem-loop Sequence
          UUAUCUGCUUCACAGUCCAUUAUUACUUUUGGUACGCGCUAGGCCAGACUCAAACUCGUACCGUGAGUAAUAAUGCACUGUGGCAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0026206
          miRBase ID: ccr-miR-126-5p
          Mature miRNA Sequence: CAUUAUUACUUUUGGUACGCG
          Chromosome Location: NA

          Dog

          Stem-loop ID cfa-mir-126
          Stem-loop Accession # MI0008154
          Stem-loop Sequence
          CAUUAUUACUUUUGGUACGCGCUGUGACACUUCAAACUCGUACCGUGAGUAAUAAUGCG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006730
          miRBase ID: cfa-miR-126
          Mature miRNA Sequence: CAUUAUUACUUUUGGUACGCG
          Chromosome Location: NA

          Gadus morhua

          Stem-loop ID gmo-mir-126
          Stem-loop Accession # MI0036133
          Stem-loop Sequence
          CAUUAUUACUUUUGGUACGCGCUAUGCCACACUCAACUCGUACCGUGAGUAAUAAUGCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0044216
          miRBase ID: gmo-miR-126-5p
          Mature miRNA Sequence: CAUUAUUACUUUUGGUACGCG
          Chromosome Location: NA

          Horse

          Stem-loop ID eca-mir-126
          Stem-loop Accession # MI0012919
          Stem-loop Sequence
          CGACGGGACAUUAUUACUUUUGGUACGCGCUGUGACACUUCAAACUCGUACCGUGAGUAAUAAUGCGCCGUCG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0013176
          miRBase ID: eca-miR-126-5p
          Mature miRNA Sequence: CAUUAUUACUUUUGGUACGCG
          Chromosome Location: NA

          Ictalurus punctatus

          Stem-loop ID ipu-mir-126b
          Stem-loop Accession # MI0024482
          Stem-loop Sequence
          CAUUAUUACUUUUGGUACGCGCUAUGCCACUCUCAACUCGUACCGUGAGUAAUAGUGCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0029401
          miRBase ID: ipu-miR-126b
          Mature miRNA Sequence: CAUUAUUACUUUUGGUACGCG
          Chromosome Location: NA

          Metriaclima zebra

          Stem-loop ID mze-mir-126
          Stem-loop Accession # MI0034063
          Stem-loop Sequence
          CAUUAUUACUUUUGGUACGCGCUAUGCCACUCUCAACUCGUACCGUGAGUAAUAAUGCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0042303
          miRBase ID: mze-miR-126
          Mature miRNA Sequence: CAUUAUUACUUUUGGUACGCG
          Chromosome Location: NA

          Microcebus murinus

          Stem-loop ID mmr-mir-126
          Stem-loop Accession # MI0039852
          Stem-loop Sequence
          CAUUAUUACUUUUGGUACGCGCUGUGACACUUCAAACUCGUACCGUGAGUAAUAAUGCG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049139
          miRBase ID: mmr-miR-126
          Mature miRNA Sequence: CAUUAUUACUUUUGGUACGCG
          Chromosome Location: NA

          Monodelphis domestica

          Stem-loop ID mdo-mir-126
          Stem-loop Accession # MI0012499
          Stem-loop Sequence
          GGGUCAUUAUUACUUUUGGUACGCGCUGUGACACAUCAAACUCGUACCGUGAGUAAUAAUGCGCCAUUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0012743
          miRBase ID: mdo-miR-126-5p
          Mature miRNA Sequence: CAUUAUUACUUUUGGUACGCG
          Chromosome Location: NA

          Neolamprologus brichardi

          Stem-loop ID nbr-mir-126
          Stem-loop Accession # MI0034319
          Stem-loop Sequence
          CAUUAUUACUUUUGGUACGCGCUAUGCCACUCUCAACUCGUACCGUGAGUAAUAAUGCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0042489
          miRBase ID: nbr-miR-126
          Mature miRNA Sequence: CAUUAUUACUUUUGGUACGCG
          Chromosome Location: NA

          Nomascus leucogenys

          Stem-loop ID nle-mir-126
          Stem-loop Accession # MI0040078
          Stem-loop Sequence
          CAUUAUUACUUUUGGUACGCGCUGUGACACUUCAAACUCGUACCGUGAGUAAUAAUGCG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049342
          miRBase ID: nle-miR-126
          Mature miRNA Sequence: CAUUAUUACUUUUGGUACGCG
          Chromosome Location: NA

          Oreochromis niloticus

          Stem-loop ID oni-mir-126
          Stem-loop Accession # MI0034574
          Stem-loop Sequence
          CAUUAUUACUUUUGGUACGCGCUAUGCCACUCUCAACUCGUACCGUGAGUAAUAAUGCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0042673
          miRBase ID: oni-miR-126
          Mature miRNA Sequence: CAUUAUUACUUUUGGUACGCG
          Chromosome Location: NA

          Ornithorhynchus anatinus

          Stem-loop ID oan-mir-126
          Stem-loop Accession # MI0006861
          Stem-loop Sequence
          GAAGCAUCAGUGACAGCUGGCAACUGCCCAUUAUUACUUUUGGUACGCGCUGUGACGCUUCAAACUCGUACCGUGAGUAAUAAUGCGUGGCUGGCAGCACUGGCAGCGCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0007125
          miRBase ID: oan-miR-126-5p
          Mature miRNA Sequence: CAUUAUUACUUUUGGUACGCG
          Chromosome Location: NA

          Oryzias latipes

          Stem-loop ID ola-mir-126
          Stem-loop Accession # MI0019483
          Stem-loop Sequence
          CGCAACAUGGCCAGCUUUGCGGUCCAUUAUUACUUUUGGUACGCGCUAUGCCACUCUCAACUCGUACCGUGAGUAAUAAUGCACUGCGACUGGUGCCGACUCCG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0022602
          miRBase ID: ola-miR-126-5p
          Mature miRNA Sequence: CAUUAUUACUUUUGGUACGCG
          Chromosome Location: NA

          Pig

          Stem-loop ID ssc-mir-126
          Stem-loop Accession # MI0016619
          Stem-loop Sequence
          CGACGGGACAUUAUUACUUUUGGUACGCGCUGUGACACUUCAAACUCGUACCGUGAGUAAUAAUGCGCUGUCG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0018377
          miRBase ID: ssc-miR-126-5p
          Mature miRNA Sequence: CAUUAUUACUUUUGGUACGCG
          Chromosome Location: NA

          Pundamilia nyererei

          Stem-loop ID pny-mir-126
          Stem-loop Accession # MI0034837
          Stem-loop Sequence
          CAUUAUUACUUUUGGUACGCGCUAUGCCACUCUCAACUCGUACCGUGAGUAAUAAUGCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0042879
          miRBase ID: pny-miR-126
          Mature miRNA Sequence: CAUUAUUACUUUUGGUACGCG
          Chromosome Location: NA

          Python bivittatus

          Stem-loop ID pbv-mir-126
          Stem-loop Accession # MI0030218
          Stem-loop Sequence
          GCGGCUCAUUAUUACUUUUGGUACGCGCUGUGCCACUUCAAACUCGUACCGUGAGUAAUAAUGCGCGGAGGGCCAGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038918
          miRBase ID: pbv-miR-126-5p
          Mature miRNA Sequence: CAUUAUUACUUUUGGUACGCG
          Chromosome Location: NA

          Salmo salar

          Stem-loop ID ssa-mir-126-1
          Stem-loop Accession # MI0026457
          Stem-loop Sequence
          CAUUAUUACUUUUGGUACGCGCUAGGCCAGUCUCAAACUCGUACCGUGAGUAAUAAUGCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0032311
          miRBase ID: ssa-miR-126-5p
          Mature miRNA Sequence: CAUUAUUACUUUUGGUACGCG
          Chromosome Location: NA

          Stem-loop ID ssa-mir-126-2
          Stem-loop Accession # MI0026458
          Stem-loop Sequence
          CAUUAUUACUUUUGGUACGCGCUAUGCCAAACUCUACUCGUACCGUGAGUAAUAAUGCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0032311
          miRBase ID: ssa-miR-126-5p
          Mature miRNA Sequence: CAUUAUUACUUUUGGUACGCG
          Chromosome Location: NA

          Stem-loop ID ssa-mir-126-3
          Stem-loop Accession # MI0026459
          Stem-loop Sequence
          CAUUAUUACUUUUGGUACGCGCUAUGCCAAACUCAACUCGUACCGUGAGUAAUAAUGCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0032311
          miRBase ID: ssa-miR-126-5p
          Mature miRNA Sequence: CAUUAUUACUUUUGGUACGCG
          Chromosome Location: NA

          Stem-loop ID ssa-mir-126-4
          Stem-loop Accession # MI0026460
          Stem-loop Sequence
          CAUUAUUACUUUUGGUACGCGCUCGGCCAGUCUCAAACUCGUACCGUGAGUAAUAAUGCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0032311
          miRBase ID: ssa-miR-126-5p
          Mature miRNA Sequence: CAUUAUUACUUUUGGUACGCG
          Chromosome Location: NA

          Xenopus laevis

          Stem-loop ID xla-mir-126-1
          Stem-loop Accession # MI0038240
          Stem-loop Sequence
          CAUUAUUACUUUUGGUACGCGCUGUGUCACAUCAAACUCGUACCGUGAGUAAUAAUGCGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0046435
          miRBase ID: xla-miR-126-5p
          Mature miRNA Sequence: CAUUAUUACUUUUGGUACGCG
          Chromosome Location: NA

          Stem-loop ID xla-mir-126-2
          Stem-loop Accession # MI0038241
          Stem-loop Sequence
          CAUUAUUACUUUUGGUACGCGCUGUGUCACAUCAAACUCGUACCGUGAGUAAUAAUGCG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0046435
          miRBase ID: xla-miR-126-5p
          Mature miRNA Sequence: CAUUAUUACUUUUGGUACGCG
          Chromosome Location: NA

          Xenopus tropicalis

          Stem-loop ID xtr-mir-126
          Stem-loop Accession # MI0004827
          Stem-loop Sequence
          GGCUGUGCAUUAUUACUUUUGGUACGCGCUGUGUCACAUCAAACUCGUACCGUGAGUAAUAAUGCGCAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0003587
          miRBase ID: xtr-miR-126-5p
          Mature miRNA Sequence: CAUUAUUACUUUUGGUACGCG
          Chromosome Location: NA

          Zebrafish

          Stem-loop ID dre-mir-126a
          Stem-loop Accession # MI0001979
          Stem-loop Sequence
          GAGCCAUUUUAACUGCUUCACAGUCCAUUAUUACUUUUGGUACGCGCUAGGCCAGACUCAAACUCGUACCGUGAGUAAUAAUGCACUGUGGCAGUGGGUUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0003157
          miRBase ID: dre-miR-126a-5p
          Mature miRNA Sequence: CAUUAUUACUUUUGGUACGCG
          Chromosome Location: NA

          Stem-loop ID dre-mir-126b
          Stem-loop Accession # MI0010848
          Stem-loop Sequence
          GAGUCUUACCGGCCUCACGGUUCAUUAUUACUUUUGGUACGCGCUAUGCCACUCUCAACUCGUACCGUGAGUAAUAGUGCACUGUGACUGGUGAACACAACUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0011307
          miRBase ID: dre-miR-126b-5p
          Mature miRNA Sequence: CAUUAUUACUUUUGGUACGCG
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Hs03303230_pri
          TaqMan™ Pri-miRNA Assay : Mm03306244_pri
          TaqMan™ Pri-miRNA Assay : Rn03464585_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM10401
          Ambion® Pre-miR™ miRNA Precursor : PM10401
          TaqMan™ MicroRNA Assay : 000451
          mirVana® miRNA inhibitor : MH10401
          mirVana® miRNA mimic : MC10401
          TaqMan™ Advanced miRNA Assay : rno480908_mir
          TaqMan™ Advanced miRNA Assay : 477888_mir

          Back To Top

          Related Products

          • TaqMan® Advanced miRNA cDNA Synthesis Kit
          • TaqMan® Fast Advanced Master Mix
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-6wvps:80/100.66.75.98:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0