Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › mmu480936_mir

          Specificity testing was performed using mouse anti-targets.

          Assay Name: mmu-miR-143-5p
          Stem-loop Accession Number: MI0000257
          miRBase Version: v22.1
          Chromosome Location: Chr.18: 61649196 - 61649258 [-] on Build GRCm38
          Mature miRNA Sequence: GGUGCAGUGCUGCAUCUCUGG
          Species: Mouse, Rat, Alligator mississippiensis, Anolis carolinensis, Chicken, Chrysemys picta, Columba livia, Dasypus novemcinctus, Goat, Guinea pig, Pig, Rabbit
          Product Type: TaqMan™ Advanced miRNA Assay
          Assay ID mmu480936_mir
          Size S: 250 rxns
          Availability Inventoried
          Catalog # A25576
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000094, mir-143

          Mature miRNA Sequence:

          GGUGCAGUGCUGCAUCUCUGG

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Mouse mmu-miR-143-5p MIMAT0017006
          Rat rno-miR-143-5p MIMAT0017129
          Alligator mississippiensis ami-miR-143-5p MIMAT0038165
          Anolis carolinensis aca-miR-143-5p MIMAT0021769
          Chicken gga-miR-143-5p MIMAT0048645
          Chrysemys picta cpi-miR-143-5p MIMAT0037768
          View More Rows
          Columba livia cli-miR-143-5p MIMAT0038531
          Dasypus novemcinctus dno-miR-143-5p MIMAT0047659
          Goat chi-miR-143-5p MIMAT0035963
          Guinea pig cpo-miR-143-5p MIMAT0047010
          Pig ssc-miR-143-5p MIMAT0017374
          Rabbit ocu-miR-143-5p MIMAT0048247

          Stem-loop Details



          Mouse

          Stem-loop ID mmu-mir-143
          Stem-loop Accession # MI0000257
          Stem-loop Sequence
          CCUGAGGUGCAGUGCUGCAUCUCUGGUCAGUUGGGAGUCUGAGAUGAAGCACUGUAGCUCAGG
          Chromosome Location Chr. 18 - 61649196 - 61649258 [-] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0017006
          miRBase ID: mmu-miR-143-5p
          Mature miRNA Sequence: GGUGCAGUGCUGCAUCUCUGG
          Chromosome Location: Chr. 18 - 61649196 - 61649258 [-] on Build GRCm38

          Rat

          Stem-loop ID rno-mir-143
          Stem-loop Accession # MI0000916
          Stem-loop Sequence
          GCGGAGCGCCUGUCUCCCAGCCUGAGGUGCAGUGCUGCAUCUCUGGUCAGUUGGGAGUCUGAGAUGAAGCACUGUAGCUCAGGAAGGGAGAAGAUGUUCUGCAGC
          Chromosome Location Chr. 18 - 56971273 - 56971377 [-] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0017129
          miRBase ID: rno-miR-143-5p
          Mature miRNA Sequence: GGUGCAGUGCUGCAUCUCUGG
          Chromosome Location: Chr. 18 - 56971273 - 56971377 [-] on Build Rnor_6.0

          Alligator mississippiensis

          Stem-loop ID ami-mir-143
          Stem-loop Accession # MI0029748
          Stem-loop Sequence
          AUGUCUCCCAGCCCAAGGUGCAGUGCUGCAUCUCUGGUCAGUUGUGAGUCUGAGAUGAAGCACUGUAGCUCGGGAAGGGAGGAAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038165
          miRBase ID: ami-miR-143-5p
          Mature miRNA Sequence: GGUGCAGUGCUGCAUCUCUGG
          Chromosome Location: NA

          Anolis carolinensis

          Stem-loop ID aca-mir-143
          Stem-loop Accession # MI0018753
          Stem-loop Sequence
          AUGUCUCCCAGCCCAAGGUGCAGUGCUGCAUCUCUGGUCAGUUGUGAGUCUGAGAUGAAGCACUGUAGCUCGGGAAGGGAGGAAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0021769
          miRBase ID: aca-miR-143-5p
          Mature miRNA Sequence: GGUGCAGUGCUGCAUCUCUGG
          Chromosome Location: NA

          Chicken

          Stem-loop ID gga-mir-143
          Stem-loop Accession # MI0039505
          Stem-loop Sequence
          UCUCCCAGCCCAAGGUGCAGUGCUGCAUCUCUGGUCAAUUGGGAGUCUGAGAUGAAGCACUGUAGCUCGGGAAGGGAGGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0048645
          miRBase ID: gga-miR-143-5p
          Mature miRNA Sequence: GGUGCAGUGCUGCAUCUCUGG
          Chromosome Location: NA

          Chrysemys picta

          Stem-loop ID cpi-mir-143
          Stem-loop Accession # MI0029486
          Stem-loop Sequence
          AGCUCCGUCAAUGUCUCCCAGCCCAAGGUGCAGUGCUGCAUCUCUGGUCAGUUGUGAGUCUGAGAUGAAGCACUGUAGCUCGGGAAGGGAGGAACUGUGCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0037768
          miRBase ID: cpi-miR-143-5p
          Mature miRNA Sequence: GGUGCAGUGCUGCAUCUCUGG
          Chromosome Location: NA

          Columba livia

          Stem-loop ID cli-mir-143
          Stem-loop Accession # MI0029978
          Stem-loop Sequence
          AUGUCUCCCAGCCCAAGGUGCAGUGCUGCAUCUCUGGUCAAUUGGGAGUCUGAGAUGAAGCACUGUAGCUCGGGAAGGGAGGAACUGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038531
          miRBase ID: cli-miR-143-5p
          Mature miRNA Sequence: GGUGCAGUGCUGCAUCUCUGG
          Chromosome Location: NA

          Dasypus novemcinctus

          Stem-loop ID dno-mir-143
          Stem-loop Accession # MI0038992
          Stem-loop Sequence
          GGUGCAGUGCUGCAUCUCUGGUCAGUUGGGAGUCUGAGAUGAAGCACUGUAGCUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047659
          miRBase ID: dno-miR-143-5p
          Mature miRNA Sequence: GGUGCAGUGCUGCAUCUCUGG
          Chromosome Location: NA

          Goat

          Stem-loop ID chi-mir-143
          Stem-loop Accession # MI0030629
          Stem-loop Sequence
          GCGUCCUGUCUCCCAGCCUGAGGUGCAGUGCUGCAUCUCUGGUCAGUUGGGAGUCUGAGAUGAAGCACUGUAGCUCGGGAAGGGAGAAGUUGUUCUACAGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0035963
          miRBase ID: chi-miR-143-5p
          Mature miRNA Sequence: GGUGCAGUGCUGCAUCUCUGG
          Chromosome Location: NA

          Guinea pig

          Stem-loop ID cpo-mir-143
          Stem-loop Accession # MI0038650
          Stem-loop Sequence
          GGUGCAGUGCUGCAUCUCUGGUCAGUUGGGAGUCUGAGAUGAAGCACUGUAGCUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047010
          miRBase ID: cpo-miR-143-5p
          Mature miRNA Sequence: GGUGCAGUGCUGCAUCUCUGG
          Chromosome Location: NA

          Pig

          Stem-loop ID ssc-mir-143
          Stem-loop Accession # MI0013098
          Stem-loop Sequence
          GUCCCCCAGCCGGAGGUGCAGUGCUGCAUCUCUGGUCAGCUGGGAGUCUGAGAUGAAGCACUGUAGCUCGGGAAGGGAGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0017374
          miRBase ID: ssc-miR-143-5p
          Mature miRNA Sequence: GGUGCAGUGCUGCAUCUCUGG
          Chromosome Location: NA

          Rabbit

          Stem-loop ID ocu-mir-143
          Stem-loop Accession # MI0039301
          Stem-loop Sequence
          GGUGCAGUGCUGCAUCUCUGGUCAGUUGGGAGUCUGAGAUGAAGCACUGUAGCUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0048247
          miRBase ID: ocu-miR-143-5p
          Mature miRNA Sequence: GGUGCAGUGCUGCAUCUCUGG
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Rn03466026_pri
          TaqMan™ Pri-miRNA Assay : Mm03306564_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM19855
          Ambion® Pre-miR™ miRNA Precursor : PM19855
          TaqMan™ MicroRNA Assay : 463509_mat
          mirVana® miRNA inhibitor : MH19855
          mirVana® miRNA mimic : MC19855
          TaqMan™ Advanced miRNA Assay : rno480936_mir

          Back To Top

          Related Products

          • TaqMan® Advanced miRNA cDNA Synthesis Kit
          • TaqMan® Fast Advanced Master Mix
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline